Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94061611-94062146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX24
Entrez Gene ID
57062
Gene Name
DEAD-box helicase 24
Gene Aliases
-
UniGene
Hs.510328
Gene Chromosome Location
Chr.14: 94050922-94081212 on Build GRCh38
Transcripts
NM_020414.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1371751 c.1216C>T p.H406Y Chr.14: 94062124-94062124 on Build GRCh38
959113 c.1200C>T p.A400A Chr.14: 94062140-94062140 on Build GRCh38
2253009 c.1225G>A p.A409T Chr.14: 94062115-94062115 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs4900211 Chr.14: 94061800-94061800 on Build GRCh38
rs757327757 Chr.14: 94062107-94062107 on Build GRCh38
rs1036584422 Chr.14: 94061719-94061719 on Build GRCh38
rs925670935 Chr.14: 94061967-94061967 on Build GRCh38
rs567409436 Chr.14: 94061951-94061951 on Build GRCh38
rs969168013 Chr.14: 94061851-94061851 on Build GRCh38
rs372748772 Chr.14: 94062050-94062050 on Build GRCh38
rs114338222 Chr.14: 94062091-94062091 on Build GRCh38
rs554037367 Chr.14: 94061637-94061637 on Build GRCh38
rs949944802 Chr.14: 94061697-94061697 on Build GRCh38
rs1009103221 Chr.14: 94061743-94061743 on Build GRCh38
rs993354943 Chr.14: 94061894-94061894 on Build GRCh38
rs535035036 Chr.14: 94061975-94061975 on Build GRCh38
rs533033997 Chr.14: 94061644-94061644 on Build GRCh38
rs369471355 Chr.14: 94062118-94062118 on Build GRCh38
rs563460983 Chr.14: 94061746-94061746 on Build GRCh38
rs191052369 Chr.14: 94061816-94061816 on Build GRCh38
rs1021533729 Chr.14: 94061891-94061891 on Build GRCh38
rs1049098079 Chr.14: 94061775-94061775 on Build GRCh38
rs778040395 Chr.14: 94062089-94062089 on Build GRCh38
rs932107847 Chr.14: 94061968-94061968 on Build GRCh38
rs927332641 Chr.14: 94061877-94061877 on Build GRCh38
rs546014043 Chr.14: 94062065-94062065 on Build GRCh38
rs1050614019 Chr.14: 94061987-94061987 on Build GRCh38
rs1036094561 Chr.14: 94062011-94062011 on Build GRCh38
rs980639603 Chr.14: 94061868-94061868 on Build GRCh38
rs936954223 Chr.14: 94061646-94061646 on Build GRCh38
rs552217576 Chr.14: 94061855-94061855 on Build GRCh38
rs151163065 Chr.14: 94061692-94061692 on Build GRCh38
rs765849965 Chr.14: 94062047-94062047 on Build GRCh38
rs995321053 Chr.14: 94061767-94061767 on Build GRCh38
rs746011166 Chr.14: 94061949-94061949 on Build GRCh38
rs1055741477 Chr.14: 94061651-94061651 on Build GRCh38
rs1040351277 Chr.14: 94062125-94062125 on Build GRCh38
rs572263949 Chr.14: 94061671-94061671 on Build GRCh38
rs770395801 Chr.14: 94061997-94061997 on Build GRCh38
rs781086913 Chr.14: 94061815-94061815 on Build GRCh38
rs913220981 Chr.14: 94061910-94061910 on Build GRCh38
rs375989603 Chr.14: 94062057-94062057 on Build GRCh38
rs376682033 Chr.14: 94061992-94061992 on Build GRCh38
rs978921247 Chr.14: 94061961-94061961 on Build GRCh38
rs996364831 Chr.14: 94061710-94061710 on Build GRCh38
rs763397266 Chr.14: 94062124-94062124 on Build GRCh38
rs771731463 Chr.14: 94061725-94061726 on Build GRCh38
rs201125907 Chr.14: 94061786-94061786 on Build GRCh38
rs778828360 Chr.14: 94062123-94062123 on Build GRCh38
rs946054150 Chr.14: 94061917-94061917 on Build GRCh38
rs968209966 Chr.14: 94061907-94061907 on Build GRCh38
rs140307008 Chr.14: 94062016-94062016 on Build GRCh38
rs1036230180 Chr.14: 94062022-94062022 on Build GRCh38
rs565285910 Chr.14: 94061908-94061911 on Build GRCh38
rs576394673 Chr.14: 94061838-94061841 on Build GRCh38
rs1008634639 Chr.14: 94061993-94061993 on Build GRCh38
rs1009777768 Chr.14: 94061874-94061874 on Build GRCh38
rs919089874 Chr.14: 94062043-94062043 on Build GRCh38
rs966682102 Chr.14: 94061848-94061848 on Build GRCh38
rs781092939 Chr.14: 94062114-94062114 on Build GRCh38
rs946270236 Chr.14: 94061658-94061658 on Build GRCh38
rs557174725 Chr.14: 94062117-94062117 on Build GRCh38
rs930454054 Chr.14: 94062045-94062045 on Build GRCh38
rs890219058 Chr.14: 94061742-94061742 on Build GRCh38
rs1043306899 Chr.14: 94061937-94061938 on Build GRCh38
rs897947627 Chr.14: 94061763-94061763 on Build GRCh38
rs367697223 Chr.14: 94062087-94062087 on Build GRCh38

View Full Product Details