Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95090098-95090604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DICER1
Entrez Gene ID
23405
Gene Name
dicer 1, ribonuclease III
Gene Aliases
DCR1, Dicer, Dicer1e, HERNA, K12H4.8-LIKE, MNG1, RMSE2
UniGene
Hs.738957
Gene Chromosome Location
Chr.14: 95086228-95158010 on Build GRCh38
Transcripts
NM_001195573.1 NM_001271282.2 NM_001291628.1 NM_030621.4 NM_177438.2 XM_011536599.1 XM_011536600.2 XM_011536601.2 XM_011536602.2 XM_011536604.2 XM_011536605.2 XM_017021120.1 XM_017021121.1 XM_017021122.1 XM_017021123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3987846 c.5724C>A p.A1908A Chr.14: 95090543-95090543 on Build GRCh38
4402578 c.5705C>T p.S1902F Chr.14: 95090562-95090562 on Build GRCh38
5950475 c.5762A>G p.N1921S Chr.14: 95090505-95090505 on Build GRCh38
959242 c.5741C>T p.A1914V Chr.14: 95090526-95090526 on Build GRCh38
3370114 c.5718A>C p.R1906S Chr.14: 95090549-95090549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573788003 Chr.14: 95090159-95090159 on Build GRCh38
rs578240601 Chr.14: 95090240-95090240 on Build GRCh38
rs191996364 Chr.14: 95090432-95090432 on Build GRCh38
rs1022635227 Chr.14: 95090221-95090223 on Build GRCh38
rs200095508 Chr.14: 95090478-95090478 on Build GRCh38
rs200498201 Chr.14: 95090172-95090172 on Build GRCh38
rs770907156 Chr.14: 95090493-95090493 on Build GRCh38
rs981079616 Chr.14: 95090266-95090266 on Build GRCh38
rs140021568 Chr.14: 95090364-95090364 on Build GRCh38
rs756294748 Chr.14: 95090463-95090463 on Build GRCh38
rs762281340 Chr.14: 95090539-95090539 on Build GRCh38
rs772958808 Chr.14: 95090475-95090475 on Build GRCh38
rs35649919 Chr.14: 95090172-95090172 on Build GRCh38
rs13078 Chr.14: 95090410-95090410 on Build GRCh38
rs960882464 Chr.14: 95090356-95090356 on Build GRCh38
rs775077210 Chr.14: 95090506-95090506 on Build GRCh38
rs748693590 Chr.14: 95090455-95090455 on Build GRCh38
rs746134430 Chr.14: 95090458-95090458 on Build GRCh38
rs914042436 Chr.14: 95090372-95090372 on Build GRCh38
rs752116341 Chr.14: 95090529-95090529 on Build GRCh38
rs774611849 Chr.14: 95090494-95090494 on Build GRCh38
rs767905778 Chr.14: 95090555-95090555 on Build GRCh38
rs180926609 Chr.14: 95090208-95090208 on Build GRCh38
rs536572479 Chr.14: 95090289-95090289 on Build GRCh38
rs781520963 Chr.14: 95090453-95090453 on Build GRCh38
rs528568542 Chr.14: 95090431-95090431 on Build GRCh38
rs757682330 Chr.14: 95090450-95090450 on Build GRCh38
rs571083676 Chr.14: 95090358-95090358 on Build GRCh38
rs995276380 Chr.14: 95090531-95090531 on Build GRCh38
rs751956931 Chr.14: 95090449-95090449 on Build GRCh38
rs766170875 Chr.14: 95090163-95090163 on Build GRCh38
rs566961540 Chr.14: 95090382-95090382 on Build GRCh38
rs758288467 Chr.14: 95090130-95090130 on Build GRCh38
rs776253418 Chr.14: 95090401-95090401 on Build GRCh38
rs749282967 Chr.14: 95090468-95090468 on Build GRCh38
rs780675412 Chr.14: 95090454-95090455 on Build GRCh38
rs764747360 Chr.14: 95090505-95090505 on Build GRCh38
rs772419252 Chr.14: 95090270-95090274 on Build GRCh38
rs545752774 Chr.14: 95090255-95090255 on Build GRCh38
rs541589888 Chr.14: 95090162-95090162 on Build GRCh38
rs912470196 Chr.14: 95090501-95090501 on Build GRCh38
rs1044532710 Chr.14: 95090576-95090576 on Build GRCh38
rs772063181 Chr.14: 95090472-95090472 on Build GRCh38
rs763651908 Chr.14: 95090519-95090519 on Build GRCh38
rs186123954 Chr.14: 95090288-95090288 on Build GRCh38
rs112795042 Chr.14: 95090332-95090332 on Build GRCh38
rs762301997 Chr.14: 95090512-95090512 on Build GRCh38
rs995643319 Chr.14: 95090184-95090184 on Build GRCh38
rs758181734 Chr.14: 95090161-95090161 on Build GRCh38
rs770494720 Chr.14: 95090387-95090387 on Build GRCh38
rs1015381767 Chr.14: 95090137-95090137 on Build GRCh38
rs1051046532 Chr.14: 95090480-95090480 on Build GRCh38
rs552609115 Chr.14: 95090275-95090275 on Build GRCh38

View Full Product Details