Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95090967-95091487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DICER1
Entrez Gene ID
23405
Gene Name
dicer 1, ribonuclease III
Gene Aliases
DCR1, Dicer, Dicer1e, HERNA, K12H4.8-LIKE, MNG1, RMSE2
UniGene
Hs.738957
Gene Chromosome Location
Chr.14: 95086228-95158010 on Build GRCh38
Transcripts
NM_001195573.1 NM_001271282.2 NM_001291628.1 NM_030621.4 NM_177438.2 XM_011536599.1 XM_011536600.2 XM_011536601.2 XM_011536602.2 XM_011536604.2 XM_011536605.2 XM_017021120.1 XM_017021121.1 XM_017021122.1 XM_017021123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5727309 c.5507C>T p.P1836L Chr.14: 95091223-95091223 on Build GRCh38
470392 c.5517G>T p.R1839R Chr.14: 95091213-95091213 on Build GRCh38
3671992 c.5550C>A p.P1850P Chr.14: 95091087-95091087 on Build GRCh38
959243 c.5463G>A p.M1821I Chr.14: 95091267-95091267 on Build GRCh38
959245 c.5438A>C p.E1813A Chr.14: 95091292-95091292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376486812 Chr.14: 95091147-95091147 on Build GRCh38
rs767777044 Chr.14: 95091371-95091372 on Build GRCh38
rs886037729 Chr.14: 95091265-95091265 on Build GRCh38
rs187288681 Chr.14: 95091223-95091223 on Build GRCh38
rs764254400 Chr.14: 95091193-95091193 on Build GRCh38
rs753581649 Chr.14: 95091368-95091368 on Build GRCh38
rs942778175 Chr.14: 95091459-95091459 on Build GRCh38
rs756025825 Chr.14: 95091342-95091342 on Build GRCh38
rs533000032 Chr.14: 95091446-95091446 on Build GRCh38
rs377325189 Chr.14: 95091087-95091087 on Build GRCh38
rs879181147 Chr.14: 95091380-95091380 on Build GRCh38
rs991370383 Chr.14: 95091338-95091338 on Build GRCh38
rs758254099 Chr.14: 95091420-95091420 on Build GRCh38
rs559119073 Chr.14: 95091421-95091421 on Build GRCh38
rs745795376 Chr.14: 95091081-95091081 on Build GRCh38
rs754838493 Chr.14: 95091369-95091369 on Build GRCh38
rs1042647998 Chr.14: 95091456-95091456 on Build GRCh38
rs770330617 Chr.14: 95091408-95091408 on Build GRCh38
rs774572163 Chr.14: 95091155-95091156 on Build GRCh38
rs587778233 Chr.14: 95091214-95091214 on Build GRCh38
rs773875981 Chr.14: 95091115-95091115 on Build GRCh38
rs752826365 Chr.14: 95091186-95091186 on Build GRCh38
rs766285402 Chr.14: 95091348-95091348 on Build GRCh38
rs748931250 Chr.14: 95091095-95091095 on Build GRCh38
rs918609873 Chr.14: 95091424-95091424 on Build GRCh38
rs759709100 Chr.14: 95091179-95091179 on Build GRCh38
rs747798126 Chr.14: 95091219-95091219 on Build GRCh38
rs879066710 Chr.14: 95091171-95091171 on Build GRCh38
rs528532124 Chr.14: 95091397-95091397 on Build GRCh38
rs377685186 Chr.14: 95091259-95091259 on Build GRCh38
rs778669119 Chr.14: 95091374-95091374 on Build GRCh38
rs751615662 Chr.14: 95091339-95091339 on Build GRCh38
rs375239471 Chr.14: 95091196-95091196 on Build GRCh38
rs886037728 Chr.14: 95091336-95091336 on Build GRCh38
rs373128565 Chr.14: 95090991-95090991 on Build GRCh38
rs766703776 Chr.14: 95091173-95091173 on Build GRCh38
rs777180964 Chr.14: 95091386-95091386 on Build GRCh38
rs749621418 Chr.14: 95091019-95091020 on Build GRCh38
rs768311617 Chr.14: 95091108-95091108 on Build GRCh38
rs747510783 Chr.14: 95091226-95091226 on Build GRCh38
rs370686986 Chr.14: 95091127-95091127 on Build GRCh38
rs201134588 Chr.14: 95091336-95091336 on Build GRCh38
rs781209499 Chr.14: 95090993-95090993 on Build GRCh38
rs747847208 Chr.14: 95091376-95091376 on Build GRCh38
rs763296369 Chr.14: 95091412-95091412 on Build GRCh38
rs758618146 Chr.14: 95091191-95091191 on Build GRCh38
rs779742174 Chr.14: 95091199-95091199 on Build GRCh38
rs769532201 Chr.14: 95091084-95091084 on Build GRCh38
rs777259787 Chr.14: 95091396-95091396 on Build GRCh38
rs17191079 Chr.14: 95091354-95091354 on Build GRCh38
rs775961946 Chr.14: 95091410-95091410 on Build GRCh38
rs878855275 Chr.14: 95091210-95091210 on Build GRCh38
rs1048486911 Chr.14: 95091022-95091022 on Build GRCh38
rs763241498 Chr.14: 95091308-95091308 on Build GRCh38
rs540355845 Chr.14: 95091416-95091416 on Build GRCh38
rs754642180 Chr.14: 95091208-95091208 on Build GRCh38
rs775912475 Chr.14: 95091302-95091302 on Build GRCh38
rs910004071 Chr.14: 95091451-95091451 on Build GRCh38
rs958434315 Chr.14: 95091330-95091330 on Build GRCh38
rs998807097 Chr.14: 95091167-95091167 on Build GRCh38
rs749059266 Chr.14: 95091206-95091206 on Build GRCh38
rs191304237 Chr.14: 95091448-95091448 on Build GRCh38
rs771345378 Chr.14: 95091391-95091391 on Build GRCh38
rs773779559 Chr.14: 95091145-95091145 on Build GRCh38
rs182626158 Chr.14: 95091177-95091177 on Build GRCh38
rs746481371 Chr.14: 95091389-95091389 on Build GRCh38
rs771059329 Chr.14: 95091141-95091142 on Build GRCh38
rs772576507 Chr.14: 95091141-95091141 on Build GRCh38
rs371099251 Chr.14: 95091381-95091381 on Build GRCh38
rs764305391 Chr.14: 95091309-95091309 on Build GRCh38
rs759875628 Chr.14: 95091288-95091288 on Build GRCh38
rs752411788 Chr.14: 95091260-95091260 on Build GRCh38
rs1007089488 Chr.14: 95091131-95091131 on Build GRCh38
rs759659916 Chr.14: 95091172-95091172 on Build GRCh38
rs765497219 Chr.14: 95091184-95091184 on Build GRCh38

View Full Product Details