Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95417809-95418309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE3
Entrez Gene ID
161176
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, NET53, Nesp3
UniGene
Hs.354645
Gene Chromosome Location
Chr.14: 95416083-95516693 on Build GRCh38
Transcripts
NM_152592.3 XM_005267377.3 XM_006720063.3 XM_011536515.2 XM_017021056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3793995 c.2827C>T p.L943L Chr.14: 95417927-95417927 on Build GRCh38
3815613 c.2764A>G p.R922G Chr.14: 95417990-95417990 on Build GRCh38
6365145 c.2774G>T p.C925F Chr.14: 95417980-95417980 on Build GRCh38
393243 c.2802G>C p.L934L Chr.14: 95417952-95417952 on Build GRCh38
6365144 c.2774G>T p.C925F Chr.14: 95417980-95417980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757022479 Chr.14: 95417990-95417991 on Build GRCh38
rs12434804 Chr.14: 95418177-95418177 on Build GRCh38
rs746262942 Chr.14: 95417993-95417993 on Build GRCh38
rs919610209 Chr.14: 95418172-95418172 on Build GRCh38
rs764589082 Chr.14: 95417911-95417911 on Build GRCh38
rs965101762 Chr.14: 95418153-95418153 on Build GRCh38
rs74078423 Chr.14: 95418178-95418178 on Build GRCh38
rs540469515 Chr.14: 95418215-95418215 on Build GRCh38
rs368574040 Chr.14: 95417863-95417863 on Build GRCh38
rs1043013040 Chr.14: 95418017-95418017 on Build GRCh38
rs770618182 Chr.14: 95417834-95417834 on Build GRCh38
rs778446310 Chr.14: 95418035-95418035 on Build GRCh38
rs771485486 Chr.14: 95418058-95418058 on Build GRCh38
rs753687279 Chr.14: 95417925-95417926 on Build GRCh38
rs748810369 Chr.14: 95417984-95417984 on Build GRCh38
rs778693061 Chr.14: 95417873-95417873 on Build GRCh38
rs540014645 Chr.14: 95417902-95417902 on Build GRCh38
rs764535842 Chr.14: 95417859-95417859 on Build GRCh38
rs974034568 Chr.14: 95418125-95418125 on Build GRCh38
rs752219918 Chr.14: 95417854-95417854 on Build GRCh38
rs142276866 Chr.14: 95417974-95417974 on Build GRCh38
rs762297360 Chr.14: 95417920-95417920 on Build GRCh38
rs369096132 Chr.14: 95418013-95418013 on Build GRCh38
rs767839740 Chr.14: 95417921-95417921 on Build GRCh38
rs541841123 Chr.14: 95417961-95417961 on Build GRCh38
rs1039548334 Chr.14: 95418214-95418214 on Build GRCh38
rs766717375 Chr.14: 95417999-95417999 on Build GRCh38
rs745545054 Chr.14: 95417897-95417897 on Build GRCh38
rs890928880 Chr.14: 95417877-95417877 on Build GRCh38
rs114043326 Chr.14: 95418074-95418074 on Build GRCh38
rs150752116 Chr.14: 95417888-95417888 on Build GRCh38
rs761065073 Chr.14: 95417997-95417997 on Build GRCh38
rs912858087 Chr.14: 95418005-95418005 on Build GRCh38
rs368064896 Chr.14: 95418029-95418029 on Build GRCh38
rs534132126 Chr.14: 95418031-95418031 on Build GRCh38
rs12434757 Chr.14: 95417986-95417986 on Build GRCh38
rs770687063 Chr.14: 95417894-95417894 on Build GRCh38
rs111617306 Chr.14: 95418124-95418124 on Build GRCh38
rs375097470 Chr.14: 95417965-95417965 on Build GRCh38
rs951072631 Chr.14: 95418173-95418173 on Build GRCh38
rs754600496 Chr.14: 95417869-95417869 on Build GRCh38
rs144399075 Chr.14: 95417881-95417881 on Build GRCh38
rs764931035 Chr.14: 95418038-95418049 on Build GRCh38
rs865977788 Chr.14: 95418175-95418175 on Build GRCh38
rs777101883 Chr.14: 95418061-95418061 on Build GRCh38
rs960103608 Chr.14: 95418094-95418094 on Build GRCh38
rs115246771 Chr.14: 95418242-95418242 on Build GRCh38
rs1022007472 Chr.14: 95417915-95417915 on Build GRCh38
rs1007025031 Chr.14: 95418113-95418113 on Build GRCh38
rs552484714 Chr.14: 95418039-95418039 on Build GRCh38
rs373636276 Chr.14: 95418027-95418027 on Build GRCh38
rs895246610 Chr.14: 95418046-95418046 on Build GRCh38
rs566185678 Chr.14: 95418014-95418014 on Build GRCh38
rs144473753 Chr.14: 95417985-95417985 on Build GRCh38
rs749360218 Chr.14: 95417876-95417876 on Build GRCh38
rs762269580 Chr.14: 95417992-95417992 on Build GRCh38
rs544195830 Chr.14: 95418248-95418248 on Build GRCh38
rs753968152 Chr.14: 95417936-95417936 on Build GRCh38
rs747008248 Chr.14: 95417943-95417943 on Build GRCh38
rs781491729 Chr.14: 95417865-95417865 on Build GRCh38
rs750670879 Chr.14: 95417867-95417867 on Build GRCh38
rs73339120 Chr.14: 95417880-95417880 on Build GRCh38
rs35838903 Chr.14: 95418164-95418165 on Build GRCh38
rs2761903 Chr.14: 95418261-95418261 on Build GRCh38
rs765505660 Chr.14: 95418063-95418063 on Build GRCh38
rs998890258 Chr.14: 95418250-95418250 on Build GRCh38
rs558453966 Chr.14: 95418241-95418241 on Build GRCh38
rs750772416 Chr.14: 95417924-95417924 on Build GRCh38
rs776086121 Chr.14: 95417836-95417836 on Build GRCh38
rs200254064 Chr.14: 95417903-95417903 on Build GRCh38
rs1057150610 Chr.14: 95418275-95418275 on Build GRCh38
rs370277221 Chr.14: 95418020-95418020 on Build GRCh38
rs1043019314 Chr.14: 95418237-95418244 on Build GRCh38
rs370007886 Chr.14: 95417973-95417973 on Build GRCh38
rs779120100 Chr.14: 95417939-95417939 on Build GRCh38
rs144877155 Chr.14: 95417946-95417946 on Build GRCh38
rs766579567 Chr.14: 95417928-95417928 on Build GRCh38
rs903162314 Chr.14: 95418084-95418084 on Build GRCh38
rs781142957 Chr.14: 95418022-95418022 on Build GRCh38
rs10130647 Chr.14: 95417918-95417918 on Build GRCh38
rs759211520 Chr.14: 95417851-95417851 on Build GRCh38
rs773707451 Chr.14: 95418041-95418041 on Build GRCh38
rs762409860 Chr.14: 95417907-95417907 on Build GRCh38
rs761069662 Chr.14: 95418044-95418044 on Build GRCh38
rs377616880 Chr.14: 95418224-95418224 on Build GRCh38
rs141381311 Chr.14: 95418023-95418023 on Build GRCh38
rs1013644157 Chr.14: 95418051-95418051 on Build GRCh38
rs116644125 Chr.14: 95418062-95418062 on Build GRCh38
rs1035805908 Chr.14: 95418089-95418089 on Build GRCh38
rs776479683 Chr.14: 95417895-95417895 on Build GRCh38
rs149881773 Chr.14: 95417937-95417937 on Build GRCh38
rs371488280 Chr.14: 95418030-95418030 on Build GRCh38
rs765445342 Chr.14: 95418019-95418019 on Build GRCh38
rs998839249 Chr.14: 95418086-95418086 on Build GRCh38
rs760503239 Chr.14: 95417832-95417832 on Build GRCh38
rs201427592 Chr.14: 95418075-95418075 on Build GRCh38
rs772491087 Chr.14: 95418040-95418040 on Build GRCh38
rs200981167 Chr.14: 95418012-95418012 on Build GRCh38
rs1040663992 Chr.14: 95417841-95417841 on Build GRCh38
rs952940721 Chr.14: 95418096-95418096 on Build GRCh38
rs764718779 Chr.14: 95417852-95417852 on Build GRCh38
rs947344537 Chr.14: 95418220-95418220 on Build GRCh38
rs1027260431 Chr.14: 95418095-95418095 on Build GRCh38
rs146368343 Chr.14: 95417872-95417872 on Build GRCh38
rs1740644 Chr.14: 95418174-95418174 on Build GRCh38
rs763499489 Chr.14: 95417855-95417855 on Build GRCh38
rs756343495 Chr.14: 95417925-95417925 on Build GRCh38
rs780897044 Chr.14: 95417958-95417958 on Build GRCh38
rs750432904 Chr.14: 95418076-95418076 on Build GRCh38
rs752750922 Chr.14: 95418071-95418071 on Build GRCh38
rs1025400455 Chr.14: 95418054-95418054 on Build GRCh38
rs562415245 Chr.14: 95418281-95418281 on Build GRCh38

View Full Product Details