Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95436698-95437210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNE3
Entrez Gene ID
161176
Gene Name
spectrin repeat containing nuclear envelope family member 3
Gene Aliases
C14orf49, NET53, Nesp3
UniGene
Hs.354645
Gene Chromosome Location
Chr.14: 95416083-95516693 on Build GRCh38
Transcripts
NM_152592.3 XM_005267377.3 XM_006720063.3 XM_011536515.2 XM_017021056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5713250 c.2385_2387delTCT p.L796delL Chr.14: 95436971-95436973 on Build GRCh38
5713252 c.2385_2387delTCT p.L796delL Chr.14: 95436971-95436973 on Build GRCh38
3386765 c.2528C>G p.S843W Chr.14: 95436830-95436830 on Build GRCh38
4878395 c.2449T>C p.L817L Chr.14: 95436909-95436909 on Build GRCh38
214548 c.2387_2389delTAC p.L796delL Chr.14: 95436969-95436971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75942423 Chr.14: 95437098-95437098 on Build GRCh38
rs751938689 Chr.14: 95436977-95436977 on Build GRCh38
rs763419751 Chr.14: 95436961-95436961 on Build GRCh38
rs776027792 Chr.14: 95436953-95436953 on Build GRCh38
rs144719514 Chr.14: 95437140-95437140 on Build GRCh38
rs747419668 Chr.14: 95437177-95437177 on Build GRCh38
rs1054211805 Chr.14: 95437043-95437043 on Build GRCh38
rs759010266 Chr.14: 95436930-95436930 on Build GRCh38
rs1010041001 Chr.14: 95437090-95437090 on Build GRCh38
rs531490074 Chr.14: 95437060-95437060 on Build GRCh38
rs753460822 Chr.14: 95437097-95437097 on Build GRCh38
rs774299592 Chr.14: 95436792-95436792 on Build GRCh38
rs774046477 Chr.14: 95436754-95436754 on Build GRCh38
rs200878334 Chr.14: 95437008-95437008 on Build GRCh38
rs763509790 Chr.14: 95437027-95437027 on Build GRCh38
rs757729261 Chr.14: 95437188-95437188 on Build GRCh38
rs376157244 Chr.14: 95436815-95436815 on Build GRCh38
rs375798321 Chr.14: 95437025-95437025 on Build GRCh38
rs183905400 Chr.14: 95436997-95436997 on Build GRCh38
rs745499387 Chr.14: 95437024-95437024 on Build GRCh38
rs762220103 Chr.14: 95437031-95437031 on Build GRCh38
rs202043284 Chr.14: 95436998-95436998 on Build GRCh38
rs1002773859 Chr.14: 95437087-95437087 on Build GRCh38
rs972183523 Chr.14: 95437141-95437142 on Build GRCh38
rs765842954 Chr.14: 95436803-95436803 on Build GRCh38
rs749569675 Chr.14: 95436957-95436959 on Build GRCh38
rs531873700 Chr.14: 95436866-95436866 on Build GRCh38
rs905817519 Chr.14: 95437083-95437083 on Build GRCh38
rs773024495 Chr.14: 95436903-95436903 on Build GRCh38
rs749250357 Chr.14: 95436777-95436777 on Build GRCh38
rs759619847 Chr.14: 95437018-95437020 on Build GRCh38
rs372184224 Chr.14: 95436802-95436802 on Build GRCh38
rs537347555 Chr.14: 95436910-95436910 on Build GRCh38
rs754969110 Chr.14: 95436876-95436876 on Build GRCh38
rs770260425 Chr.14: 95436794-95436795 on Build GRCh38
rs746755589 Chr.14: 95437020-95437020 on Build GRCh38
rs757497360 Chr.14: 95436873-95436873 on Build GRCh38
rs778952658 Chr.14: 95436890-95436890 on Build GRCh38
rs747882894 Chr.14: 95436793-95436793 on Build GRCh38
rs555598757 Chr.14: 95436952-95436952 on Build GRCh38
rs369228456 Chr.14: 95436790-95436790 on Build GRCh38
rs921244093 Chr.14: 95437119-95437119 on Build GRCh38
rs147820834 Chr.14: 95436732-95436732 on Build GRCh38
rs1035552723 Chr.14: 95437091-95437091 on Build GRCh38
rs764421402 Chr.14: 95436844-95436844 on Build GRCh38
rs747975370 Chr.14: 95436896-95436896 on Build GRCh38
rs912746124 Chr.14: 95437180-95437180 on Build GRCh38
rs748132626 Chr.14: 95437007-95437007 on Build GRCh38
rs200011095 Chr.14: 95436874-95436874 on Build GRCh38
rs768731428 Chr.14: 95437022-95437023 on Build GRCh38
rs758715860 Chr.14: 95436830-95436830 on Build GRCh38
rs777967910 Chr.14: 95436895-95436896 on Build GRCh38
rs185935331 Chr.14: 95437100-95437100 on Build GRCh38
rs78849563 Chr.14: 95436723-95436723 on Build GRCh38
rs76499929 Chr.14: 95436971-95436973 on Build GRCh38
rs369215627 Chr.14: 95436865-95436865 on Build GRCh38
rs778146649 Chr.14: 95437112-95437112 on Build GRCh38
rs971398111 Chr.14: 95436811-95436811 on Build GRCh38
rs772039220 Chr.14: 95436901-95436901 on Build GRCh38
rs764511039 Chr.14: 95436965-95436965 on Build GRCh38
rs769147719 Chr.14: 95437028-95437028 on Build GRCh38
rs751153719 Chr.14: 95436956-95436956 on Build GRCh38
rs565815934 Chr.14: 95437190-95437190 on Build GRCh38
rs974356373 Chr.14: 95436954-95436954 on Build GRCh38
rs962752334 Chr.14: 95437101-95437101 on Build GRCh38
rs200146914 Chr.14: 95437023-95437023 on Build GRCh38
rs765669538 Chr.14: 95436817-95436817 on Build GRCh38
rs757433429 Chr.14: 95436984-95436984 on Build GRCh38
rs540053794 Chr.14: 95436829-95436829 on Build GRCh38
rs771879833 Chr.14: 95436800-95436800 on Build GRCh38
rs776012276 Chr.14: 95436920-95436920 on Build GRCh38
rs768700866 Chr.14: 95436780-95436780 on Build GRCh38
rs192178494 Chr.14: 95436801-95436801 on Build GRCh38
rs548109975 Chr.14: 95437145-95437145 on Build GRCh38
rs201240192 Chr.14: 95436969-95436971 on Build GRCh38
rs1016317050 Chr.14: 95437124-95437124 on Build GRCh38
rs770563232 Chr.14: 95436917-95436917 on Build GRCh38
rs36036653 Chr.14: 95437109-95437109 on Build GRCh38
rs909409683 Chr.14: 95436916-95436916 on Build GRCh38
rs57883787 Chr.14: 95437006-95437006 on Build GRCh38
rs9671893 Chr.14: 95437062-95437062 on Build GRCh38
rs528943316 Chr.14: 95437109-95437109 on Build GRCh38
rs370181203 Chr.14: 95437026-95437026 on Build GRCh38

View Full Product Details