Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96242038-96242541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDKRB2
Entrez Gene ID
624
Gene Name
bradykinin receptor B2
Gene Aliases
B2R, BK-2, BK2, BKR2, BRB2
UniGene
Hs.654542
Gene Chromosome Location
Chr.14: 96204798-96244329 on Build GRCh38
Transcripts
NM_000623.3

SNP Detail

SNP ID Chromosome Location
rs2242968 Chr.14: 96242385-96242385 on Build GRCh38
rs201997774 Chr.14: 96242237-96242237 on Build GRCh38
rs200662380 Chr.14: 96242227-96242227 on Build GRCh38
rs201410293 Chr.14: 96242191-96242191 on Build GRCh38
rs1048096280 Chr.14: 96242455-96242455 on Build GRCh38
rs1028982246 Chr.14: 96242290-96242290 on Build GRCh38
rs200905836 Chr.14: 96242097-96242097 on Build GRCh38
rs201574316 Chr.14: 96242125-96242125 on Build GRCh38
rs924691506 Chr.14: 96242148-96242148 on Build GRCh38
rs921295747 Chr.14: 96242077-96242077 on Build GRCh38
rs200393121 Chr.14: 96242121-96242121 on Build GRCh38
rs200081924 Chr.14: 96242258-96242258 on Build GRCh38
rs936625622 Chr.14: 96242422-96242422 on Build GRCh38
rs201676732 Chr.14: 96242456-96242456 on Build GRCh38
rs143560728 Chr.14: 96242343-96242343 on Build GRCh38
rs200247341 Chr.14: 96242187-96242187 on Build GRCh38
rs1013851702 Chr.14: 96242432-96242432 on Build GRCh38
rs949348855 Chr.14: 96242222-96242222 on Build GRCh38
rs867155008 Chr.14: 96242349-96242349 on Build GRCh38
rs964960590 Chr.14: 96242519-96242519 on Build GRCh38
rs114065472 Chr.14: 96242205-96242205 on Build GRCh38
rs2069591 Chr.14: 96242495-96242495 on Build GRCh38
rs370819142 Chr.14: 96242301-96242301 on Build GRCh38
rs910765198 Chr.14: 96242327-96242327 on Build GRCh38
rs201450248 Chr.14: 96242370-96242370 on Build GRCh38
rs995162818 Chr.14: 96242238-96242238 on Build GRCh38
rs978846898 Chr.14: 96242110-96242110 on Build GRCh38
rs910691592 Chr.14: 96242075-96242075 on Build GRCh38
rs201042178 Chr.14: 96242303-96242303 on Build GRCh38
rs202134083 Chr.14: 96242268-96242268 on Build GRCh38
rs200543485 Chr.14: 96242369-96242369 on Build GRCh38
rs2069592 Chr.14: 96242511-96242511 on Build GRCh38
rs1040855433 Chr.14: 96242113-96242113 on Build GRCh38
rs1009967944 Chr.14: 96242124-96242124 on Build GRCh38
rs759869326 Chr.14: 96242333-96242333 on Build GRCh38
rs373967767 Chr.14: 96242136-96242136 on Build GRCh38
rs200113595 Chr.14: 96242089-96242089 on Build GRCh38
rs201421328 Chr.14: 96242430-96242430 on Build GRCh38
rs923361073 Chr.14: 96242357-96242357 on Build GRCh38
rs1040490563 Chr.14: 96242340-96242340 on Build GRCh38
rs896902229 Chr.14: 96242449-96242449 on Build GRCh38
rs5226 Chr.14: 96242206-96242206 on Build GRCh38
rs199852386 Chr.14: 96242295-96242295 on Build GRCh38
rs1025132880 Chr.14: 96242249-96242249 on Build GRCh38
rs148890334 Chr.14: 96242240-96242240 on Build GRCh38
rs41299185 Chr.14: 96242510-96242510 on Build GRCh38
rs201734929 Chr.14: 96242256-96242256 on Build GRCh38
rs2069588 Chr.14: 96242330-96242330 on Build GRCh38
rs765071398 Chr.14: 96242377-96242377 on Build GRCh38
rs200907522 Chr.14: 96242450-96242450 on Build GRCh38
rs968673658 Chr.14: 96242099-96242099 on Build GRCh38
rs199828613 Chr.14: 96242428-96242428 on Build GRCh38
rs886857983 Chr.14: 96242241-96242241 on Build GRCh38
rs13306220 Chr.14: 96242091-96242091 on Build GRCh38
rs935206586 Chr.14: 96242158-96242158 on Build GRCh38
rs199498029 Chr.14: 96242476-96242476 on Build GRCh38
rs115774314 Chr.14: 96242262-96242262 on Build GRCh38
rs1026688493 Chr.14: 96242496-96242496 on Build GRCh38
rs202117207 Chr.14: 96242166-96242166 on Build GRCh38
rs199505173 Chr.14: 96242221-96242221 on Build GRCh38
rs202074566 Chr.14: 96242103-96242103 on Build GRCh38
rs1049263114 Chr.14: 96242068-96242068 on Build GRCh38
rs2069589 Chr.14: 96242379-96242379 on Build GRCh38
rs560183247 Chr.14: 96242393-96242393 on Build GRCh38
rs200717123 Chr.14: 96242250-96242250 on Build GRCh38
rs1017854970 Chr.14: 96242512-96242512 on Build GRCh38
rs2069590 Chr.14: 96242445-96242445 on Build GRCh38
rs200311570 Chr.14: 96242429-96242429 on Build GRCh38
rs199754651 Chr.14: 96242198-96242198 on Build GRCh38
rs200583746 Chr.14: 96242480-96242480 on Build GRCh38
rs1033480202 Chr.14: 96242233-96242233 on Build GRCh38
rs997132762 Chr.14: 96242274-96242274 on Build GRCh38
rs201157943 Chr.14: 96242418-96242418 on Build GRCh38
rs577161706 Chr.14: 96242117-96242117 on Build GRCh38

View Full Product Details