Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96242233-96242733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDKRB2
Entrez Gene ID
624
Gene Name
bradykinin receptor B2
Gene Aliases
B2R, BK-2, BK2, BKR2, BRB2
UniGene
Hs.654542
Gene Chromosome Location
Chr.14: 96204798-96244329 on Build GRCh38
Transcripts
NM_000623.3

SNP Detail

SNP ID Chromosome Location
rs112642055 Chr.14: 96242582-96242582 on Build GRCh38
rs2242968 Chr.14: 96242385-96242385 on Build GRCh38
rs949292775 Chr.14: 96242612-96242612 on Build GRCh38
rs1048096280 Chr.14: 96242455-96242455 on Build GRCh38
rs945460210 Chr.14: 96242537-96242537 on Build GRCh38
rs1028982246 Chr.14: 96242290-96242290 on Build GRCh38
rs200557132 Chr.14: 96242627-96242627 on Build GRCh38
rs200081924 Chr.14: 96242258-96242258 on Build GRCh38
rs1006573216 Chr.14: 96242693-96242693 on Build GRCh38
rs936625622 Chr.14: 96242422-96242422 on Build GRCh38
rs200357797 Chr.14: 96242640-96242640 on Build GRCh38
rs201676732 Chr.14: 96242456-96242456 on Build GRCh38
rs143560728 Chr.14: 96242343-96242343 on Build GRCh38
rs1013851702 Chr.14: 96242432-96242432 on Build GRCh38
rs2069591 Chr.14: 96242495-96242495 on Build GRCh38
rs867155008 Chr.14: 96242349-96242349 on Build GRCh38
rs964960590 Chr.14: 96242519-96242519 on Build GRCh38
rs370819142 Chr.14: 96242301-96242301 on Build GRCh38
rs201400176 Chr.14: 96242681-96242681 on Build GRCh38
rs910765198 Chr.14: 96242327-96242327 on Build GRCh38
rs755883719 Chr.14: 96242585-96242585 on Build GRCh38
rs201450248 Chr.14: 96242370-96242370 on Build GRCh38
rs202217547 Chr.14: 96242599-96242599 on Build GRCh38
rs201042178 Chr.14: 96242303-96242303 on Build GRCh38
rs202134083 Chr.14: 96242268-96242268 on Build GRCh38
rs779665575 Chr.14: 96242646-96242646 on Build GRCh38
rs200543485 Chr.14: 96242369-96242369 on Build GRCh38
rs2069592 Chr.14: 96242511-96242511 on Build GRCh38
rs759869326 Chr.14: 96242333-96242333 on Build GRCh38
rs895086082 Chr.14: 96242609-96242609 on Build GRCh38
rs1017929016 Chr.14: 96242710-96242710 on Build GRCh38
rs896902229 Chr.14: 96242449-96242449 on Build GRCh38
rs923361073 Chr.14: 96242357-96242357 on Build GRCh38
rs1040490563 Chr.14: 96242340-96242340 on Build GRCh38
rs201421328 Chr.14: 96242430-96242430 on Build GRCh38
rs201708437 Chr.14: 96242528-96242528 on Build GRCh38
rs936663059 Chr.14: 96242568-96242568 on Build GRCh38
rs199644339 Chr.14: 96242637-96242637 on Build GRCh38
rs199852386 Chr.14: 96242295-96242295 on Build GRCh38
rs41299185 Chr.14: 96242510-96242510 on Build GRCh38
rs201734929 Chr.14: 96242256-96242256 on Build GRCh38
rs2069588 Chr.14: 96242330-96242330 on Build GRCh38
rs765071398 Chr.14: 96242377-96242377 on Build GRCh38
rs200907522 Chr.14: 96242450-96242450 on Build GRCh38
rs199828613 Chr.14: 96242428-96242428 on Build GRCh38
rs192962712 Chr.14: 96242711-96242711 on Build GRCh38
rs201873675 Chr.14: 96242543-96242543 on Build GRCh38
rs1046668842 Chr.14: 96242616-96242616 on Build GRCh38
rs199498029 Chr.14: 96242476-96242476 on Build GRCh38
rs115774314 Chr.14: 96242262-96242262 on Build GRCh38
rs1026688493 Chr.14: 96242496-96242496 on Build GRCh38
rs574561251 Chr.14: 96242610-96242610 on Build GRCh38
rs200886163 Chr.14: 96242542-96242542 on Build GRCh38
rs926408265 Chr.14: 96242630-96242630 on Build GRCh38
rs2069593 Chr.14: 96242628-96242628 on Build GRCh38
rs925325391 Chr.14: 96242546-96242546 on Build GRCh38
rs2069589 Chr.14: 96242379-96242379 on Build GRCh38
rs371042075 Chr.14: 96242621-96242621 on Build GRCh38
rs560183247 Chr.14: 96242393-96242393 on Build GRCh38
rs1017854970 Chr.14: 96242512-96242512 on Build GRCh38
rs200311570 Chr.14: 96242429-96242429 on Build GRCh38
rs2069590 Chr.14: 96242445-96242445 on Build GRCh38
rs200583746 Chr.14: 96242480-96242480 on Build GRCh38
rs997132762 Chr.14: 96242274-96242274 on Build GRCh38
rs200098225 Chr.14: 96242536-96242536 on Build GRCh38
rs750035237 Chr.14: 96242575-96242575 on Build GRCh38
rs200200402 Chr.14: 96242558-96242558 on Build GRCh38
rs201157943 Chr.14: 96242418-96242418 on Build GRCh38
rs200855525 Chr.14: 96242586-96242586 on Build GRCh38

View Full Product Details