Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96243231-96243738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDKRB2
Entrez Gene ID
624
Gene Name
bradykinin receptor B2
Gene Aliases
B2R, BK-2, BK2, BKR2, BRB2
UniGene
Hs.654542
Gene Chromosome Location
Chr.14: 96204798-96244329 on Build GRCh38
Transcripts
NM_000623.3

SNP Detail

SNP ID Chromosome Location
rs201352503 Chr.14: 96243713-96243713 on Build GRCh38
rs111933870 Chr.14: 96243448-96243448 on Build GRCh38
rs200322671 Chr.14: 96243538-96243538 on Build GRCh38
rs1041752866 Chr.14: 96243422-96243422 on Build GRCh38
rs200529240 Chr.14: 96243546-96243546 on Build GRCh38
rs200149243 Chr.14: 96243636-96243636 on Build GRCh38
rs200263353 Chr.14: 96243705-96243705 on Build GRCh38
rs112800751 Chr.14: 96243440-96243440 on Build GRCh38
rs918542372 Chr.14: 96243554-96243554 on Build GRCh38
rs997875261 Chr.14: 96243352-96243352 on Build GRCh38
rs199831419 Chr.14: 96243286-96243286 on Build GRCh38
rs371467739 Chr.14: 96243651-96243651 on Build GRCh38
rs201156230 Chr.14: 96243257-96243257 on Build GRCh38
rs181901959 Chr.14: 96243512-96243512 on Build GRCh38
rs111413870 Chr.14: 96243541-96243541 on Build GRCh38
rs199508762 Chr.14: 96243340-96243340 on Build GRCh38
rs902767866 Chr.14: 96243432-96243432 on Build GRCh38
rs1053843922 Chr.14: 96243484-96243484 on Build GRCh38
rs202120220 Chr.14: 96243696-96243696 on Build GRCh38
rs199698927 Chr.14: 96243560-96243560 on Build GRCh38
rs201964448 Chr.14: 96243392-96243392 on Build GRCh38
rs373547737 Chr.14: 96243473-96243473 on Build GRCh38
rs200839277 Chr.14: 96243599-96243599 on Build GRCh38
rs201872393 Chr.14: 96243434-96243434 on Build GRCh38
rs974155269 Chr.14: 96243546-96243560 on Build GRCh38
rs200096088 Chr.14: 96243686-96243686 on Build GRCh38
rs531325959 Chr.14: 96243284-96243284 on Build GRCh38
rs200091753 Chr.14: 96243397-96243397 on Build GRCh38
rs200238113 Chr.14: 96243301-96243301 on Build GRCh38
rs935713037 Chr.14: 96243580-96243580 on Build GRCh38
rs200981135 Chr.14: 96243386-96243386 on Build GRCh38
rs144391658 Chr.14: 96243482-96243497 on Build GRCh38
rs369857475 Chr.14: 96243417-96243417 on Build GRCh38
rs200747021 Chr.14: 96243498-96243498 on Build GRCh38
rs200782024 Chr.14: 96243647-96243647 on Build GRCh38
rs201786252 Chr.14: 96243351-96243351 on Build GRCh38
rs201367675 Chr.14: 96243497-96243497 on Build GRCh38
rs1027730222 Chr.14: 96243475-96243475 on Build GRCh38
rs1053222845 Chr.14: 96243618-96243618 on Build GRCh38
rs201002921 Chr.14: 96243563-96243563 on Build GRCh38
rs201837321 Chr.14: 96243477-96243477 on Build GRCh38
rs199688381 Chr.14: 96243316-96243316 on Build GRCh38
rs1005388667 Chr.14: 96243519-96243519 on Build GRCh38
rs200609067 Chr.14: 96243496-96243496 on Build GRCh38
rs201793160 Chr.14: 96243677-96243677 on Build GRCh38
rs892243997 Chr.14: 96243487-96243487 on Build GRCh38
rs982150596 Chr.14: 96243552-96243552 on Build GRCh38
rs111641549 Chr.14: 96243402-96243402 on Build GRCh38
rs199656317 Chr.14: 96243500-96243500 on Build GRCh38
rs201674904 Chr.14: 96243334-96243334 on Build GRCh38
rs1001529150 Chr.14: 96243707-96243707 on Build GRCh38
rs113463024 Chr.14: 96243374-96243374 on Build GRCh38
rs905889986 Chr.14: 96243704-96243704 on Build GRCh38
rs1023708765 Chr.14: 96243540-96243540 on Build GRCh38
rs201536679 Chr.14: 96243315-96243315 on Build GRCh38
rs201403893 Chr.14: 96243539-96243539 on Build GRCh38
rs1015398030 Chr.14: 96243525-96243525 on Build GRCh38
rs201220162 Chr.14: 96243431-96243431 on Build GRCh38
rs1046251 Chr.14: 96243679-96243679 on Build GRCh38
rs922395305 Chr.14: 96243711-96243711 on Build GRCh38
rs200997007 Chr.14: 96243333-96243333 on Build GRCh38
rs201223591 Chr.14: 96243691-96243691 on Build GRCh38
rs775506564 Chr.14: 96243516-96243516 on Build GRCh38
rs892044361 Chr.14: 96243436-96243436 on Build GRCh38
rs201729839 Chr.14: 96243553-96243553 on Build GRCh38
rs913368914 Chr.14: 96243646-96243646 on Build GRCh38
rs984029247 Chr.14: 96243601-96243601 on Build GRCh38
rs566250182 Chr.14: 96243569-96243569 on Build GRCh38
rs909754373 Chr.14: 96243650-96243650 on Build GRCh38
rs878953306 Chr.14: 96243679-96243680 on Build GRCh38
rs201531634 Chr.14: 96243606-96243606 on Build GRCh38
rs952559746 Chr.14: 96243502-96243502 on Build GRCh38
rs200587531 Chr.14: 96243346-96243346 on Build GRCh38
rs1014473039 Chr.14: 96243439-96243439 on Build GRCh38
rs13306218 Chr.14: 96243466-96243466 on Build GRCh38
rs13306217 Chr.14: 96243465-96243465 on Build GRCh38
rs201838517 Chr.14: 96243591-96243591 on Build GRCh38
rs111359094 Chr.14: 96243326-96243326 on Build GRCh38
rs551890783 Chr.14: 96243658-96243658 on Build GRCh38
rs199852145 Chr.14: 96243594-96243594 on Build GRCh38

View Full Product Details