Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96437547-96438068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK7
Entrez Gene ID
122481
Gene Name
adenylate kinase 7
Gene Aliases
AK 7
UniGene
Hs.667462
Gene Chromosome Location
Chr.14: 96392111-96489427 on Build GRCh38
Transcripts
NM_152327.3 XM_006720021.2 XM_006720022.2 XM_011536403.2 XM_017020956.1 XM_017020957.1 XM_017020958.1 XM_017020959.1 XM_017020960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4617558 c.642T>C p.A214A Chr.14: 96437867-96437867 on Build GRCh38
4053363 c.690+1G>A p.? Chr.14: 96437916-96437916 on Build GRCh38
3499592 c.655G>A p.G219R Chr.14: 96437880-96437880 on Build GRCh38
6252011 c.622G>T p.A208S Chr.14: 96437847-96437847 on Build GRCh38
7310321 c.610-5A>T p.? Chr.14: 96437830-96437830 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775781680 Chr.14: 96437790-96437790 on Build GRCh38
rs756025716 Chr.14: 96437832-96437832 on Build GRCh38
rs150648522 Chr.14: 96437877-96437877 on Build GRCh38
rs778534347 Chr.14: 96437855-96437855 on Build GRCh38
rs896091070 Chr.14: 96437608-96437608 on Build GRCh38
rs367856999 Chr.14: 96437777-96437777 on Build GRCh38
rs924370227 Chr.14: 96437730-96437730 on Build GRCh38
rs140456762 Chr.14: 96437618-96437618 on Build GRCh38
rs377456389 Chr.14: 96437935-96437935 on Build GRCh38
rs201330148 Chr.14: 96437856-96437856 on Build GRCh38
rs775945196 Chr.14: 96437967-96437967 on Build GRCh38
rs139741113 Chr.14: 96437884-96437884 on Build GRCh38
rs754738536 Chr.14: 96437932-96437932 on Build GRCh38
rs572486060 Chr.14: 96437715-96437715 on Build GRCh38
rs558260298 Chr.14: 96437942-96437942 on Build GRCh38
rs766096366 Chr.14: 96437835-96437835 on Build GRCh38
rs186036906 Chr.14: 96437960-96437960 on Build GRCh38
rs563459379 Chr.14: 96437704-96437704 on Build GRCh38
rs753702630 Chr.14: 96437931-96437931 on Build GRCh38
rs147444886 Chr.14: 96437906-96437906 on Build GRCh38
rs996593910 Chr.14: 96437982-96437982 on Build GRCh38
rs554775360 Chr.14: 96437644-96437644 on Build GRCh38
rs150396858 Chr.14: 96437951-96437951 on Build GRCh38
rs199760145 Chr.14: 96437836-96437836 on Build GRCh38
rs530982568 Chr.14: 96437720-96437720 on Build GRCh38
rs201546130 Chr.14: 96437885-96437885 on Build GRCh38
rs550666818 Chr.14: 96437930-96437930 on Build GRCh38
rs770205923 Chr.14: 96437896-96437896 on Build GRCh38
rs767437918 Chr.14: 96437808-96437808 on Build GRCh38
rs930812163 Chr.14: 96437796-96437796 on Build GRCh38
rs752148565 Chr.14: 96437703-96437703 on Build GRCh38
rs917494482 Chr.14: 96437745-96437745 on Build GRCh38
rs764099443 Chr.14: 96437916-96437916 on Build GRCh38
rs900477378 Chr.14: 96437971-96437971 on Build GRCh38
rs1050431889 Chr.14: 96437922-96437922 on Build GRCh38
rs760376366 Chr.14: 96437891-96437891 on Build GRCh38
rs948986594 Chr.14: 96437747-96437747 on Build GRCh38
rs549146406 Chr.14: 96437768-96437768 on Build GRCh38
rs182057750 Chr.14: 96437662-96437662 on Build GRCh38
rs753576637 Chr.14: 96437845-96437845 on Build GRCh38
rs762931603 Chr.14: 96437802-96437802 on Build GRCh38
rs542863751 Chr.14: 96437636-96437636 on Build GRCh38
rs746219659 Chr.14: 96437895-96437895 on Build GRCh38
rs765489493 Chr.14: 96437786-96437786 on Build GRCh38
rs545061117 Chr.14: 96437697-96437697 on Build GRCh38
rs11624031 Chr.14: 96437671-96437671 on Build GRCh38
rs777148549 Chr.14: 96437893-96437893 on Build GRCh38
rs929577101 Chr.14: 96438003-96438003 on Build GRCh38
rs770258785 Chr.14: 96437962-96437962 on Build GRCh38
rs750302341 Chr.14: 96437826-96437826 on Build GRCh38
rs45628241 Chr.14: 96437680-96437680 on Build GRCh38
rs146644995 Chr.14: 96437959-96437959 on Build GRCh38
rs771113927 Chr.14: 96437807-96437808 on Build GRCh38
rs575334419 Chr.14: 96437575-96437575 on Build GRCh38
rs960430648 Chr.14: 96437734-96437734 on Build GRCh38
rs143022548 Chr.14: 96437902-96437902 on Build GRCh38
rs754687055 Chr.14: 96437851-96437851 on Build GRCh38
rs760546995 Chr.14: 96437928-96437928 on Build GRCh38
rs777290080 Chr.14: 96437864-96437864 on Build GRCh38
rs1013312589 Chr.14: 96437735-96437735 on Build GRCh38
rs759519981 Chr.14: 96437786-96437786 on Build GRCh38
rs968554061 Chr.14: 96437882-96437882 on Build GRCh38
rs372680114 Chr.14: 96437818-96437818 on Build GRCh38
rs899240467 Chr.14: 96437652-96437652 on Build GRCh38
rs370081121 Chr.14: 96437947-96437947 on Build GRCh38
rs1040349733 Chr.14: 96437969-96437969 on Build GRCh38
rs369947338 Chr.14: 96437936-96437936 on Build GRCh38
rs965287925 Chr.14: 96437687-96437687 on Build GRCh38
rs111711167 Chr.14: 96437853-96437853 on Build GRCh38
rs757826277 Chr.14: 96437859-96437859 on Build GRCh38
rs189456619 Chr.14: 96437656-96437656 on Build GRCh38
rs1018929655 Chr.14: 96437843-96437843 on Build GRCh38
rs746469329 Chr.14: 96437955-96437955 on Build GRCh38
rs996679957 Chr.14: 96437863-96437863 on Build GRCh38
rs992488897 Chr.14: 96437900-96437900 on Build GRCh38
rs776309966 Chr.14: 96437661-96437661 on Build GRCh38

View Full Product Details