Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99495409-99495921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNK
Entrez Gene ID
8812
Gene Name
cyclin K
Gene Aliases
CPR4
UniGene
Hs.510409
Gene Chromosome Location
Chr.14: 99481402-99511515 on Build GRCh38
Transcripts
NM_001099402.1 XM_005268154.4 XM_011537275.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
699667 c.395T>A p.F132Y Chr.14: 99495613-99495613 on Build GRCh38
3499658 c.411G>A p.K137K Chr.14: 99495629-99495629 on Build GRCh38
7140496 c.388G>T p.G130C Chr.14: 99495606-99495606 on Build GRCh38
6208425 c.310G>T p.G104W Chr.14: 99495528-99495528 on Build GRCh38
5724530 c.390C>T p.G130G Chr.14: 99495608-99495608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979069327 Chr.14: 99495894-99495894 on Build GRCh38
rs916273501 Chr.14: 99495449-99495449 on Build GRCh38
rs3918071 Chr.14: 99495841-99495845 on Build GRCh38
rs923576349 Chr.14: 99495897-99495897 on Build GRCh38
rs914802672 Chr.14: 99495882-99495882 on Build GRCh38
rs1015053067 Chr.14: 99495728-99495728 on Build GRCh38
rs1044269692 Chr.14: 99495872-99495872 on Build GRCh38
rs868692667 Chr.14: 99495531-99495531 on Build GRCh38
rs777518234 Chr.14: 99495674-99495675 on Build GRCh38
rs760766785 Chr.14: 99495494-99495494 on Build GRCh38
rs757090553 Chr.14: 99495545-99495545 on Build GRCh38
rs199582133 Chr.14: 99495474-99495474 on Build GRCh38
rs747885973 Chr.14: 99495475-99495477 on Build GRCh38
rs564310227 Chr.14: 99495473-99495473 on Build GRCh38
rs1035880255 Chr.14: 99495838-99495838 on Build GRCh38
rs182575606 Chr.14: 99495870-99495870 on Build GRCh38
rs934382947 Chr.14: 99495782-99495785 on Build GRCh38
rs1002923242 Chr.14: 99495835-99495835 on Build GRCh38
rs571934588 Chr.14: 99495458-99495458 on Build GRCh38
rs772265080 Chr.14: 99495480-99495480 on Build GRCh38
rs143133451 Chr.14: 99495433-99495433 on Build GRCh38
rs571830140 Chr.14: 99495722-99495722 on Build GRCh38
rs368300306 Chr.14: 99495500-99495500 on Build GRCh38
rs190257611 Chr.14: 99495580-99495580 on Build GRCh38
rs755839795 Chr.14: 99495639-99495641 on Build GRCh38
rs201534587 Chr.14: 99495643-99495643 on Build GRCh38
rs567024566 Chr.14: 99495530-99495530 on Build GRCh38
rs748585481 Chr.14: 99495660-99495660 on Build GRCh38
rs755533872 Chr.14: 99495620-99495620 on Build GRCh38
rs2069492 Chr.14: 99495493-99495493 on Build GRCh38
rs769132864 Chr.14: 99495464-99495464 on Build GRCh38
rs764283011 Chr.14: 99495602-99495602 on Build GRCh38
rs376645856 Chr.14: 99495590-99495590 on Build GRCh38
rs757354038 Chr.14: 99495889-99495889 on Build GRCh38
rs992129437 Chr.14: 99495441-99495441 on Build GRCh38
rs551635560 Chr.14: 99495707-99495707 on Build GRCh38
rs766500297 Chr.14: 99495497-99495497 on Build GRCh38
rs565877072 Chr.14: 99495659-99495659 on Build GRCh38
rs978379354 Chr.14: 99495747-99495747 on Build GRCh38
rs200951416 Chr.14: 99495677-99495677 on Build GRCh38
rs755675979 Chr.14: 99495593-99495593 on Build GRCh38
rs751458769 Chr.14: 99495518-99495518 on Build GRCh38
rs528935102 Chr.14: 99495819-99495819 on Build GRCh38
rs368952076 Chr.14: 99495596-99495596 on Build GRCh38
rs147439837 Chr.14: 99495842-99495842 on Build GRCh38
rs935043382 Chr.14: 99495579-99495579 on Build GRCh38
rs1050752455 Chr.14: 99495649-99495649 on Build GRCh38
rs374722616 Chr.14: 99495671-99495671 on Build GRCh38
rs148239455 Chr.14: 99495472-99495472 on Build GRCh38
rs774760059 Chr.14: 99495467-99495467 on Build GRCh38
rs368133103 Chr.14: 99495451-99495451 on Build GRCh38
rs749821472 Chr.14: 99495462-99495462 on Build GRCh38
rs764123517 Chr.14: 99495509-99495509 on Build GRCh38
rs142910964 Chr.14: 99495762-99495762 on Build GRCh38
rs372362013 Chr.14: 99495581-99495581 on Build GRCh38

View Full Product Details