Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99501264-99501770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNK
Entrez Gene ID
8812
Gene Name
cyclin K
Gene Aliases
CPR4
UniGene
Hs.510409
Gene Chromosome Location
Chr.14: 99481402-99511515 on Build GRCh38
Transcripts
NM_001099402.1 XM_005268154.4 XM_011537275.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1371934 c.568A>G p.N190D Chr.14: 99501406-99501406 on Build GRCh38
1300951 c.520G>C p.D174H Chr.14: 99501358-99501358 on Build GRCh38
6623060 c.575+1G>T p.? Chr.14: 99501414-99501414 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1042894771 Chr.14: 99501498-99501498 on Build GRCh38
rs555687081 Chr.14: 99501470-99501470 on Build GRCh38
rs760944464 Chr.14: 99501550-99501550 on Build GRCh38
rs982842489 Chr.14: 99501409-99501409 on Build GRCh38
rs904427767 Chr.14: 99501504-99501504 on Build GRCh38
rs562656983 Chr.14: 99501626-99501626 on Build GRCh38
rs771876941 Chr.14: 99501421-99501421 on Build GRCh38
rs753268667 Chr.14: 99501326-99501326 on Build GRCh38
rs753517300 Chr.14: 99501441-99501441 on Build GRCh38
rs759137082 Chr.14: 99501447-99501447 on Build GRCh38
rs546310610 Chr.14: 99501513-99501513 on Build GRCh38
rs892467744 Chr.14: 99501580-99501580 on Build GRCh38
rs908214268 Chr.14: 99501425-99501425 on Build GRCh38
rs966604104 Chr.14: 99501422-99501422 on Build GRCh38
rs1013640614 Chr.14: 99501588-99501588 on Build GRCh38
rs758021988 Chr.14: 99501671-99501671 on Build GRCh38
rs766091421 Chr.14: 99501437-99501437 on Build GRCh38
rs1002355041 Chr.14: 99501615-99501615 on Build GRCh38
rs574365010 Chr.14: 99501494-99501494 on Build GRCh38
rs542140118 Chr.14: 99501642-99501642 on Build GRCh38
rs746022344 Chr.14: 99501368-99501368 on Build GRCh38
rs761862068 Chr.14: 99501393-99501393 on Build GRCh38
rs778352010 Chr.14: 99501320-99501320 on Build GRCh38
rs778240539 Chr.14: 99501336-99501336 on Build GRCh38
rs747329091 Chr.14: 99501337-99501337 on Build GRCh38
rs199594007 Chr.14: 99501375-99501375 on Build GRCh38
rs539807872 Chr.14: 99501553-99501553 on Build GRCh38
rs921047659 Chr.14: 99501485-99501485 on Build GRCh38
rs781579005 Chr.14: 99501346-99501346 on Build GRCh38
rs538635589 Chr.14: 99501688-99501688 on Build GRCh38
rs576835489 Chr.14: 99501600-99501600 on Build GRCh38
rs751470489 Chr.14: 99501727-99501727 on Build GRCh38
rs766288269 Chr.14: 99501446-99501447 on Build GRCh38
rs895074415 Chr.14: 99501691-99501691 on Build GRCh38
rs2144797 Chr.14: 99501659-99501659 on Build GRCh38
rs553733370 Chr.14: 99501562-99501562 on Build GRCh38
rs1010749505 Chr.14: 99501698-99501698 on Build GRCh38
rs139198946 Chr.14: 99501335-99501335 on Build GRCh38
rs545898021 Chr.14: 99501608-99501608 on Build GRCh38
rs186971252 Chr.14: 99501295-99501295 on Build GRCh38
rs773130579 Chr.14: 99501428-99501428 on Build GRCh38
rs760397544 Chr.14: 99501433-99501433 on Build GRCh38
rs965008049 Chr.14: 99501707-99501707 on Build GRCh38
rs771243618 Chr.14: 99501338-99501338 on Build GRCh38
rs754486460 Chr.14: 99501311-99501311 on Build GRCh38
rs192835632 Chr.14: 99501682-99501682 on Build GRCh38
rs3918088 Chr.14: 99501630-99501630 on Build GRCh38
rs371238020 Chr.14: 99501384-99501384 on Build GRCh38

View Full Product Details