Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99894535-99895037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML1
Entrez Gene ID
2009
Gene Name
echinoderm microtubule associated protein like 1
Gene Aliases
ELP79, EMAP, EMAPL, HuEMAP
UniGene
Hs.12451
Gene Chromosome Location
Chr.14: 99737699-99942060 on Build GRCh38
Transcripts
NM_001008707.1 NM_004434.2 XM_005267397.1 XM_005267398.3 XM_005267399.4 XM_005267400.4 XM_011536540.2 XM_011536541.1 XM_011536542.2 XM_017021074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653748 c.637C>T p.R213C Chr.14: 99894661-99894661 on Build GRCh38
6231526 c.718C>A p.L240I Chr.14: 99894742-99894742 on Build GRCh38
4933141 c.731G>T p.W244L Chr.14: 99894755-99894755 on Build GRCh38
258777 c.673G>C p.D225H Chr.14: 99894697-99894697 on Build GRCh38
7329193 c.638G>A p.R213H Chr.14: 99894662-99894662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759686790 Chr.14: 99894673-99894673 on Build GRCh38
rs770994133 Chr.14: 99894670-99894670 on Build GRCh38
rs774587329 Chr.14: 99894804-99894804 on Build GRCh38
rs574216824 Chr.14: 99894916-99894916 on Build GRCh38
rs769908049 Chr.14: 99894782-99894782 on Build GRCh38
rs1029677311 Chr.14: 99894908-99894908 on Build GRCh38
rs1040081824 Chr.14: 99894850-99894850 on Build GRCh38
rs764402431 Chr.14: 99894682-99894682 on Build GRCh38
rs752060828 Chr.14: 99894692-99894692 on Build GRCh38
rs1014914052 Chr.14: 99894573-99894573 on Build GRCh38
rs749083976 Chr.14: 99894656-99894656 on Build GRCh38
rs371809821 Chr.14: 99894808-99894808 on Build GRCh38
rs778328432 Chr.14: 99894589-99894589 on Build GRCh38
rs779922760 Chr.14: 99894654-99894654 on Build GRCh38
rs143655192 Chr.14: 99894584-99894584 on Build GRCh38
rs936034263 Chr.14: 99894987-99894987 on Build GRCh38
rs768458744 Chr.14: 99894740-99894740 on Build GRCh38
rs773198914 Chr.14: 99894776-99894776 on Build GRCh38
rs752972653 Chr.14: 99894679-99894679 on Build GRCh38
rs755746063 Chr.14: 99894722-99894722 on Build GRCh38
rs771907467 Chr.14: 99894773-99894773 on Build GRCh38
rs756756760 Chr.14: 99894603-99894603 on Build GRCh38
rs759168069 Chr.14: 99894627-99894629 on Build GRCh38
rs1006506096 Chr.14: 99894566-99894566 on Build GRCh38
rs773928137 Chr.14: 99894607-99894607 on Build GRCh38
rs748061411 Chr.14: 99894768-99894768 on Build GRCh38
rs866231714 Chr.14: 99894798-99894798 on Build GRCh38
rs763351218 Chr.14: 99894680-99894680 on Build GRCh38
rs766410926 Chr.14: 99894583-99894583 on Build GRCh38
rs1055155649 Chr.14: 99894638-99894638 on Build GRCh38
rs1035728730 Chr.14: 99895004-99895004 on Build GRCh38
rs144068185 Chr.14: 99894750-99894750 on Build GRCh38
rs1015193722 Chr.14: 99894991-99894991 on Build GRCh38
rs769578207 Chr.14: 99894617-99894617 on Build GRCh38
rs886037937 Chr.14: 99894754-99894754 on Build GRCh38
rs780594449 Chr.14: 99894604-99894604 on Build GRCh38
rs975252 Chr.14: 99894968-99894968 on Build GRCh38
rs976322792 Chr.14: 99894905-99894905 on Build GRCh38
rs1003283607 Chr.14: 99895000-99895000 on Build GRCh38
rs752052142 Chr.14: 99894591-99894591 on Build GRCh38
rs776703881 Chr.14: 99894672-99894672 on Build GRCh38
rs867039760 Chr.14: 99894841-99894841 on Build GRCh38
rs7144394 Chr.14: 99894735-99894735 on Build GRCh38
rs780786090 Chr.14: 99894709-99894709 on Build GRCh38
rs199720899 Chr.14: 99894605-99894605 on Build GRCh38
rs928714320 Chr.14: 99894612-99894612 on Build GRCh38
rs763112095 Chr.14: 99894785-99894785 on Build GRCh38
rs952266154 Chr.14: 99894563-99894563 on Build GRCh38
rs1006453777 Chr.14: 99894986-99894986 on Build GRCh38
rs368700243 Chr.14: 99894778-99894778 on Build GRCh38
rs961870032 Chr.14: 99894859-99894859 on Build GRCh38
rs749951880 Chr.14: 99894719-99894719 on Build GRCh38
rs757722177 Chr.14: 99894705-99894705 on Build GRCh38
rs946619191 Chr.14: 99894772-99894772 on Build GRCh38
rs111225983 Chr.14: 99894965-99894965 on Build GRCh38
rs761687162 Chr.14: 99894665-99894665 on Build GRCh38
rs768385829 Chr.14: 99894659-99894659 on Build GRCh38
rs901584498 Chr.14: 99894895-99894895 on Build GRCh38
rs1015287978 Chr.14: 99894835-99894835 on Build GRCh38
rs754179826 Chr.14: 99894966-99894966 on Build GRCh38
rs779832984 Chr.14: 99894733-99894733 on Build GRCh38
rs374460569 Chr.14: 99894585-99894585 on Build GRCh38
rs775655719 Chr.14: 99894783-99894783 on Build GRCh38
rs577805530 Chr.14: 99894661-99894661 on Build GRCh38
rs764467094 Chr.14: 99894800-99894800 on Build GRCh38
rs897791700 Chr.14: 99894992-99894992 on Build GRCh38

View Full Product Details