Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 100128389-100128898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVL
Entrez Gene ID
51466
Gene Name
Enah/Vasp-like
Gene Aliases
RNB6
UniGene
Hs.125867
Gene Chromosome Location
Chr.14: 99971454-100144236 on Build GRCh38
Transcripts
NM_016337.2 XM_005267749.3 XM_011536828.2 XM_017021363.1 XM_017021364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3493755 c.716G>A p.R239Q Chr.14: 100128747-100128747 on Build GRCh38
1678251 c.680C>T p.A227V Chr.14: 100128711-100128711 on Build GRCh38
1749061 c.528C>A p.S176R Chr.14: 100128559-100128559 on Build GRCh38
4622888 c.590C>T p.T197I Chr.14: 100128621-100128621 on Build GRCh38
1707879 c.569_570CC>TT p.P190L Chr.14: 100128600-100128601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373669362 Chr.14: 100128745-100128745 on Build GRCh38
rs776080759 Chr.14: 100128604-100128606 on Build GRCh38
rs1037080885 Chr.14: 100128828-100128828 on Build GRCh38
rs776930698 Chr.14: 100128613-100128613 on Build GRCh38
rs375445728 Chr.14: 100128597-100128597 on Build GRCh38
rs1001661317 Chr.14: 100128415-100128417 on Build GRCh38
rs200866953 Chr.14: 100128752-100128752 on Build GRCh38
rs553404714 Chr.14: 100128450-100128450 on Build GRCh38
rs780940581 Chr.14: 100128598-100128598 on Build GRCh38
rs755399246 Chr.14: 100128578-100128578 on Build GRCh38
rs762378972 Chr.14: 100128786-100128786 on Build GRCh38
rs769974988 Chr.14: 100128774-100128774 on Build GRCh38
rs755416403 Chr.14: 100128469-100128469 on Build GRCh38
rs1048422636 Chr.14: 100128491-100128491 on Build GRCh38
rs746584136 Chr.14: 100128595-100128595 on Build GRCh38
rs764638056 Chr.14: 100128652-100128652 on Build GRCh38
rs371979522 Chr.14: 100128769-100128769 on Build GRCh38
rs776280776 Chr.14: 100128522-100128522 on Build GRCh38
rs909855551 Chr.14: 100128811-100128811 on Build GRCh38
rs763440319 Chr.14: 100128626-100128626 on Build GRCh38
rs561579418 Chr.14: 100128763-100128763 on Build GRCh38
rs748116903 Chr.14: 100128766-100128766 on Build GRCh38
rs766671985 Chr.14: 100128565-100128565 on Build GRCh38
rs368142656 Chr.14: 100128767-100128767 on Build GRCh38
rs149868896 Chr.14: 100128695-100128695 on Build GRCh38
rs763441126 Chr.14: 100128634-100128637 on Build GRCh38
rs765819981 Chr.14: 100128610-100128616 on Build GRCh38
rs775529615 Chr.14: 100128537-100128537 on Build GRCh38
rs767968647 Chr.14: 100128787-100128787 on Build GRCh38
rs988236591 Chr.14: 100128740-100128740 on Build GRCh38
rs773404009 Chr.14: 100128606-100128606 on Build GRCh38
rs867584550 Chr.14: 100128584-100128584 on Build GRCh38
rs747614488 Chr.14: 100128471-100128471 on Build GRCh38
rs765609851 Chr.14: 100128580-100128580 on Build GRCh38
rs201601257 Chr.14: 100128784-100128784 on Build GRCh38
rs771053611 Chr.14: 100128709-100128709 on Build GRCh38
rs762247179 Chr.14: 100128604-100128605 on Build GRCh38
rs530975071 Chr.14: 100128683-100128683 on Build GRCh38
rs924994799 Chr.14: 100128733-100128733 on Build GRCh38
rs371072374 Chr.14: 100128596-100128596 on Build GRCh38
rs755767739 Chr.14: 100128755-100128755 on Build GRCh38
rs777345841 Chr.14: 100128472-100128472 on Build GRCh38
rs367737727 Chr.14: 100128600-100128600 on Build GRCh38
rs765634073 Chr.14: 100128617-100128617 on Build GRCh38
rs111801307 Chr.14: 100128493-100128493 on Build GRCh38
rs775813248 Chr.14: 100128715-100128715 on Build GRCh38
rs754119420 Chr.14: 100128566-100128566 on Build GRCh38
rs753644754 Chr.14: 100128761-100128761 on Build GRCh38
rs773561817 Chr.14: 100128620-100128629 on Build GRCh38
rs772745657 Chr.14: 100128788-100128788 on Build GRCh38
rs528151175 Chr.14: 100128790-100128790 on Build GRCh38
rs752083896 Chr.14: 100128744-100128744 on Build GRCh38
rs760875516 Chr.14: 100128610-100128610 on Build GRCh38
rs755798736 Chr.14: 100128678-100128678 on Build GRCh38
rs759759324 Chr.14: 100128616-100128616 on Build GRCh38
rs985850682 Chr.14: 100128723-100128723 on Build GRCh38
rs377134346 Chr.14: 100128782-100128782 on Build GRCh38
rs771018080 Chr.14: 100128612-100128612 on Build GRCh38
rs544913047 Chr.14: 100128746-100128746 on Build GRCh38
rs944303942 Chr.14: 100128439-100128439 on Build GRCh38
rs536656592 Chr.14: 100128835-100128835 on Build GRCh38
rs897661635 Chr.14: 100128844-100128844 on Build GRCh38
rs866421043 Chr.14: 100128708-100128708 on Build GRCh38
rs986240086 Chr.14: 100128870-100128870 on Build GRCh38
rs779023969 Chr.14: 100128685-100128685 on Build GRCh38
rs764547953 Chr.14: 100128743-100128743 on Build GRCh38
rs780661759 Chr.14: 100128659-100128659 on Build GRCh38
rs751040383 Chr.14: 100128656-100128656 on Build GRCh38
rs746606863 Chr.14: 100128590-100128590 on Build GRCh38
rs997725403 Chr.14: 100128833-100128833 on Build GRCh38
rs769654642 Chr.14: 100128601-100128601 on Build GRCh38
rs938562646 Chr.14: 100128847-100128847 on Build GRCh38
rs565113043 Chr.14: 100128620-100128620 on Build GRCh38
rs201901077 Chr.14: 100128783-100128783 on Build GRCh38
rs546854320 Chr.14: 100128560-100128560 on Build GRCh38
rs201550659 Chr.14: 100128710-100128710 on Build GRCh38
rs745727356 Chr.14: 100128523-100128523 on Build GRCh38
rs550413526 Chr.14: 100128857-100128857 on Build GRCh38
rs766832916 Chr.14: 100128643-100128643 on Build GRCh38
rs763352727 Chr.14: 100128742-100128742 on Build GRCh38
rs370658826 Chr.14: 100128539-100128539 on Build GRCh38
rs924791775 Chr.14: 100128412-100128412 on Build GRCh38
rs780082762 Chr.14: 100128603-100128603 on Build GRCh38
rs200017971 Chr.14: 100128592-100128592 on Build GRCh38
rs768770640 Chr.14: 100128605-100128605 on Build GRCh38
rs947142811 Chr.14: 100128643-100128643 on Build GRCh38
rs779892342 Chr.14: 100128756-100128756 on Build GRCh38
rs770055323 Chr.14: 100128713-100128713 on Build GRCh38
rs768161285 Chr.14: 100128598-100128598 on Build GRCh38
rs139055077 Chr.14: 100128682-100128682 on Build GRCh38
rs746666025 Chr.14: 100128475-100128475 on Build GRCh38
rs745987828 Chr.14: 100128773-100128773 on Build GRCh38
rs750105789 Chr.14: 100128661-100128661 on Build GRCh38
rs753213998 Chr.14: 100128589-100128589 on Build GRCh38
rs551499169 Chr.14: 100128793-100128793 on Build GRCh38
rs773502273 Chr.14: 100128559-100128559 on Build GRCh38
rs932673075 Chr.14: 100128480-100128480 on Build GRCh38
rs749243042 Chr.14: 100128604-100128604 on Build GRCh38
rs756826325 Chr.14: 100128658-100128658 on Build GRCh38
rs377528297 Chr.14: 100128527-100128527 on Build GRCh38
rs936369985 Chr.14: 100128735-100128735 on Build GRCh38
rs1021228978 Chr.14: 100128704-100128704 on Build GRCh38
rs767719598 Chr.14: 100128546-100128546 on Build GRCh38
rs571146445 Chr.14: 100128827-100128827 on Build GRCh38
rs777257901 Chr.14: 100128594-100128594 on Build GRCh38
rs775182748 Chr.14: 100128589-100128590 on Build GRCh38
rs765065722 Chr.14: 100128866-100128866 on Build GRCh38
rs976989699 Chr.14: 100128440-100128440 on Build GRCh38
rs1003205663 Chr.14: 100128671-100128671 on Build GRCh38
rs952088932 Chr.14: 100128460-100128460 on Build GRCh38

View Full Product Details