Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 101964389-101964910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1H1
Entrez Gene ID
1778
Gene Name
dynein cytoplasmic 1 heavy chain 1
Gene Aliases
CMT2O, DHC1, DHC1a, DNCH1, DNCL, DNECL, DYHC, Dnchc1, HL-3, SMALED1, p22
UniGene
Hs.614080
Gene Chromosome Location
Chr.14: 101964528-102050798 on Build GRCh38
Transcripts
NM_001376.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748709 c.96G>T p.K32N Chr.14: 101964787-101964787 on Build GRCh38
6565826 c.139G>A p.E47K Chr.14: 101964830-101964830 on Build GRCh38
6381095 c.215C>A p.P72Q Chr.14: 101964906-101964906 on Build GRCh38
3689938 c.115C>T p.P39S Chr.14: 101964806-101964806 on Build GRCh38
6214152 c.208T>G p.S70A Chr.14: 101964899-101964899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760083766 Chr.14: 101964769-101964769 on Build GRCh38
rs780018678 Chr.14: 101964834-101964834 on Build GRCh38
rs756016984 Chr.14: 101964833-101964833 on Build GRCh38
rs546390193 Chr.14: 101964532-101964532 on Build GRCh38
rs532206018 Chr.14: 101964606-101964606 on Build GRCh38
rs577413889 Chr.14: 101964886-101964886 on Build GRCh38
rs944435270 Chr.14: 101964597-101964597 on Build GRCh38
rs2273436 Chr.14: 101964639-101964639 on Build GRCh38
rs749252604 Chr.14: 101964850-101964850 on Build GRCh38
rs369550020 Chr.14: 101964689-101964689 on Build GRCh38
rs774412818 Chr.14: 101964713-101964713 on Build GRCh38
rs745720896 Chr.14: 101964825-101964825 on Build GRCh38
rs917189373 Chr.14: 101964458-101964458 on Build GRCh38
rs748270608 Chr.14: 101964721-101964721 on Build GRCh38
rs554497564 Chr.14: 101964874-101964874 on Build GRCh38
rs751157210 Chr.14: 101964809-101964809 on Build GRCh38
rs554577662 Chr.14: 101964680-101964680 on Build GRCh38
rs537857759 Chr.14: 101964679-101964679 on Build GRCh38
rs1033704180 Chr.14: 101964595-101964595 on Build GRCh38
rs372404851 Chr.14: 101964841-101964841 on Build GRCh38
rs772158466 Chr.14: 101964729-101964729 on Build GRCh38
rs773538369 Chr.14: 101964665-101964665 on Build GRCh38
rs769682126 Chr.14: 101964710-101964710 on Build GRCh38
rs903873576 Chr.14: 101964788-101964788 on Build GRCh38
rs757038886 Chr.14: 101964706-101964706 on Build GRCh38
rs17511858 Chr.14: 101964687-101964687 on Build GRCh38
rs529909315 Chr.14: 101964478-101964478 on Build GRCh38
rs940605237 Chr.14: 101964459-101964459 on Build GRCh38
rs777022618 Chr.14: 101964875-101964875 on Build GRCh38
rs1009092704 Chr.14: 101964548-101964548 on Build GRCh38
rs113463845 Chr.14: 101964514-101964514 on Build GRCh38
rs777616661 Chr.14: 101964703-101964703 on Build GRCh38
rs766681689 Chr.14: 101964673-101964673 on Build GRCh38
rs756874444 Chr.14: 101964815-101964815 on Build GRCh38
rs552280450 Chr.14: 101964609-101964609 on Build GRCh38
rs17511851 Chr.14: 101964645-101964645 on Build GRCh38
rs1002259356 Chr.14: 101964594-101964594 on Build GRCh38
rs968044796 Chr.14: 101964591-101964591 on Build GRCh38
rs757934129 Chr.14: 101964791-101964791 on Build GRCh38
rs17540561 Chr.14: 101964599-101964599 on Build GRCh38
rs752231360 Chr.14: 101964784-101964784 on Build GRCh38
rs781032145 Chr.14: 101964820-101964820 on Build GRCh38
rs765882472 Chr.14: 101964774-101964774 on Build GRCh38
rs937150153 Chr.14: 101964412-101964412 on Build GRCh38
rs767796841 Chr.14: 101964660-101964660 on Build GRCh38
rs763730356 Chr.14: 101964796-101964796 on Build GRCh38
rs762013096 Chr.14: 101964650-101964650 on Build GRCh38
rs1043956265 Chr.14: 101964783-101964783 on Build GRCh38
rs2273437 Chr.14: 101964737-101964737 on Build GRCh38
rs780854206 Chr.14: 101964709-101964709 on Build GRCh38
rs1050140788 Chr.14: 101964513-101964513 on Build GRCh38
rs773381038 Chr.14: 101964730-101964730 on Build GRCh38
rs745849309 Chr.14: 101964567-101964569 on Build GRCh38
rs566619022 Chr.14: 101964570-101964570 on Build GRCh38
rs867811283 Chr.14: 101964704-101964704 on Build GRCh38
rs992724811 Chr.14: 101964451-101964451 on Build GRCh38
rs768423968 Chr.14: 101964841-101964842 on Build GRCh38
rs776917333 Chr.14: 101964761-101964761 on Build GRCh38
rs772070566 Chr.14: 101964852-101964852 on Build GRCh38
rs769763690 Chr.14: 101964607-101964619 on Build GRCh38
rs1040549528 Chr.14: 101964551-101964551 on Build GRCh38
rs369058949 Chr.14: 101964799-101964799 on Build GRCh38
rs971971468 Chr.14: 101964608-101964608 on Build GRCh38
rs917082431 Chr.14: 101964605-101964605 on Build GRCh38
rs930396391 Chr.14: 101964506-101964507 on Build GRCh38
rs1040602539 Chr.14: 101964690-101964690 on Build GRCh38
rs564940277 Chr.14: 101964507-101964507 on Build GRCh38
rs373527972 Chr.14: 101964727-101964727 on Build GRCh38
rs935645209 Chr.14: 101964808-101964808 on Build GRCh38
rs892742037 Chr.14: 101964555-101964555 on Build GRCh38
rs766704602 Chr.14: 101964754-101964754 on Build GRCh38
rs761048563 Chr.14: 101964668-101964668 on Build GRCh38
rs917016410 Chr.14: 101964748-101964748 on Build GRCh38
rs17540554 Chr.14: 101964461-101964461 on Build GRCh38
rs960762590 Chr.14: 101964598-101964598 on Build GRCh38
rs886050367 Chr.14: 101964555-101964555 on Build GRCh38
rs1009817478 Chr.14: 101964563-101964563 on Build GRCh38
rs770303875 Chr.14: 101964888-101964888 on Build GRCh38
rs868465219 Chr.14: 101964526-101964526 on Build GRCh38

View Full Product Details