Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102509094-102509632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD9
Entrez Gene ID
122416
Gene Name
ankyrin repeat domain 9
Gene Aliases
-
UniGene
Hs.432945
Gene Chromosome Location
Chr.14: 102503649-102509791 on Build GRCh38
Transcripts
NM_152326.2 XM_005267310.2 XM_005267311.3

SNP Detail

SNP ID Chromosome Location
rs975185839 Chr.14: 102509477-102509477 on Build GRCh38
rs886740067 Chr.14: 102509358-102509358 on Build GRCh38
rs921129748 Chr.14: 102509209-102509209 on Build GRCh38
rs1020233244 Chr.14: 102509277-102509277 on Build GRCh38
rs1029543037 Chr.14: 102509560-102509560 on Build GRCh38
rs555059591 Chr.14: 102509327-102509327 on Build GRCh38
rs199721054 Chr.14: 102509457-102509458 on Build GRCh38
rs113587425 Chr.14: 102509206-102509206 on Build GRCh38
rs893910340 Chr.14: 102509277-102509278 on Build GRCh38
rs950371955 Chr.14: 102509335-102509335 on Build GRCh38
rs1016181673 Chr.14: 102509364-102509364 on Build GRCh38
rs200622845 Chr.14: 102509541-102509541 on Build GRCh38
rs986698738 Chr.14: 102509576-102509576 on Build GRCh38
rs3742445 Chr.14: 102509215-102509215 on Build GRCh38
rs1052491033 Chr.14: 102509266-102509266 on Build GRCh38
rs1053101319 Chr.14: 102509192-102509192 on Build GRCh38
rs962443235 Chr.14: 102509419-102509419 on Build GRCh38
rs1045196475 Chr.14: 102509308-102509308 on Build GRCh38
rs12882912 Chr.14: 102509416-102509416 on Build GRCh38
rs3742446 Chr.14: 102509235-102509235 on Build GRCh38
rs1016666814 Chr.14: 102509408-102509408 on Build GRCh38
rs911887405 Chr.14: 102509236-102509236 on Build GRCh38
rs144538418 Chr.14: 102509123-102509123 on Build GRCh38
rs868645885 Chr.14: 102509440-102509440 on Build GRCh38
rs890078191 Chr.14: 102509239-102509239 on Build GRCh38
rs1005929445 Chr.14: 102509360-102509360 on Build GRCh38
rs938786747 Chr.14: 102509186-102509186 on Build GRCh38
rs924524728 Chr.14: 102509587-102509587 on Build GRCh38
rs979964606 Chr.14: 102509149-102509149 on Build GRCh38
rs1028155966 Chr.14: 102509501-102509501 on Build GRCh38
rs951156172 Chr.14: 102509150-102509150 on Build GRCh38
rs928722795 Chr.14: 102509163-102509163 on Build GRCh38
rs1004305436 Chr.14: 102509371-102509371 on Build GRCh38
rs1034689094 Chr.14: 102509125-102509125 on Build GRCh38
rs1061924 Chr.14: 102509318-102509318 on Build GRCh38
rs901575955 Chr.14: 102509306-102509306 on Build GRCh38
rs1007007007 Chr.14: 102509262-102509262 on Build GRCh38
rs962515317 Chr.14: 102509182-102509182 on Build GRCh38
rs1016697863 Chr.14: 102509173-102509173 on Build GRCh38

View Full Product Details