Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102922575-102923064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN
Entrez Gene ID
81693
Gene Name
amnion associated transmembrane protein
Gene Aliases
PRO1028, amnionless
UniGene
Hs.534494
Gene Chromosome Location
Chr.14: 102922430-102930842 on Build GRCh38
Transcripts
NM_030943.3 XM_011537202.2 XM_011537203.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6243844 c.43G>T p.A15S Chr.14: 102922731-102922731 on Build GRCh38
3667616 c.9C>G p.V3V Chr.14: 102922697-102922697 on Build GRCh38
4627238 c.20T>C p.V7A Chr.14: 102922708-102922708 on Build GRCh38
6463351 c.16_17delCG p.R6fs*43 Chr.14: 102922704-102922705 on Build GRCh38
296889 c.42C>T p.C14C Chr.14: 102922730-102922730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024306768 Chr.14: 102922934-102922934 on Build GRCh38
rs552003719 Chr.14: 102922791-102922791 on Build GRCh38
rs930833237 Chr.14: 102922697-102922697 on Build GRCh38
rs759591055 Chr.14: 102922665-102922665 on Build GRCh38
rs765164769 Chr.14: 102922742-102922742 on Build GRCh38
rs386834172 Chr.14: 102922732-102922732 on Build GRCh38
rs540052204 Chr.14: 102922827-102922827 on Build GRCh38
rs759036156 Chr.14: 102922769-102922769 on Build GRCh38
rs752133468 Chr.14: 102922684-102922684 on Build GRCh38
rs200460988 Chr.14: 102922706-102922706 on Build GRCh38
rs1040721072 Chr.14: 102922896-102922896 on Build GRCh38
rs545740577 Chr.14: 102922986-102922986 on Build GRCh38
rs1009899789 Chr.14: 102922680-102922681 on Build GRCh38
rs888604488 Chr.14: 102922928-102922928 on Build GRCh38
rs532118413 Chr.14: 102922789-102922789 on Build GRCh38
rs745417738 Chr.14: 102922694-102922694 on Build GRCh38
rs376932320 Chr.14: 102922683-102922683 on Build GRCh38
rs948134934 Chr.14: 102922612-102922612 on Build GRCh38
rs768522233 Chr.14: 102922709-102922709 on Build GRCh38
rs568052855 Chr.14: 102922835-102922835 on Build GRCh38
rs533063234 Chr.14: 102923017-102923017 on Build GRCh38
rs889436372 Chr.14: 102922695-102922695 on Build GRCh38
rs764306556 Chr.14: 102922681-102922681 on Build GRCh38
rs916588993 Chr.14: 102922606-102922606 on Build GRCh38
rs538102706 Chr.14: 102922924-102922924 on Build GRCh38
rs776734357 Chr.14: 102922663-102922663 on Build GRCh38
rs755562339 Chr.14: 102922702-102922702 on Build GRCh38
rs57687948 Chr.14: 102922615-102922615 on Build GRCh38
rs753478163 Chr.14: 102922939-102922940 on Build GRCh38
rs1047234748 Chr.14: 102922914-102922914 on Build GRCh38
rs956795833 Chr.14: 102922861-102922861 on Build GRCh38
rs958829979 Chr.14: 102922637-102922637 on Build GRCh38
rs1035031423 Chr.14: 102922961-102922961 on Build GRCh38
rs978651612 Chr.14: 102923003-102923003 on Build GRCh38
rs1043411398 Chr.14: 102922960-102922960 on Build GRCh38
rs746143548 Chr.14: 102922653-102922654 on Build GRCh38
rs112794792 Chr.14: 102922623-102922623 on Build GRCh38
rs772049847 Chr.14: 102922737-102922737 on Build GRCh38
rs747910380 Chr.14: 102922730-102922730 on Build GRCh38
rs969778231 Chr.14: 102922848-102922848 on Build GRCh38
rs560170773 Chr.14: 102922786-102922786 on Build GRCh38
rs959001635 Chr.14: 102922830-102922830 on Build GRCh38
rs900882249 Chr.14: 102922898-102922898 on Build GRCh38
rs143226658 Chr.14: 102922858-102922858 on Build GRCh38
rs892112866 Chr.14: 102922917-102922917 on Build GRCh38
rs769523726 Chr.14: 102922654-102922654 on Build GRCh38
rs1039978266 Chr.14: 102922741-102922741 on Build GRCh38
rs769770182 Chr.14: 102922723-102922723 on Build GRCh38
rs1040134458 Chr.14: 102922916-102922916 on Build GRCh38
rs1041084526 Chr.14: 102923018-102923018 on Build GRCh38
rs745544144 Chr.14: 102922650-102922650 on Build GRCh38
rs59793431 Chr.14: 102922602-102922602 on Build GRCh38
rs767176145 Chr.14: 102922860-102922860 on Build GRCh38
rs2295829 Chr.14: 102922666-102922666 on Build GRCh38
rs756319589 Chr.14: 102922931-102922931 on Build GRCh38
rs562492639 Chr.14: 102922989-102922989 on Build GRCh38
rs540951191 Chr.14: 102922958-102922958 on Build GRCh38
rs538734434 Chr.14: 102922617-102922617 on Build GRCh38
rs752010055 Chr.14: 102922756-102922756 on Build GRCh38
rs560265119 Chr.14: 102922811-102922812 on Build GRCh38
rs761472053 Chr.14: 102922740-102922740 on Build GRCh38
rs1190225 Chr.14: 102922877-102922877 on Build GRCh38
rs531423591 Chr.14: 102923007-102923007 on Build GRCh38
rs181448681 Chr.14: 102922975-102922975 on Build GRCh38
rs772035824 Chr.14: 102923035-102923035 on Build GRCh38
rs911093628 Chr.14: 102923006-102923006 on Build GRCh38
rs1025818394 Chr.14: 102922782-102922782 on Build GRCh38
rs116464826 Chr.14: 102922957-102922957 on Build GRCh38
rs952915295 Chr.14: 102922787-102922787 on Build GRCh38
rs777888037 Chr.14: 102923010-102923010 on Build GRCh38
rs561570970 Chr.14: 102922704-102922704 on Build GRCh38
rs972032697 Chr.14: 102923024-102923024 on Build GRCh38
rs2295828 Chr.14: 102922662-102922662 on Build GRCh38
rs749162280 Chr.14: 102922655-102922655 on Build GRCh38
rs780798513 Chr.14: 102922649-102922649 on Build GRCh38
rs758794765 Chr.14: 102922679-102922679 on Build GRCh38
rs372711546 Chr.14: 102922755-102922755 on Build GRCh38
rs189112558 Chr.14: 102922859-102922859 on Build GRCh38
rs775709395 Chr.14: 102922670-102922672 on Build GRCh38
rs1024302338 Chr.14: 102922857-102922858 on Build GRCh38
rs913086493 Chr.14: 102922871-102922871 on Build GRCh38
rs772396092 Chr.14: 102922660-102922660 on Build GRCh38
rs779387275 Chr.14: 102922776-102922776 on Build GRCh38
rs768328215 Chr.14: 102922656-102922656 on Build GRCh38
rs575646706 Chr.14: 102922893-102922893 on Build GRCh38
rs988171680 Chr.14: 102922976-102922976 on Build GRCh38
rs900952948 Chr.14: 102922757-102922757 on Build GRCh38
rs532186843 Chr.14: 102922887-102922887 on Build GRCh38
rs757797348 Chr.14: 102922685-102922685 on Build GRCh38
rs531412005 Chr.14: 102922811-102922811 on Build GRCh38
rs750958756 Chr.14: 102922765-102922765 on Build GRCh38
rs566547172 Chr.14: 102922862-102922862 on Build GRCh38
rs764845014 Chr.14: 102922744-102922744 on Build GRCh38
rs1022657715 Chr.14: 102922771-102922771 on Build GRCh38
rs1014332872 Chr.14: 102922998-102922998 on Build GRCh38
rs775795140 Chr.14: 102922754-102922754 on Build GRCh38
rs969702078 Chr.14: 102922940-102922940 on Build GRCh38
rs184031634 Chr.14: 102922768-102922768 on Build GRCh38
rs555185838 Chr.14: 102922950-102922950 on Build GRCh38
rs755318316 Chr.14: 102922881-102922881 on Build GRCh38
rs972340659 Chr.14: 102922676-102922676 on Build GRCh38
rs1043346509 Chr.14: 102922878-102922878 on Build GRCh38
rs548366137 Chr.14: 102922823-102922823 on Build GRCh38
rs386834168 Chr.14: 102922702-102922702 on Build GRCh38
rs1002128045 Chr.14: 102922977-102922977 on Build GRCh38
rs554473083 Chr.14: 102922966-102922966 on Build GRCh38
rs114710627 Chr.14: 102922644-102922644 on Build GRCh38
rs927345662 Chr.14: 102922635-102922635 on Build GRCh38
rs575202288 Chr.14: 102922761-102922761 on Build GRCh38
rs369434844 Chr.14: 102922738-102922738 on Build GRCh38
rs562564195 Chr.14: 102922794-102922794 on Build GRCh38
rs993935377 Chr.14: 102922913-102922913 on Build GRCh38

View Full Product Details