Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102923809-102924315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN
Entrez Gene ID
81693
Gene Name
amnion associated transmembrane protein
Gene Aliases
PRO1028, amnionless
UniGene
Hs.534494
Gene Chromosome Location
Chr.14: 102922430-102930842 on Build GRCh38
Transcripts
NM_030943.3 XM_011537202.2 XM_011537203.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3983112 c.207+9G>T p.? Chr.14: 102923988-102923988 on Build GRCh38
5162313 c.193G>A p.A65T Chr.14: 102923965-102923965 on Build GRCh38
6599280 c.194C>T p.A65V Chr.14: 102923966-102923966 on Build GRCh38
6599279 c.152C>T p.P51L Chr.14: 102923819-102923819 on Build GRCh38
6075293 c.163-1G>A p.? Chr.14: 102923934-102923934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571044395 Chr.14: 102924226-102924226 on Build GRCh38
rs532327934 Chr.14: 102924213-102924223 on Build GRCh38
rs1038175710 Chr.14: 102924171-102924171 on Build GRCh38
rs1190226 Chr.14: 102924046-102924046 on Build GRCh38
rs113776350 Chr.14: 102923842-102923842 on Build GRCh38
rs368988450 Chr.14: 102923990-102923990 on Build GRCh38
rs540867109 Chr.14: 102924012-102924012 on Build GRCh38
rs781248844 Chr.14: 102923931-102923931 on Build GRCh38
rs777863436 Chr.14: 102923923-102923923 on Build GRCh38
rs767064693 Chr.14: 102923864-102923865 on Build GRCh38
rs762514501 Chr.14: 102923899-102923899 on Build GRCh38
rs775021246 Chr.14: 102923839-102923839 on Build GRCh38
rs781459651 Chr.14: 102923993-102923993 on Build GRCh38
rs866956966 Chr.14: 102923965-102923965 on Build GRCh38
rs773867275 Chr.14: 102923952-102923952 on Build GRCh38
rs759479529 Chr.14: 102923979-102923979 on Build GRCh38
rs371713937 Chr.14: 102923969-102923969 on Build GRCh38
rs777014561 Chr.14: 102923890-102923890 on Build GRCh38
rs993798264 Chr.14: 102924136-102924136 on Build GRCh38
rs755514962 Chr.14: 102924019-102924020 on Build GRCh38
rs750576347 Chr.14: 102923995-102923995 on Build GRCh38
rs746289915 Chr.14: 102923932-102923932 on Build GRCh38
rs778994887 Chr.14: 102923891-102923891 on Build GRCh38
rs368767424 Chr.14: 102924039-102924039 on Build GRCh38
rs749734745 Chr.14: 102923946-102923946 on Build GRCh38
rs75944538 Chr.14: 102924260-102924260 on Build GRCh38
rs199714929 Chr.14: 102923880-102923880 on Build GRCh38
rs772996972 Chr.14: 102923968-102923968 on Build GRCh38
rs756480860 Chr.14: 102924007-102924007 on Build GRCh38
rs896442195 Chr.14: 102924236-102924236 on Build GRCh38
rs942125544 Chr.14: 102924144-102924144 on Build GRCh38
rs766219642 Chr.14: 102923977-102923977 on Build GRCh38
rs911042300 Chr.14: 102924212-102924212 on Build GRCh38
rs77888366 Chr.14: 102923895-102923895 on Build GRCh38
rs547698885 Chr.14: 102924185-102924185 on Build GRCh38
rs1013243189 Chr.14: 102924019-102924019 on Build GRCh38
rs549220135 Chr.14: 102924283-102924283 on Build GRCh38
rs898308988 Chr.14: 102924179-102924179 on Build GRCh38
rs754018127 Chr.14: 102923844-102923844 on Build GRCh38
rs577736389 Chr.14: 102924050-102924050 on Build GRCh38
rs752265131 Chr.14: 102924017-102924018 on Build GRCh38
rs767280834 Chr.14: 102923854-102923854 on Build GRCh38
rs74082982 Chr.14: 102924136-102924136 on Build GRCh38
rs1024755085 Chr.14: 102924076-102924076 on Build GRCh38
rs563397369 Chr.14: 102924063-102924063 on Build GRCh38
rs933415944 Chr.14: 102924279-102924279 on Build GRCh38
rs144729558 Chr.14: 102923962-102923962 on Build GRCh38
rs780121742 Chr.14: 102923862-102923862 on Build GRCh38
rs201557681 Chr.14: 102923867-102923867 on Build GRCh38
rs1055448287 Chr.14: 102924190-102924190 on Build GRCh38
rs983457775 Chr.14: 102924106-102924106 on Build GRCh38
rs781731359 Chr.14: 102923874-102923874 on Build GRCh38
rs769613083 Chr.14: 102924228-102924228 on Build GRCh38
rs367879875 Chr.14: 102923924-102923924 on Build GRCh38
rs757268891 Chr.14: 102923991-102923991 on Build GRCh38
rs770137078 Chr.14: 102923934-102923934 on Build GRCh38
rs753764931 Chr.14: 102923915-102923915 on Build GRCh38
rs771222631 Chr.14: 102923887-102923887 on Build GRCh38
rs751216705 Chr.14: 102923849-102923849 on Build GRCh38
rs764759870 Chr.14: 102923834-102923834 on Build GRCh38
rs756164471 Chr.14: 102923860-102923860 on Build GRCh38
rs765056352 Chr.14: 102923988-102923988 on Build GRCh38
rs375774640 Chr.14: 102923978-102923978 on Build GRCh38
rs761475340 Chr.14: 102923909-102923909 on Build GRCh38
rs766494238 Chr.14: 102923911-102923911 on Build GRCh38
rs767244412 Chr.14: 102923910-102923910 on Build GRCh38
rs773033666 Chr.14: 102924241-102924241 on Build GRCh38
rs1047826818 Chr.14: 102923908-102923908 on Build GRCh38
rs531742126 Chr.14: 102924064-102924064 on Build GRCh38
rs61731158 Chr.14: 102923937-102923937 on Build GRCh38
rs1024639553 Chr.14: 102923906-102923906 on Build GRCh38
rs769243461 Chr.14: 102924014-102924014 on Build GRCh38
rs758550140 Chr.14: 102923877-102923877 on Build GRCh38
rs2749514 Chr.14: 102924184-102924184 on Build GRCh38
rs535042576 Chr.14: 102923903-102923903 on Build GRCh38
rs757005037 Chr.14: 102923851-102923851 on Build GRCh38
rs933306677 Chr.14: 102924102-102924102 on Build GRCh38
rs548601484 Chr.14: 102924097-102924097 on Build GRCh38
rs868608694 Chr.14: 102924192-102924192 on Build GRCh38
rs758423517 Chr.14: 102923922-102923922 on Build GRCh38
rs771819877 Chr.14: 102923964-102923964 on Build GRCh38
rs747938509 Chr.14: 102924067-102924067 on Build GRCh38
rs757540694 Chr.14: 102923929-102923929 on Build GRCh38
rs2749513 Chr.14: 102924051-102924051 on Build GRCh38
rs750347092 Chr.14: 102923856-102923856 on Build GRCh38
rs777940681 Chr.14: 102923841-102923841 on Build GRCh38
rs199629027 Chr.14: 102923947-102923947 on Build GRCh38
rs768142079 Chr.14: 102923900-102923900 on Build GRCh38
rs923327834 Chr.14: 102924263-102924263 on Build GRCh38
rs747197642 Chr.14: 102923884-102923884 on Build GRCh38
rs779277289 Chr.14: 102924022-102924022 on Build GRCh38
rs182043665 Chr.14: 102924062-102924062 on Build GRCh38

View Full Product Details