Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102932682-102933181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPB
Entrez Gene ID
9578
Gene Name
CDC42 binding protein kinase beta
Gene Aliases
MRCKB
UniGene
Hs.654634
Gene Chromosome Location
Chr.14: 102932379-103057405 on Build GRCh38
Transcripts
NM_006035.3 XM_005268227.1 XM_005268228.1 XM_005268229.1

SNP Detail

SNP ID Chromosome Location
rs533551183 Chr.14: 102932787-102932787 on Build GRCh38
rs902148713 Chr.14: 102932796-102932796 on Build GRCh38
rs530158794 Chr.14: 102933095-102933095 on Build GRCh38
rs35903000 Chr.14: 102932897-102932897 on Build GRCh38
rs146914181 Chr.14: 102932902-102932906 on Build GRCh38
rs960663636 Chr.14: 102932850-102932850 on Build GRCh38
rs1020956100 Chr.14: 102933070-102933070 on Build GRCh38
rs933627467 Chr.14: 102932975-102932975 on Build GRCh38
rs11442 Chr.14: 102933141-102933141 on Build GRCh38
rs1004313922 Chr.14: 102932826-102932826 on Build GRCh38
rs895062523 Chr.14: 102932836-102932836 on Build GRCh38
rs3203840 Chr.14: 102932966-102932966 on Build GRCh38
rs966768175 Chr.14: 102932929-102932929 on Build GRCh38
rs886872321 Chr.14: 102932797-102932797 on Build GRCh38
rs964945324 Chr.14: 102933122-102933122 on Build GRCh38
rs542119219 Chr.14: 102933047-102933047 on Build GRCh38
rs1000959546 Chr.14: 102932822-102932822 on Build GRCh38
rs943518321 Chr.14: 102932748-102932748 on Build GRCh38
rs780169060 Chr.14: 102932767-102932767 on Build GRCh38
rs896761597 Chr.14: 102933033-102933033 on Build GRCh38
rs923463172 Chr.14: 102933006-102933006 on Build GRCh38
rs886559399 Chr.14: 102933117-102933117 on Build GRCh38
rs35675176 Chr.14: 102932865-102932865 on Build GRCh38
rs4708 Chr.14: 102933111-102933111 on Build GRCh38
rs925802925 Chr.14: 102932933-102932933 on Build GRCh38
rs938169338 Chr.14: 102933156-102933156 on Build GRCh38
rs985029233 Chr.14: 102932715-102932715 on Build GRCh38
rs975316378 Chr.14: 102932952-102932952 on Build GRCh38
rs1034350164 Chr.14: 102932848-102932848 on Build GRCh38
rs1013870306 Chr.14: 102933050-102933050 on Build GRCh38
rs965000786 Chr.14: 102932990-102932990 on Build GRCh38
rs939686375 Chr.14: 102933127-102933127 on Build GRCh38
rs935922083 Chr.14: 102932760-102932760 on Build GRCh38
rs748114936 Chr.14: 102932928-102932928 on Build GRCh38
rs552658031 Chr.14: 102933010-102933010 on Build GRCh38
rs1002793503 Chr.14: 102933011-102933011 on Build GRCh38
rs897797262 Chr.14: 102932901-102932901 on Build GRCh38
rs373450892 Chr.14: 102933092-102933092 on Build GRCh38
rs538561316 Chr.14: 102932761-102932761 on Build GRCh38
rs898686530 Chr.14: 102932841-102932841 on Build GRCh38
rs921157061 Chr.14: 102933104-102933104 on Build GRCh38
rs1002940412 Chr.14: 102932876-102932876 on Build GRCh38
rs1017083306 Chr.14: 102932833-102932833 on Build GRCh38
rs35864966 Chr.14: 102932906-102932906 on Build GRCh38
rs1002458583 Chr.14: 102932847-102932847 on Build GRCh38
rs908208943 Chr.14: 102933123-102933123 on Build GRCh38
rs562363379 Chr.14: 102932989-102932989 on Build GRCh38
rs575796613 Chr.14: 102932991-102932991 on Build GRCh38
rs971434156 Chr.14: 102932874-102932875 on Build GRCh38
rs35042334 Chr.14: 102933147-102933147 on Build GRCh38
rs35056762 Chr.14: 102932771-102932771 on Build GRCh38
rs925804004 Chr.14: 102932756-102932756 on Build GRCh38
rs1053669269 Chr.14: 102932834-102932834 on Build GRCh38
rs1033907784 Chr.14: 102933032-102933032 on Build GRCh38
rs192976483 Chr.14: 102932739-102932739 on Build GRCh38
rs796271130 Chr.14: 102932925-102932925 on Build GRCh38
rs1042062911 Chr.14: 102932788-102932788 on Build GRCh38
rs935870925 Chr.14: 102932938-102932938 on Build GRCh38
rs559641501 Chr.14: 102932710-102932710 on Build GRCh38
rs933658674 Chr.14: 102932781-102932781 on Build GRCh38
rs561789318 Chr.14: 102933088-102933088 on Build GRCh38
rs921185901 Chr.14: 102932974-102932974 on Build GRCh38
rs1038089906 Chr.14: 102933140-102933140 on Build GRCh38
rs1055686186 Chr.14: 102933157-102933157 on Build GRCh38
rs977273550 Chr.14: 102932749-102932749 on Build GRCh38
rs1050735871 Chr.14: 102932984-102932984 on Build GRCh38
rs974962889 Chr.14: 102933124-102933124 on Build GRCh38
rs1035740133 Chr.14: 102933014-102933014 on Build GRCh38
rs1000953552 Chr.14: 102933090-102933090 on Build GRCh38
rs553872308 Chr.14: 102932794-102932794 on Build GRCh38
rs979955040 Chr.14: 102932931-102932931 on Build GRCh38
rs986707955 Chr.14: 102933119-102933119 on Build GRCh38
rs945836641 Chr.14: 102932743-102932743 on Build GRCh38
rs1039255566 Chr.14: 102932759-102932759 on Build GRCh38
rs574377072 Chr.14: 102932780-102932780 on Build GRCh38
rs909460047 Chr.14: 102932745-102932745 on Build GRCh38
rs960408162 Chr.14: 102933043-102933043 on Build GRCh38
rs977846231 Chr.14: 102933031-102933031 on Build GRCh38
rs1053897298 Chr.14: 102933056-102933056 on Build GRCh38
rs28715432 Chr.14: 102932896-102932896 on Build GRCh38
rs1038567328 Chr.14: 102932998-102932998 on Build GRCh38
rs1006475220 Chr.14: 102932887-102932887 on Build GRCh38
rs957191107 Chr.14: 102933093-102933093 on Build GRCh38
rs149875632 Chr.14: 102933106-102933106 on Build GRCh38
rs184599136 Chr.14: 102932817-102932817 on Build GRCh38

View Full Product Details