Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102939524-102940026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42BPB
Entrez Gene ID
9578
Gene Name
CDC42 binding protein kinase beta
Gene Aliases
MRCKB
UniGene
Hs.654634
Gene Chromosome Location
Chr.14: 102932379-103057405 on Build GRCh38
Transcripts
NM_006035.3 XM_005268227.1 XM_005268228.1 XM_005268229.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5893757 c.4619C>T p.S1540F Chr.14: 102939920-102939920 on Build GRCh38
2025250 c.4668C>T p.F1556F Chr.14: 102939871-102939871 on Build GRCh38
6624387 c.4621G>A p.D1541N Chr.14: 102939918-102939918 on Build GRCh38
5522346 c.4807G>A p.V1603M Chr.14: 102939630-102939630 on Build GRCh38
1368380 c.4708C>T p.R1570* Chr.14: 102939831-102939831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202016085 Chr.14: 102939944-102939944 on Build GRCh38
rs779185138 Chr.14: 102939818-102939818 on Build GRCh38
rs758971557 Chr.14: 102939822-102939822 on Build GRCh38
rs751934399 Chr.14: 102939978-102939978 on Build GRCh38
rs370794007 Chr.14: 102939980-102939980 on Build GRCh38
rs574304884 Chr.14: 102939976-102939976 on Build GRCh38
rs775479309 Chr.14: 102939891-102939891 on Build GRCh38
rs774459084 Chr.14: 102939955-102939955 on Build GRCh38
rs762832845 Chr.14: 102939898-102939898 on Build GRCh38
rs201479761 Chr.14: 102939966-102939966 on Build GRCh38
rs778755078 Chr.14: 102939582-102939585 on Build GRCh38
rs17101081 Chr.14: 102939738-102939738 on Build GRCh38
rs373717720 Chr.14: 102939970-102939970 on Build GRCh38
rs769269988 Chr.14: 102939596-102939596 on Build GRCh38
rs772535709 Chr.14: 102939753-102939753 on Build GRCh38
rs771717134 Chr.14: 102939958-102939959 on Build GRCh38
rs201017984 Chr.14: 102939705-102939705 on Build GRCh38
rs775397673 Chr.14: 102939788-102939788 on Build GRCh38
rs769470267 Chr.14: 102939874-102939874 on Build GRCh38
rs563312788 Chr.14: 102939557-102939557 on Build GRCh38
rs371308106 Chr.14: 102939763-102939763 on Build GRCh38
rs928663657 Chr.14: 102940001-102940001 on Build GRCh38
rs774652544 Chr.14: 102939598-102939598 on Build GRCh38
rs140004721 Chr.14: 102939830-102939830 on Build GRCh38
rs754647455 Chr.14: 102939999-102939999 on Build GRCh38
rs201739098 Chr.14: 102939599-102939599 on Build GRCh38
rs765460474 Chr.14: 102939967-102939967 on Build GRCh38
rs774370821 Chr.14: 102939911-102939911 on Build GRCh38
rs368182073 Chr.14: 102939783-102939783 on Build GRCh38
rs572199745 Chr.14: 102939606-102939606 on Build GRCh38
rs750593843 Chr.14: 102939802-102939802 on Build GRCh38
rs757447340 Chr.14: 102939642-102939642 on Build GRCh38
rs745550984 Chr.14: 102939871-102939871 on Build GRCh38
rs752024361 Chr.14: 102939929-102939929 on Build GRCh38
rs748911420 Chr.14: 102939998-102939998 on Build GRCh38
rs781591159 Chr.14: 102939575-102939575 on Build GRCh38
rs748499505 Chr.14: 102939821-102939821 on Build GRCh38
rs909183338 Chr.14: 102939664-102939664 on Build GRCh38
rs373755648 Chr.14: 102939875-102939875 on Build GRCh38
rs60475222 Chr.14: 102939832-102939832 on Build GRCh38
rs766329158 Chr.14: 102939917-102939917 on Build GRCh38
rs116889152 Chr.14: 102939592-102939592 on Build GRCh38
rs188937393 Chr.14: 102939811-102939811 on Build GRCh38
rs373046706 Chr.14: 102939981-102939981 on Build GRCh38
rs765442114 Chr.14: 102939923-102939923 on Build GRCh38
rs139297022 Chr.14: 102939817-102939818 on Build GRCh38
rs755631432 Chr.14: 102939983-102939983 on Build GRCh38
rs776437116 Chr.14: 102939922-102939922 on Build GRCh38
rs764484900 Chr.14: 102939973-102939973 on Build GRCh38
rs148066338 Chr.14: 102939727-102939727 on Build GRCh38
rs779495697 Chr.14: 102939751-102939751 on Build GRCh38
rs368319342 Chr.14: 102939794-102939794 on Build GRCh38
rs765236814 Chr.14: 102939621-102939621 on Build GRCh38
rs745958701 Chr.14: 102939757-102939757 on Build GRCh38
rs563814555 Chr.14: 102939928-102939928 on Build GRCh38
rs759399853 Chr.14: 102939772-102939772 on Build GRCh38
rs772766652 Chr.14: 102939756-102939756 on Build GRCh38
rs770300795 Chr.14: 102939863-102939863 on Build GRCh38
rs749598805 Chr.14: 102939594-102939594 on Build GRCh38
rs772024223 Chr.14: 102939963-102939963 on Build GRCh38
rs775335460 Chr.14: 102939960-102939961 on Build GRCh38
rs760828996 Chr.14: 102939916-102939916 on Build GRCh38
rs770113836 Chr.14: 102939581-102939581 on Build GRCh38
rs772986036 Chr.14: 102939953-102939953 on Build GRCh38
rs374818759 Chr.14: 102939770-102939770 on Build GRCh38
rs542788488 Chr.14: 102939580-102939580 on Build GRCh38
rs753065038 Chr.14: 102939925-102939925 on Build GRCh38
rs771520786 Chr.14: 102939600-102939600 on Build GRCh38
rs369556425 Chr.14: 102939964-102939964 on Build GRCh38
rs1009900223 Chr.14: 102939918-102939918 on Build GRCh38
rs780932163 Chr.14: 102939870-102939870 on Build GRCh38
rs544013491 Chr.14: 102939915-102939915 on Build GRCh38
rs370955304 Chr.14: 102939558-102939558 on Build GRCh38
rs767750407 Chr.14: 102939654-102939654 on Build GRCh38
rs768715462 Chr.14: 102939900-102939900 on Build GRCh38
rs978643502 Chr.14: 102939683-102939683 on Build GRCh38
rs375217140 Chr.14: 102939732-102939732 on Build GRCh38
rs757357137 Chr.14: 102939564-102939564 on Build GRCh38
rs377554520 Chr.14: 102939972-102939972 on Build GRCh38
rs763037873 Chr.14: 102939792-102939792 on Build GRCh38
rs768519250 Chr.14: 102939946-102939946 on Build GRCh38
rs751666492 Chr.14: 102939563-102939563 on Build GRCh38
rs760369636 Chr.14: 102939601-102939601 on Build GRCh38
rs749106487 Chr.14: 102939945-102939945 on Build GRCh38
rs759726532 Chr.14: 102939920-102939920 on Build GRCh38
rs150003464 Chr.14: 102939718-102939718 on Build GRCh38
rs200207520 Chr.14: 102939752-102939752 on Build GRCh38
rs755400950 Chr.14: 102939817-102939817 on Build GRCh38
rs774624337 Chr.14: 102939742-102939742 on Build GRCh38
rs778901502 Chr.14: 102940003-102940003 on Build GRCh38
rs373428218 Chr.14: 102939812-102939812 on Build GRCh38
rs745708875 Chr.14: 102939779-102939780 on Build GRCh38
rs758705817 Chr.14: 102939926-102939926 on Build GRCh38
rs150855245 Chr.14: 102939640-102939640 on Build GRCh38
rs1019328882 Chr.14: 102939886-102939886 on Build GRCh38
rs761695164 Chr.14: 102939962-102939962 on Build GRCh38
rs753957301 Chr.14: 102939816-102939816 on Build GRCh38
rs139323146 Chr.14: 102939714-102939714 on Build GRCh38
rs202209179 Chr.14: 102939933-102939933 on Build GRCh38
rs200373977 Chr.14: 102939996-102939996 on Build GRCh38
rs759189098 Chr.14: 102939620-102939620 on Build GRCh38
rs745374413 Chr.14: 102939935-102939935 on Build GRCh38
rs776557594 Chr.14: 102939607-102939607 on Build GRCh38
rs753922027 Chr.14: 102939919-102939919 on Build GRCh38
rs759594063 Chr.14: 102939965-102939965 on Build GRCh38

View Full Product Details