Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103102296-103102783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L4
Entrez Gene ID
91828
Gene Name
exocyst complex component 3 like 4
Gene Aliases
C14orf73
UniGene
Hs.37712
Gene Chromosome Location
Chr.14: 103094202-103110562 on Build GRCh38
Transcripts
NM_001077594.1 XM_011537323.2 XM_011537324.2 XM_011537325.2 XM_011537327.2 XM_011537328.2 XM_011537329.2 XM_011537330.2 XM_011537331.1 XM_011537332.2 XM_011537333.2 XM_011537334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5764695 c.990C>T p.D330D Chr.14: 103102713-103102713 on Build GRCh38
4657457 c.840C>G p.A280A Chr.14: 103102563-103102563 on Build GRCh38
6657144 c.703C>A p.L235M Chr.14: 103102426-103102426 on Build GRCh38
5549510 c.956C>A p.A319D Chr.14: 103102679-103102679 on Build GRCh38
3749260 c.921A>C p.P307P Chr.14: 103102644-103102644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182654404 Chr.14: 103102467-103102467 on Build GRCh38
rs779801856 Chr.14: 103102314-103102314 on Build GRCh38
rs570284843 Chr.14: 103102603-103102603 on Build GRCh38
rs1017107033 Chr.14: 103102641-103102641 on Build GRCh38
rs549913822 Chr.14: 103102541-103102541 on Build GRCh38
rs543367128 Chr.14: 103102381-103102381 on Build GRCh38
rs35565431 Chr.14: 103102517-103102517 on Build GRCh38
rs959545655 Chr.14: 103102665-103102665 on Build GRCh38
rs919870195 Chr.14: 103102664-103102664 on Build GRCh38
rs768949981 Chr.14: 103102617-103102617 on Build GRCh38
rs900738164 Chr.14: 103102512-103102512 on Build GRCh38
rs777868304 Chr.14: 103102697-103102697 on Build GRCh38
rs763424509 Chr.14: 103102415-103102415 on Build GRCh38
rs752048300 Chr.14: 103102379-103102379 on Build GRCh38
rs756330078 Chr.14: 103102716-103102716 on Build GRCh38
rs760179539 Chr.14: 103102635-103102635 on Build GRCh38
rs758329486 Chr.14: 103102555-103102555 on Build GRCh38
rs752630387 Chr.14: 103102554-103102554 on Build GRCh38
rs1039404777 Chr.14: 103102363-103102363 on Build GRCh38
rs770174979 Chr.14: 103102412-103102412 on Build GRCh38
rs749262903 Chr.14: 103102315-103102315 on Build GRCh38
rs780428527 Chr.14: 103102614-103102614 on Build GRCh38
rs369448990 Chr.14: 103102654-103102654 on Build GRCh38
rs780476836 Chr.14: 103102560-103102563 on Build GRCh38
rs749512841 Chr.14: 103102746-103102746 on Build GRCh38
rs778716701 Chr.14: 103102405-103102405 on Build GRCh38
rs765749804 Chr.14: 103102368-103102368 on Build GRCh38
rs529560631 Chr.14: 103102611-103102611 on Build GRCh38
rs769939065 Chr.14: 103102613-103102613 on Build GRCh38
rs762042524 Chr.14: 103102621-103102621 on Build GRCh38
rs753418981 Chr.14: 103102456-103102456 on Build GRCh38
rs373971768 Chr.14: 103102691-103102691 on Build GRCh38
rs770979269 Chr.14: 103102409-103102409 on Build GRCh38
rs934288256 Chr.14: 103102726-103102726 on Build GRCh38
rs1035039389 Chr.14: 103102627-103102627 on Build GRCh38
rs755258284 Chr.14: 103102761-103102761 on Build GRCh38
rs541514251 Chr.14: 103102435-103102435 on Build GRCh38
rs924125667 Chr.14: 103102710-103102710 on Build GRCh38
rs1042132333 Chr.14: 103102704-103102704 on Build GRCh38
rs990541835 Chr.14: 103102676-103102676 on Build GRCh38
rs1053949729 Chr.14: 103102725-103102725 on Build GRCh38
rs769108922 Chr.14: 103102720-103102726 on Build GRCh38
rs996792137 Chr.14: 103102755-103102755 on Build GRCh38
rs371500721 Chr.14: 103102619-103102619 on Build GRCh38
rs764984920 Chr.14: 103102685-103102685 on Build GRCh38
rs1042583251 Chr.14: 103102482-103102482 on Build GRCh38
rs10142200 Chr.14: 103102392-103102392 on Build GRCh38
rs139497928 Chr.14: 103102616-103102616 on Build GRCh38
rs914491676 Chr.14: 103102728-103102728 on Build GRCh38
rs867290580 Chr.14: 103102399-103102399 on Build GRCh38
rs996225502 Chr.14: 103102591-103102591 on Build GRCh38
rs983407806 Chr.14: 103102496-103102496 on Build GRCh38
rs754741864 Chr.14: 103102400-103102400 on Build GRCh38
rs921003472 Chr.14: 103102525-103102525 on Build GRCh38
rs751608160 Chr.14: 103102449-103102449 on Build GRCh38
rs764533102 Chr.14: 103102374-103102374 on Build GRCh38
rs199896285 Chr.14: 103102339-103102339 on Build GRCh38
rs78542708 Chr.14: 103102471-103102471 on Build GRCh38
rs776826779 Chr.14: 103102728-103102728 on Build GRCh38
rs1004228874 Chr.14: 103102554-103102562 on Build GRCh38
rs773648920 Chr.14: 103102629-103102629 on Build GRCh38
rs745371400 Chr.14: 103102392-103102393 on Build GRCh38
rs1023694216 Chr.14: 103102420-103102420 on Build GRCh38
rs769966284 Chr.14: 103102437-103102437 on Build GRCh38
rs948560309 Chr.14: 103102744-103102744 on Build GRCh38
rs778951767 Chr.14: 103102762-103102762 on Build GRCh38
rs887081848 Chr.14: 103102631-103102631 on Build GRCh38
rs372851552 Chr.14: 103102317-103102317 on Build GRCh38
rs192143764 Chr.14: 103102359-103102359 on Build GRCh38
rs768609520 Chr.14: 103102318-103102318 on Build GRCh38
rs563565267 Chr.14: 103102382-103102382 on Build GRCh38
rs77071436 Chr.14: 103102644-103102644 on Build GRCh38
rs902650716 Chr.14: 103102499-103102499 on Build GRCh38
rs763372631 Chr.14: 103102370-103102370 on Build GRCh38
rs758546204 Chr.14: 103102344-103102344 on Build GRCh38
rs750124863 Chr.14: 103102384-103102384 on Build GRCh38
rs752571642 Chr.14: 103102690-103102690 on Build GRCh38
rs772653678 Chr.14: 103102628-103102628 on Build GRCh38
rs767039907 Chr.14: 103102383-103102383 on Build GRCh38
rs975140244 Chr.14: 103102504-103102504 on Build GRCh38
rs775808393 Chr.14: 103102414-103102414 on Build GRCh38
rs573862329 Chr.14: 103102390-103102390 on Build GRCh38
rs759209408 Chr.14: 103102677-103102677 on Build GRCh38
rs762358612 Chr.14: 103102429-103102429 on Build GRCh38
rs934614512 Chr.14: 103102588-103102588 on Build GRCh38
rs933696477 Chr.14: 103102551-103102551 on Build GRCh38
rs377044258 Chr.14: 103102720-103102720 on Build GRCh38
rs760128226 Chr.14: 103102442-103102442 on Build GRCh38
rs751477109 Chr.14: 103102702-103102702 on Build GRCh38
rs778553720 Chr.14: 103102346-103102347 on Build GRCh38
rs533328127 Chr.14: 103102520-103102520 on Build GRCh38
rs755730426 Chr.14: 103102389-103102389 on Build GRCh38
rs1004470558 Chr.14: 103102636-103102636 on Build GRCh38
rs772584540 Chr.14: 103102519-103102520 on Build GRCh38
rs1032572104 Chr.14: 103102503-103102503 on Build GRCh38
rs1028058552 Chr.14: 103102680-103102680 on Build GRCh38
rs9324055 Chr.14: 103102612-103102612 on Build GRCh38
rs924242897 Chr.14: 103102562-103102562 on Build GRCh38
rs936848333 Chr.14: 103102719-103102719 on Build GRCh38
rs774787403 Chr.14: 103102424-103102424 on Build GRCh38
rs747409697 Chr.14: 103102630-103102631 on Build GRCh38
rs202097758 Chr.14: 103102369-103102369 on Build GRCh38
rs200540457 Chr.14: 103102727-103102727 on Build GRCh38
rs537010809 Chr.14: 103102732-103102732 on Build GRCh38
rs528327540 Chr.14: 103102416-103102416 on Build GRCh38
rs896273218 Chr.14: 103102464-103102464 on Build GRCh38
rs1001131814 Chr.14: 103102502-103102502 on Build GRCh38
rs777816991 Chr.14: 103102610-103102610 on Build GRCh38
rs549586706 Chr.14: 103102436-103102436 on Build GRCh38
rs776780047 Chr.14: 103102350-103102350 on Build GRCh38
rs774292899 Chr.14: 103102322-103102322 on Build GRCh38

View Full Product Details