Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103108315-103108817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L4
Entrez Gene ID
91828
Gene Name
exocyst complex component 3 like 4
Gene Aliases
C14orf73
UniGene
Hs.37712
Gene Chromosome Location
Chr.14: 103094202-103110562 on Build GRCh38
Transcripts
NM_001077594.1 XM_011537323.2 XM_011537324.2 XM_011537325.2 XM_011537327.2 XM_011537328.2 XM_011537329.2 XM_011537330.2 XM_011537331.1 XM_011537332.2 XM_011537333.2 XM_011537334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3772879 c.1865C>T p.A622V Chr.14: 103108406-103108406 on Build GRCh38
1300409 c.1927G>T p.D643Y Chr.14: 103108468-103108468 on Build GRCh38
4647745 c.1855G>A p.G619S Chr.14: 103108396-103108396 on Build GRCh38
2025368 c.1912G>A p.E638K Chr.14: 103108453-103108453 on Build GRCh38
6657139 c.1969G>A p.D657N Chr.14: 103108510-103108510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970894365 Chr.14: 103108468-103108468 on Build GRCh38
rs763741958 Chr.14: 103108449-103108449 on Build GRCh38
rs573105103 Chr.14: 103108734-103108734 on Build GRCh38
rs761252650 Chr.14: 103108551-103108551 on Build GRCh38
rs768198090 Chr.14: 103108547-103108547 on Build GRCh38
rs142206397 Chr.14: 103108774-103108774 on Build GRCh38
rs760055414 Chr.14: 103108434-103108434 on Build GRCh38
rs575293675 Chr.14: 103108681-103108681 on Build GRCh38
rs755794707 Chr.14: 103108510-103108510 on Build GRCh38
rs754387947 Chr.14: 103108361-103108361 on Build GRCh38
rs895068584 Chr.14: 103108647-103108647 on Build GRCh38
rs41301467 Chr.14: 103108631-103108631 on Build GRCh38
rs747551119 Chr.14: 103108562-103108562 on Build GRCh38
rs766998057 Chr.14: 103108357-103108357 on Build GRCh38
rs777358465 Chr.14: 103108371-103108371 on Build GRCh38
rs1046112073 Chr.14: 103108655-103108655 on Build GRCh38
rs755773836 Chr.14: 103108652-103108652 on Build GRCh38
rs912768185 Chr.14: 103108744-103108744 on Build GRCh38
rs752275463 Chr.14: 103108437-103108437 on Build GRCh38
rs762591338 Chr.14: 103108493-103108493 on Build GRCh38
rs370314474 Chr.14: 103108392-103108392 on Build GRCh38
rs373450066 Chr.14: 103108344-103108344 on Build GRCh38
rs1035489668 Chr.14: 103108617-103108617 on Build GRCh38
rs759023360 Chr.14: 103108362-103108362 on Build GRCh38
rs148718670 Chr.14: 103108478-103108478 on Build GRCh38
rs772142668 Chr.14: 103108347-103108347 on Build GRCh38
rs776126370 Chr.14: 103108539-103108539 on Build GRCh38
rs756097471 Chr.14: 103108466-103108466 on Build GRCh38
rs761184509 Chr.14: 103108422-103108422 on Build GRCh38
rs556935029 Chr.14: 103108684-103108684 on Build GRCh38
rs536595646 Chr.14: 103108721-103108721 on Build GRCh38
rs772349414 Chr.14: 103108402-103108402 on Build GRCh38
rs749739825 Chr.14: 103108541-103108541 on Build GRCh38
rs770348480 Chr.14: 103108484-103108484 on Build GRCh38
rs947920194 Chr.14: 103108649-103108649 on Build GRCh38
rs757574274 Chr.14: 103108518-103108518 on Build GRCh38
rs763584877 Chr.14: 103108495-103108495 on Build GRCh38
rs370767688 Chr.14: 103108351-103108351 on Build GRCh38
rs2274685 Chr.14: 103108733-103108733 on Build GRCh38
rs751213812 Chr.14: 103108452-103108452 on Build GRCh38
rs376043981 Chr.14: 103108393-103108393 on Build GRCh38
rs781176657 Chr.14: 103108391-103108391 on Build GRCh38
rs549171249 Chr.14: 103108477-103108477 on Build GRCh38
rs763591813 Chr.14: 103108716-103108716 on Build GRCh38
rs775795456 Chr.14: 103108486-103108486 on Build GRCh38
rs987659318 Chr.14: 103108775-103108775 on Build GRCh38
rs777968401 Chr.14: 103108515-103108515 on Build GRCh38
rs372356744 Chr.14: 103108461-103108461 on Build GRCh38
rs771407866 Chr.14: 103108427-103108427 on Build GRCh38
rs762312566 Chr.14: 103108447-103108447 on Build GRCh38
rs41304367 Chr.14: 103108548-103108548 on Build GRCh38
rs961704395 Chr.14: 103108675-103108675 on Build GRCh38
rs113602008 Chr.14: 103108733-103108733 on Build GRCh38
rs766402144 Chr.14: 103108512-103108512 on Build GRCh38
rs767118564 Chr.14: 103108503-103108503 on Build GRCh38
rs867137674 Chr.14: 103108499-103108499 on Build GRCh38
rs756979257 Chr.14: 103108378-103108378 on Build GRCh38
rs191971953 Chr.14: 103108706-103108706 on Build GRCh38
rs377097077 Chr.14: 103108513-103108513 on Build GRCh38
rs142666277 Chr.14: 103108435-103108435 on Build GRCh38
rs199536153 Chr.14: 103108509-103108509 on Build GRCh38
rs920663239 Chr.14: 103108604-103108604 on Build GRCh38
rs1013662737 Chr.14: 103108643-103108643 on Build GRCh38
rs912693085 Chr.14: 103108504-103108504 on Build GRCh38
rs376532811 Chr.14: 103108350-103108350 on Build GRCh38
rs1015612851 Chr.14: 103108665-103108665 on Build GRCh38
rs750369988 Chr.14: 103108462-103108462 on Build GRCh38
rs2274686 Chr.14: 103108530-103108530 on Build GRCh38
rs931197607 Chr.14: 103108622-103108622 on Build GRCh38
rs947098167 Chr.14: 103108788-103108788 on Build GRCh38
rs759429676 Chr.14: 103108488-103108488 on Build GRCh38
rs10144543 Chr.14: 103108377-103108377 on Build GRCh38
rs777326406 Chr.14: 103108432-103108432 on Build GRCh38
rs143373036 Chr.14: 103108471-103108471 on Build GRCh38
rs752330030 Chr.14: 103108369-103108369 on Build GRCh38
rs781214501 Chr.14: 103108525-103108525 on Build GRCh38
rs1029082484 Chr.14: 103108772-103108772 on Build GRCh38
rs111501373 Chr.14: 103108343-103108343 on Build GRCh38
rs746172335 Chr.14: 103108483-103108483 on Build GRCh38
rs746248737 Chr.14: 103108526-103108526 on Build GRCh38
rs553500199 Chr.14: 103108421-103108421 on Build GRCh38
rs1001363898 Chr.14: 103108408-103108408 on Build GRCh38
rs368664108 Chr.14: 103108401-103108401 on Build GRCh38
rs371928931 Chr.14: 103108666-103108666 on Build GRCh38
rs774042905 Chr.14: 103108498-103108498 on Build GRCh38
rs758399381 Chr.14: 103108473-103108473 on Build GRCh38
rs777165139 Chr.14: 103108479-103108479 on Build GRCh38
rs773498066 Chr.14: 103108419-103108419 on Build GRCh38
rs757105801 Chr.14: 103108453-103108453 on Build GRCh38
rs373552730 Chr.14: 103108370-103108370 on Build GRCh38
rs944187194 Chr.14: 103108773-103108773 on Build GRCh38

View Full Product Details