Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103521016-103521535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKB
Entrez Gene ID
1152
Gene Name
creatine kinase B
Gene Aliases
B-CK, BCK, CKBB, HEL-211, HEL-S-29
UniGene
Hs.173724
Gene Chromosome Location
Chr.14: 103519658-103522859 on Build GRCh38
Transcripts
NM_001823.4 XM_017020951.1 XM_017020952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4147841 c.606G>A p.L202L Chr.14: 103521310-103521310 on Build GRCh38
7180659 c.568G>C p.D190H Chr.14: 103521348-103521348 on Build GRCh38
4136234 c.498C>G p.D166E Chr.14: 103521418-103521418 on Build GRCh38
6426509 c.494T>C p.L165P Chr.14: 103521422-103521422 on Build GRCh38
5620499 c.517T>C p.Y173H Chr.14: 103521399-103521399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760338292 Chr.14: 103521217-103521220 on Build GRCh38
rs762471123 Chr.14: 103521161-103521161 on Build GRCh38
rs980059690 Chr.14: 103521038-103521038 on Build GRCh38
rs374321801 Chr.14: 103521476-103521476 on Build GRCh38
rs759797744 Chr.14: 103521444-103521444 on Build GRCh38
rs199890528 Chr.14: 103521217-103521217 on Build GRCh38
rs370276279 Chr.14: 103521249-103521249 on Build GRCh38
rs1024113733 Chr.14: 103521176-103521176 on Build GRCh38
rs753527319 Chr.14: 103521416-103521416 on Build GRCh38
rs952821803 Chr.14: 103521164-103521164 on Build GRCh38
rs990375963 Chr.14: 103521050-103521051 on Build GRCh38
rs771562633 Chr.14: 103521130-103521130 on Build GRCh38
rs368610740 Chr.14: 103521239-103521239 on Build GRCh38
rs760682236 Chr.14: 103521317-103521317 on Build GRCh38
rs755489688 Chr.14: 103521069-103521069 on Build GRCh38
rs775331914 Chr.14: 103521301-103521301 on Build GRCh38
rs775455742 Chr.14: 103521231-103521231 on Build GRCh38
rs745686895 Chr.14: 103521296-103521296 on Build GRCh38
rs776553779 Chr.14: 103521220-103521220 on Build GRCh38
rs575158421 Chr.14: 103521505-103521505 on Build GRCh38
rs774680391 Chr.14: 103521235-103521235 on Build GRCh38
rs766619543 Chr.14: 103521325-103521325 on Build GRCh38
rs202115891 Chr.14: 103521460-103521460 on Build GRCh38
rs746626440 Chr.14: 103521212-103521212 on Build GRCh38
rs774233084 Chr.14: 103521438-103521438 on Build GRCh38
rs1044526035 Chr.14: 103521103-103521103 on Build GRCh38
rs544177157 Chr.14: 103521507-103521507 on Build GRCh38
rs934912918 Chr.14: 103521040-103521040 on Build GRCh38
rs994376078 Chr.14: 103521157-103521157 on Build GRCh38
rs990429761 Chr.14: 103521206-103521207 on Build GRCh38
rs750712872 Chr.14: 103521240-103521240 on Build GRCh38
rs754012287 Chr.14: 103521348-103521348 on Build GRCh38
rs941862478 Chr.14: 103521343-103521343 on Build GRCh38
rs756879188 Chr.14: 103521467-103521468 on Build GRCh38
rs750103975 Chr.14: 103521484-103521484 on Build GRCh38
rs760766301 Chr.14: 103521439-103521439 on Build GRCh38
rs770978803 Chr.14: 103521441-103521441 on Build GRCh38
rs368048391 Chr.14: 103521312-103521312 on Build GRCh38
rs969446283 Chr.14: 103521191-103521191 on Build GRCh38
rs907283690 Chr.14: 103521126-103521126 on Build GRCh38
rs369278146 Chr.14: 103521485-103521485 on Build GRCh38
rs777417115 Chr.14: 103521259-103521259 on Build GRCh38
rs767133418 Chr.14: 103521481-103521481 on Build GRCh38
rs1035441563 Chr.14: 103521475-103521475 on Build GRCh38
rs765604703 Chr.14: 103521381-103521381 on Build GRCh38
rs780397194 Chr.14: 103521046-103521046 on Build GRCh38
rs956173859 Chr.14: 103521203-103521203 on Build GRCh38
rs749139920 Chr.14: 103521486-103521486 on Build GRCh38
rs753209091 Chr.14: 103521382-103521382 on Build GRCh38
rs888633414 Chr.14: 103521409-103521409 on Build GRCh38
rs745508721 Chr.14: 103521402-103521402 on Build GRCh38
rs1041327622 Chr.14: 103521143-103521143 on Build GRCh38
rs565861001 Chr.14: 103521078-103521078 on Build GRCh38
rs546351693 Chr.14: 103521050-103521050 on Build GRCh38
rs1005640118 Chr.14: 103521170-103521170 on Build GRCh38
rs746716173 Chr.14: 103521266-103521266 on Build GRCh38
rs977838121 Chr.14: 103521196-103521196 on Build GRCh38
rs200890896 Chr.14: 103521391-103521391 on Build GRCh38
rs766533899 Chr.14: 103521243-103521243 on Build GRCh38
rs370427943 Chr.14: 103521215-103521223 on Build GRCh38
rs769546303 Chr.14: 103521407-103521407 on Build GRCh38
rs528522014 Chr.14: 103521138-103521138 on Build GRCh38
rs759343818 Chr.14: 103521225-103521225 on Build GRCh38
rs759868311 Chr.14: 103521367-103521367 on Build GRCh38
rs889478478 Chr.14: 103521137-103521137 on Build GRCh38
rs753223914 Chr.14: 103521250-103521250 on Build GRCh38
rs982505834 Chr.14: 103521199-103521199 on Build GRCh38
rs1025808393 Chr.14: 103521163-103521163 on Build GRCh38
rs913019324 Chr.14: 103521077-103521077 on Build GRCh38
rs911012665 Chr.14: 103521503-103521503 on Build GRCh38
rs373730099 Chr.14: 103521280-103521280 on Build GRCh38
rs938554433 Chr.14: 103521064-103521064 on Build GRCh38
rs954621077 Chr.14: 103521178-103521178 on Build GRCh38
rs781011154 Chr.14: 103521393-103521393 on Build GRCh38
rs901402911 Chr.14: 103521150-103521150 on Build GRCh38
rs756319486 Chr.14: 103521241-103521241 on Build GRCh38
rs988655343 Chr.14: 103521223-103521223 on Build GRCh38
rs753117577 Chr.14: 103521462-103521462 on Build GRCh38
rs747645661 Chr.14: 103521096-103521096 on Build GRCh38
rs200266751 Chr.14: 103521417-103521417 on Build GRCh38
rs763157009 Chr.14: 103521466-103521466 on Build GRCh38
rs1052882927 Chr.14: 103521074-103521074 on Build GRCh38
rs756873696 Chr.14: 103521480-103521480 on Build GRCh38
rs768646865 Chr.14: 103521424-103521424 on Build GRCh38
rs769578998 Chr.14: 103521297-103521297 on Build GRCh38
rs765086884 Chr.14: 103521227-103521227 on Build GRCh38
rs1037618915 Chr.14: 103521368-103521368 on Build GRCh38
rs947146408 Chr.14: 103521101-103521101 on Build GRCh38
rs767291625 Chr.14: 103521213-103521214 on Build GRCh38
rs1041105054 Chr.14: 103521351-103521351 on Build GRCh38
rs968552046 Chr.14: 103521494-103521494 on Build GRCh38
rs537451279 Chr.14: 103521245-103521245 on Build GRCh38
rs137901801 Chr.14: 103521145-103521146 on Build GRCh38
rs938785111 Chr.14: 103521271-103521271 on Build GRCh38
rs778366070 Chr.14: 103521387-103521387 on Build GRCh38
rs764724502 Chr.14: 103521470-103521470 on Build GRCh38
rs36002620 Chr.14: 103521386-103521386 on Build GRCh38
rs776880074 Chr.14: 103521443-103521443 on Build GRCh38
rs959941529 Chr.14: 103521221-103521221 on Build GRCh38
rs1016125181 Chr.14: 103521152-103521152 on Build GRCh38
rs763838800 Chr.14: 103521284-103521285 on Build GRCh38
rs773414293 Chr.14: 103521146-103521151 on Build GRCh38
rs770716903 Chr.14: 103521279-103521279 on Build GRCh38
rs558681211 Chr.14: 103521498-103521498 on Build GRCh38
rs1042750270 Chr.14: 103521131-103521131 on Build GRCh38
rs748351557 Chr.14: 103521146-103521148 on Build GRCh38
rs1023746346 Chr.14: 103521454-103521454 on Build GRCh38
rs1047304467 Chr.14: 103521392-103521392 on Build GRCh38
rs773296929 Chr.14: 103521315-103521315 on Build GRCh38

View Full Product Details