Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103556154-103556680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG5
Entrez Gene ID
9529
Gene Name
BCL2 associated athanogene 5
Gene Aliases
BAG-5
UniGene
Hs.5443
Gene Chromosome Location
Chr.14: 103556544-103562814 on Build GRCh38
Transcripts
NM_001015048.2 NM_001015049.2 NM_004873.3

SNP Detail

SNP ID Chromosome Location
rs914124261 Chr.14: 103556518-103556518 on Build GRCh38
rs11376839 Chr.14: 103556457-103556458 on Build GRCh38
rs925412787 Chr.14: 103556528-103556528 on Build GRCh38
rs560178831 Chr.14: 103556227-103556227 on Build GRCh38
rs1021176126 Chr.14: 103556563-103556564 on Build GRCh38
rs531955574 Chr.14: 103556308-103556308 on Build GRCh38
rs966934689 Chr.14: 103556520-103556520 on Build GRCh38
rs921533000 Chr.14: 103556478-103556478 on Build GRCh38
rs535394525 Chr.14: 103556493-103556493 on Build GRCh38
rs1001001538 Chr.14: 103556563-103556564 on Build GRCh38
rs748803091 Chr.14: 103556284-103556284 on Build GRCh38
rs141918742 Chr.14: 103556323-103556323 on Build GRCh38
rs540393236 Chr.14: 103556177-103556177 on Build GRCh38
rs397806601 Chr.14: 103556473-103556474 on Build GRCh38
rs185198644 Chr.14: 103556563-103556563 on Build GRCh38
rs1051427315 Chr.14: 103556445-103556445 on Build GRCh38
rs7154948 Chr.14: 103556607-103556607 on Build GRCh38
rs1032506516 Chr.14: 103556574-103556574 on Build GRCh38
rs980424721 Chr.14: 103556245-103556245 on Build GRCh38
rs980002270 Chr.14: 103556523-103556523 on Build GRCh38
rs938086637 Chr.14: 103556465-103556466 on Build GRCh38
rs990904075 Chr.14: 103556609-103556609 on Build GRCh38
rs867633510 Chr.14: 103556318-103556318 on Build GRCh38
rs887017542 Chr.14: 103556658-103556658 on Build GRCh38
rs998124541 Chr.14: 103556443-103556443 on Build GRCh38
rs965543056 Chr.14: 103556230-103556230 on Build GRCh38
rs1016725763 Chr.14: 103556476-103556476 on Build GRCh38
rs960623466 Chr.14: 103556501-103556501 on Build GRCh38
rs569660069 Chr.14: 103556481-103556481 on Build GRCh38
rs192816540 Chr.14: 103556328-103556328 on Build GRCh38
rs943462486 Chr.14: 103556536-103556536 on Build GRCh38
rs576973924 Chr.14: 103556646-103556646 on Build GRCh38
rs562542418 Chr.14: 103556288-103556288 on Build GRCh38
rs148469077 Chr.14: 103556434-103556434 on Build GRCh38
rs76005268 Chr.14: 103556562-103556562 on Build GRCh38
rs1001915235 Chr.14: 103556488-103556488 on Build GRCh38
rs923290245 Chr.14: 103556583-103556583 on Build GRCh38
rs370713600 Chr.14: 103556513-103556513 on Build GRCh38
rs1006472398 Chr.14: 103556243-103556243 on Build GRCh38
rs189772962 Chr.14: 103556282-103556282 on Build GRCh38
rs925359652 Chr.14: 103556390-103556390 on Build GRCh38
rs750625354 Chr.14: 103556628-103556628 on Build GRCh38
rs931126522 Chr.14: 103556512-103556512 on Build GRCh38
rs564579945 Chr.14: 103556475-103556475 on Build GRCh38
rs982568898 Chr.14: 103556206-103556206 on Build GRCh38
rs926902794 Chr.14: 103556253-103556253 on Build GRCh38
rs576913167 Chr.14: 103556277-103556277 on Build GRCh38
rs886969759 Chr.14: 103556473-103556473 on Build GRCh38
rs939586701 Chr.14: 103556317-103556317 on Build GRCh38
rs780569735 Chr.14: 103556456-103556457 on Build GRCh38
rs1039011457 Chr.14: 103556414-103556414 on Build GRCh38
rs966888670 Chr.14: 103556411-103556411 on Build GRCh38
rs990938891 Chr.14: 103556474-103556474 on Build GRCh38
rs547188693 Chr.14: 103556384-103556384 on Build GRCh38
rs900815678 Chr.14: 103556552-103556552 on Build GRCh38
rs527856063 Chr.14: 103556345-103556345 on Build GRCh38
rs7154587 Chr.14: 103556413-103556413 on Build GRCh38
rs570593350 Chr.14: 103556388-103556388 on Build GRCh38

View Full Product Details