Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103913883-103914387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf2
Entrez Gene ID
9556
Gene Name
chromosome 14 open reading frame 2
Gene Aliases
MP68, PLPM
UniGene
Hs.109052
Gene Chromosome Location
Chr.14: 103912288-103921566 on Build GRCh38
Transcripts
NM_001127393.1 NM_004894.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5601124 c.148+4_148+8delAGATT p.? Chr.14: 103913953-103913957 on Build GRCh38
4853540 c.195G>A p.L65L Chr.14: 103913965-103913965 on Build GRCh38
4853541 c.144G>A p.L48L Chr.14: 103913965-103913965 on Build GRCh38
953680 c.131G>T p.R44I Chr.14: 103913978-103913978 on Build GRCh38
5601123 c.199+4_199+8delAGATT p.? Chr.14: 103913953-103913957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769419419 Chr.14: 103913999-103914000 on Build GRCh38
rs896254118 Chr.14: 103914297-103914297 on Build GRCh38
rs888180329 Chr.14: 103914214-103914214 on Build GRCh38
rs934074690 Chr.14: 103914167-103914167 on Build GRCh38
rs142886349 Chr.14: 103914229-103914229 on Build GRCh38
rs200773885 Chr.14: 103913970-103913970 on Build GRCh38
rs917411756 Chr.14: 103914124-103914124 on Build GRCh38
rs764920786 Chr.14: 103914275-103914275 on Build GRCh38
rs745624520 Chr.14: 103913926-103913926 on Build GRCh38
rs761409929 Chr.14: 103914263-103914263 on Build GRCh38
rs151059352 Chr.14: 103914329-103914329 on Build GRCh38
rs1036612649 Chr.14: 103914160-103914160 on Build GRCh38
rs773430486 Chr.14: 103914004-103914004 on Build GRCh38
rs776684852 Chr.14: 103913914-103913914 on Build GRCh38
rs993820917 Chr.14: 103914203-103914203 on Build GRCh38
rs760903915 Chr.14: 103914013-103914013 on Build GRCh38
rs370793148 Chr.14: 103913943-103913943 on Build GRCh38
rs767646086 Chr.14: 103914002-103914002 on Build GRCh38
rs565105771 Chr.14: 103914194-103914194 on Build GRCh38
rs1032225669 Chr.14: 103914097-103914097 on Build GRCh38
rs769668721 Chr.14: 103913932-103913932 on Build GRCh38
rs1048114818 Chr.14: 103914212-103914212 on Build GRCh38
rs762486965 Chr.14: 103914002-103914002 on Build GRCh38
rs758226443 Chr.14: 103914363-103914363 on Build GRCh38
rs763649553 Chr.14: 103914226-103914226 on Build GRCh38
rs975414576 Chr.14: 103914126-103914126 on Build GRCh38
rs185400295 Chr.14: 103914237-103914237 on Build GRCh38
rs950290247 Chr.14: 103914155-103914155 on Build GRCh38
rs868612066 Chr.14: 103914273-103914273 on Build GRCh38
rs750123270 Chr.14: 103914343-103914343 on Build GRCh38
rs533646018 Chr.14: 103914230-103914230 on Build GRCh38
rs754409261 Chr.14: 103914020-103914020 on Build GRCh38
rs572652145 Chr.14: 103913952-103913952 on Build GRCh38
rs761138928 Chr.14: 103914162-103914162 on Build GRCh38
rs1018074923 Chr.14: 103914290-103914290 on Build GRCh38
rs747862330 Chr.14: 103913992-103913996 on Build GRCh38
rs755259278 Chr.14: 103914021-103914021 on Build GRCh38
rs376771450 Chr.14: 103914025-103914025 on Build GRCh38
rs375097597 Chr.14: 103913954-103913954 on Build GRCh38
rs979346035 Chr.14: 103914080-103914080 on Build GRCh38
rs111415409 Chr.14: 103914082-103914082 on Build GRCh38
rs921370777 Chr.14: 103914152-103914152 on Build GRCh38
rs1054780551 Chr.14: 103914272-103914272 on Build GRCh38
rs553833325 Chr.14: 103913992-103913992 on Build GRCh38
rs902354045 Chr.14: 103914334-103914334 on Build GRCh38
rs8009847 Chr.14: 103914001-103914001 on Build GRCh38
rs1042120632 Chr.14: 103914331-103914331 on Build GRCh38
rs776038225 Chr.14: 103914036-103914036 on Build GRCh38
rs746913520 Chr.14: 103913971-103913974 on Build GRCh38
rs766775292 Chr.14: 103914018-103914018 on Build GRCh38
rs765775621 Chr.14: 103914021-103914022 on Build GRCh38
rs543440529 Chr.14: 103914205-103914205 on Build GRCh38
rs1055936795 Chr.14: 103914076-103914076 on Build GRCh38
rs957612783 Chr.14: 103914121-103914121 on Build GRCh38
rs898723231 Chr.14: 103914258-103914258 on Build GRCh38
rs998016305 Chr.14: 103914326-103914326 on Build GRCh38
rs772621828 Chr.14: 103914000-103914000 on Build GRCh38
rs113631950 Chr.14: 103914362-103914362 on Build GRCh38
rs769192690 Chr.14: 103914264-103914265 on Build GRCh38
rs1038599541 Chr.14: 103914243-103914243 on Build GRCh38
rs768488399 Chr.14: 103913976-103913980 on Build GRCh38

View Full Product Details