Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103914935-103915346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf2
Entrez Gene ID
9556
Gene Name
chromosome 14 open reading frame 2
Gene Aliases
MP68, PLPM
UniGene
Hs.109052
Gene Chromosome Location
Chr.14: 103912288-103921566 on Build GRCh38
Transcripts
NM_001127393.1 NM_004894.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1300415 c.167G>A p.R56Q Chr.14: 103915074-103915074 on Build GRCh38
1300416 c.116G>A p.R39Q Chr.14: 103915074-103915074 on Build GRCh38
1135528 c.89T>C p.M30T Chr.14: 103915152-103915152 on Build GRCh38
6614311 c.153C>T p.I51I Chr.14: 103915088-103915088 on Build GRCh38
4186852 c.75G>A p.W25* Chr.14: 103915115-103915115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053419 Chr.14: 103915165-103915165 on Build GRCh38
rs747259696 Chr.14: 103915090-103915090 on Build GRCh38
rs375366415 Chr.14: 103915021-103915021 on Build GRCh38
rs894420427 Chr.14: 103914995-103914995 on Build GRCh38
rs772463787 Chr.14: 103915325-103915325 on Build GRCh38
rs780109497 Chr.14: 103915193-103915193 on Build GRCh38
rs371303152 Chr.14: 103915022-103915022 on Build GRCh38
rs763709667 Chr.14: 103915056-103915056 on Build GRCh38
rs936395639 Chr.14: 103915153-103915153 on Build GRCh38
rs749080256 Chr.14: 103915087-103915087 on Build GRCh38
rs778866537 Chr.14: 103915217-103915217 on Build GRCh38
rs530829960 Chr.14: 103915037-103915037 on Build GRCh38
rs753891703 Chr.14: 103915170-103915171 on Build GRCh38
rs545265668 Chr.14: 103914977-103914977 on Build GRCh38
rs753857525 Chr.14: 103915198-103915198 on Build GRCh38
rs754772315 Chr.14: 103915212-103915212 on Build GRCh38
rs779922073 Chr.14: 103915083-103915083 on Build GRCh38
rs140374540 Chr.14: 103915061-103915061 on Build GRCh38
rs764470239 Chr.14: 103915128-103915128 on Build GRCh38
rs149160111 Chr.14: 103915074-103915074 on Build GRCh38
rs931775782 Chr.14: 103915173-103915173 on Build GRCh38
rs546356392 Chr.14: 103915098-103915098 on Build GRCh38
rs368173941 Chr.14: 103915075-103915075 on Build GRCh38
rs373753548 Chr.14: 103915174-103915174 on Build GRCh38
rs372604665 Chr.14: 103915036-103915036 on Build GRCh38
rs1054064732 Chr.14: 103915163-103915163 on Build GRCh38
rs771182349 Chr.14: 103915231-103915231 on Build GRCh38
rs566276365 Chr.14: 103915116-103915116 on Build GRCh38
rs537363863 Chr.14: 103915250-103915250 on Build GRCh38
rs188036235 Chr.14: 103915094-103915094 on Build GRCh38
rs77167399 Chr.14: 103914981-103914981 on Build GRCh38
rs758824252 Chr.14: 103915051-103915051 on Build GRCh38
rs776956038 Chr.14: 103915034-103915034 on Build GRCh38
rs759604495 Chr.14: 103915103-103915103 on Build GRCh38
rs768613036 Chr.14: 103915088-103915088 on Build GRCh38
rs757394820 Chr.14: 103915200-103915201 on Build GRCh38
rs560737987 Chr.14: 103915062-103915062 on Build GRCh38
rs763269504 Chr.14: 103915127-103915127 on Build GRCh38
rs753060541 Chr.14: 103915046-103915046 on Build GRCh38
rs747136713 Chr.14: 103915226-103915226 on Build GRCh38
rs201065575 Chr.14: 103915089-103915089 on Build GRCh38
rs112876390 Chr.14: 103915304-103915304 on Build GRCh38
rs555765748 Chr.14: 103915285-103915285 on Build GRCh38
rs890347635 Chr.14: 103915293-103915293 on Build GRCh38
rs765456053 Chr.14: 103915042-103915042 on Build GRCh38
rs923713408 Chr.14: 103915111-103915111 on Build GRCh38
rs781331303 Chr.14: 103915233-103915233 on Build GRCh38
rs1015536161 Chr.14: 103915306-103915306 on Build GRCh38
rs746132558 Chr.14: 103915303-103915303 on Build GRCh38
rs368692211 Chr.14: 103915176-103915176 on Build GRCh38
rs1041443863 Chr.14: 103915300-103915300 on Build GRCh38
rs751161089 Chr.14: 103915143-103915143 on Build GRCh38
rs571559919 Chr.14: 103915243-103915243 on Build GRCh38
rs557285040 Chr.14: 103915269-103915269 on Build GRCh38
rs897280172 Chr.14: 103915307-103915307 on Build GRCh38

View Full Product Details