Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104587642-104588145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf180
Entrez Gene ID
400258
Gene Name
chromosome 14 open reading frame 180
Gene Aliases
C14orf77, NRAC
UniGene
Hs.153827
Gene Chromosome Location
Chr.14: 104579684-104589847 on Build GRCh38
Transcripts
NM_001008404.2 NM_001286399.1 NM_001286400.1 XM_005267638.3 XM_011536764.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1368488 c.128_129insC p.S46fs*39 Chr.14: 104587765-104587766 on Build GRCh38
4533523 c.198G>A p.S66S Chr.14: 104587835-104587835 on Build GRCh38
1242046 c.129delC p.S46fs*3 Chr.14: 104587766-104587766 on Build GRCh38
6139885 c.117C>G p.D39E Chr.14: 104587754-104587754 on Build GRCh38
4878363 c.202C>T p.R68C Chr.14: 104587839-104587839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760999693 Chr.14: 104587913-104587913 on Build GRCh38
rs909862607 Chr.14: 104588099-104588099 on Build GRCh38
rs763551047 Chr.14: 104587784-104587785 on Build GRCh38
rs199521419 Chr.14: 104587840-104587840 on Build GRCh38
rs772370422 Chr.14: 104587900-104587900 on Build GRCh38
rs761345073 Chr.14: 104587766-104587766 on Build GRCh38
rs773817993 Chr.14: 104587773-104587773 on Build GRCh38
rs201727131 Chr.14: 104587724-104587724 on Build GRCh38
rs144428844 Chr.14: 104587851-104587851 on Build GRCh38
rs745775991 Chr.14: 104587882-104587882 on Build GRCh38
rs149488814 Chr.14: 104588063-104588063 on Build GRCh38
rs754270740 Chr.14: 104587857-104587857 on Build GRCh38
rs748580951 Chr.14: 104587835-104587835 on Build GRCh38
rs916217831 Chr.14: 104587821-104587821 on Build GRCh38
rs761848816 Chr.14: 104588073-104588073 on Build GRCh38
rs764621310 Chr.14: 104587765-104587766 on Build GRCh38
rs947794468 Chr.14: 104587679-104587679 on Build GRCh38
rs750544509 Chr.14: 104587790-104587790 on Build GRCh38
rs770865389 Chr.14: 104587810-104587810 on Build GRCh38
rs775358570 Chr.14: 104587892-104587892 on Build GRCh38
rs144973367 Chr.14: 104587746-104587746 on Build GRCh38
rs751622949 Chr.14: 104587788-104587788 on Build GRCh38
rs981860147 Chr.14: 104587968-104587968 on Build GRCh38
rs949659391 Chr.14: 104587680-104587680 on Build GRCh38
rs766243629 Chr.14: 104587771-104587771 on Build GRCh38
rs751916748 Chr.14: 104587909-104587910 on Build GRCh38
rs144894622 Chr.14: 104587865-104587865 on Build GRCh38
rs112399099 Chr.14: 104587792-104587792 on Build GRCh38
rs34869170 Chr.14: 104588016-104588017 on Build GRCh38
rs766366010 Chr.14: 104587919-104587919 on Build GRCh38
rs773899450 Chr.14: 104587745-104587745 on Build GRCh38
rs994005845 Chr.14: 104587895-104587895 on Build GRCh38
rs1043302535 Chr.14: 104588052-104588052 on Build GRCh38
rs201529094 Chr.14: 104587786-104587786 on Build GRCh38
rs141850398 Chr.14: 104587864-104587864 on Build GRCh38
rs916791278 Chr.14: 104587671-104587671 on Build GRCh38
rs771773957 Chr.14: 104587806-104587806 on Build GRCh38
rs952621350 Chr.14: 104587803-104587803 on Build GRCh38
rs1023404298 Chr.14: 104588031-104588031 on Build GRCh38
rs759906999 Chr.14: 104587922-104587922 on Build GRCh38
rs972681427 Chr.14: 104587757-104587757 on Build GRCh38
rs76292733 Chr.14: 104587748-104587748 on Build GRCh38
rs781458489 Chr.14: 104587736-104587736 on Build GRCh38
rs751639012 Chr.14: 104587729-104587729 on Build GRCh38
rs755265727 Chr.14: 104587796-104587796 on Build GRCh38
rs113768792 Chr.14: 104587859-104587859 on Build GRCh38
rs761346458 Chr.14: 104587709-104587709 on Build GRCh38
rs568340704 Chr.14: 104587690-104587690 on Build GRCh38
rs984425000 Chr.14: 104588083-104588083 on Build GRCh38
rs911247689 Chr.14: 104588056-104588056 on Build GRCh38
rs947650646 Chr.14: 104588049-104588049 on Build GRCh38
rs749617172 Chr.14: 104587794-104587794 on Build GRCh38
rs776406536 Chr.14: 104587812-104587812 on Build GRCh38
rs576367205 Chr.14: 104587741-104587741 on Build GRCh38
rs999198899 Chr.14: 104587826-104587826 on Build GRCh38
rs765126356 Chr.14: 104587778-104587778 on Build GRCh38
rs192674070 Chr.14: 104587841-104587841 on Build GRCh38
rs780407065 Chr.14: 104587739-104587739 on Build GRCh38
rs1005442746 Chr.14: 104587931-104587931 on Build GRCh38
rs746422041 Chr.14: 104587699-104587699 on Build GRCh38
rs1003677136 Chr.14: 104588051-104588051 on Build GRCh38
rs762652719 Chr.14: 104587703-104587703 on Build GRCh38
rs901268023 Chr.14: 104588100-104588100 on Build GRCh38
rs772776625 Chr.14: 104587761-104587761 on Build GRCh38
rs139748841 Chr.14: 104587839-104587839 on Build GRCh38
rs988946081 Chr.14: 104588012-104588012 on Build GRCh38
rs754008742 Chr.14: 104587920-104587920 on Build GRCh38
rs759269377 Chr.14: 104587773-104587773 on Build GRCh38
rs116669858 Chr.14: 104587998-104587998 on Build GRCh38
rs756575881 Chr.14: 104587791-104587791 on Build GRCh38
rs564349827 Chr.14: 104587687-104587687 on Build GRCh38
rs377302831 Chr.14: 104587713-104587713 on Build GRCh38
rs775785207 Chr.14: 104587702-104587702 on Build GRCh38
rs73361819 Chr.14: 104587707-104587707 on Build GRCh38
rs553984960 Chr.14: 104587712-104587712 on Build GRCh38
rs9788527 Chr.14: 104587923-104587923 on Build GRCh38
rs1025443974 Chr.14: 104587915-104587915 on Build GRCh38
rs555264092 Chr.14: 104587871-104587871 on Build GRCh38
rs772655925 Chr.14: 104587807-104587807 on Build GRCh38
rs759555265 Chr.14: 104587834-104587834 on Build GRCh38
rs773547359 Chr.14: 104587906-104587906 on Build GRCh38
rs138014749 Chr.14: 104587936-104587936 on Build GRCh38
rs146244955 Chr.14: 104587876-104587876 on Build GRCh38
rs770313320 Chr.14: 104587872-104587872 on Build GRCh38
rs370128922 Chr.14: 104587785-104587785 on Build GRCh38
rs556254029 Chr.14: 104588022-104588022 on Build GRCh38
rs572278637 Chr.14: 104587781-104587781 on Build GRCh38
rs762870829 Chr.14: 104587893-104587893 on Build GRCh38
rs927714816 Chr.14: 104587991-104587991 on Build GRCh38
rs755277534 Chr.14: 104587795-104587795 on Build GRCh38
rs776267399 Chr.14: 104587874-104587874 on Build GRCh38
rs202177288 Chr.14: 104587783-104587783 on Build GRCh38
rs999480316 Chr.14: 104587944-104587944 on Build GRCh38
rs1053853551 Chr.14: 104587982-104587982 on Build GRCh38
rs769783996 Chr.14: 104587887-104587887 on Build GRCh38
rs35883949 Chr.14: 104587981-104587982 on Build GRCh38
rs777437346 Chr.14: 104587869-104587869 on Build GRCh38
rs1051647396 Chr.14: 104587935-104587935 on Build GRCh38
rs768205649 Chr.14: 104587706-104587706 on Build GRCh38
rs140952034 Chr.14: 104587662-104587662 on Build GRCh38
rs374207195 Chr.14: 104587701-104587701 on Build GRCh38
rs1023742718 Chr.14: 104587932-104587932 on Build GRCh38
rs766855232 Chr.14: 104587877-104587877 on Build GRCh38
rs768313484 Chr.14: 104587743-104587743 on Build GRCh38
rs964094816 Chr.14: 104588106-104588106 on Build GRCh38
rs373047135 Chr.14: 104587710-104587710 on Build GRCh38
rs776699068 Chr.14: 104588062-104588062 on Build GRCh38
rs746368743 Chr.14: 104587738-104587738 on Build GRCh38
rs375304625 Chr.14: 104587802-104587802 on Build GRCh38
rs61743131 Chr.14: 104587852-104587852 on Build GRCh38
rs199942352 Chr.14: 104587725-104587725 on Build GRCh38
rs757387751 Chr.14: 104587735-104587735 on Build GRCh38
rs184171404 Chr.14: 104588035-104588035 on Build GRCh38
rs760596404 Chr.14: 104587767-104587767 on Build GRCh38

View Full Product Details