Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104937566-104938077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHNAK2
Entrez Gene ID
113146
Gene Name
AHNAK nucleoprotein 2
Gene Aliases
C14orf78
UniGene
Hs.441783
Gene Chromosome Location
Chr.14: 104937253-104978357 on Build GRCh38
Transcripts
NM_138420.2 XM_005267299.1 XM_011536411.2 XM_017020965.1

Gene Symbol
PLD4
Entrez Gene ID
122618
Gene Name
phospholipase D family member 4
Gene Aliases
C14orf175
UniGene
Hs.407101
Gene Chromosome Location
Chr.14: 104924806-104937789 on Build GRCh38
Transcripts
NM_138420.2 XM_005267299.1 XM_011536411.2 XM_017020965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2026664 c.17379T>C p.N5793N Chr.14: 104938072-104938072 on Build GRCh38
2026665 c.2289T>C p.N763N Chr.14: 104938072-104938072 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs893906412 Chr.14: 104937915-104937915 on Build GRCh38
rs193118009 Chr.14: 104937757-104937757 on Build GRCh38
rs974820977 Chr.14: 104937743-104937743 on Build GRCh38
rs188930441 Chr.14: 104937671-104937671 on Build GRCh38
rs1052823448 Chr.14: 104937610-104937610 on Build GRCh38
rs935377497 Chr.14: 104937930-104937930 on Build GRCh38
rs953004733 Chr.14: 104937833-104937833 on Build GRCh38
rs897582720 Chr.14: 104937997-104937997 on Build GRCh38
rs1018751810 Chr.14: 104937996-104937996 on Build GRCh38
rs79161962 Chr.14: 104937677-104937677 on Build GRCh38
rs1052056954 Chr.14: 104937803-104937803 on Build GRCh38
rs950537122 Chr.14: 104938028-104938028 on Build GRCh38
rs1035600559 Chr.14: 104938013-104938013 on Build GRCh38
rs1003195684 Chr.14: 104937999-104937999 on Build GRCh38
rs185589455 Chr.14: 104937847-104937847 on Build GRCh38
rs1027341343 Chr.14: 104938035-104938035 on Build GRCh38
rs948214964 Chr.14: 104937891-104937891 on Build GRCh38
rs1050322748 Chr.14: 104937974-104937974 on Build GRCh38
rs779782003 Chr.14: 104937897-104937897 on Build GRCh38
rs747334439 Chr.14: 104937905-104937905 on Build GRCh38
rs1048257 Chr.14: 104938047-104938047 on Build GRCh38
rs78726317 Chr.14: 104938048-104938048 on Build GRCh38
rs572103420 Chr.14: 104937940-104937940 on Build GRCh38
rs1008735426 Chr.14: 104937993-104937993 on Build GRCh38
rs758896698 Chr.14: 104937879-104937879 on Build GRCh38
rs963054340 Chr.14: 104937730-104937730 on Build GRCh38
rs552338664 Chr.14: 104937788-104937788 on Build GRCh38
rs779522525 Chr.14: 104937632-104937632 on Build GRCh38
rs893729641 Chr.14: 104937616-104937616 on Build GRCh38
rs953174717 Chr.14: 104937672-104937672 on Build GRCh38
rs1028846204 Chr.14: 104937665-104937665 on Build GRCh38
rs970389646 Chr.14: 104937644-104937644 on Build GRCh38
rs1011208537 Chr.14: 104937658-104937658 on Build GRCh38
rs993624777 Chr.14: 104938034-104938034 on Build GRCh38
rs1046450721 Chr.14: 104937595-104937595 on Build GRCh38
rs748921467 Chr.14: 104938022-104938022 on Build GRCh38
rs916775986 Chr.14: 104937875-104937875 on Build GRCh38
rs1052457899 Chr.14: 104937944-104937944 on Build GRCh38
rs984319326 Chr.14: 104937870-104937870 on Build GRCh38
rs769766602 Chr.14: 104938050-104938051 on Build GRCh38
rs999529019 Chr.14: 104937599-104937599 on Build GRCh38
rs568243880 Chr.14: 104937804-104937804 on Build GRCh38
rs777155301 Chr.14: 104937973-104937973 on Build GRCh38
rs144991997 Chr.14: 104937906-104937906 on Build GRCh38
rs138792829 Chr.14: 104937945-104937945 on Build GRCh38
rs906576150 Chr.14: 104937596-104937596 on Build GRCh38
rs1018699694 Chr.14: 104937692-104937692 on Build GRCh38
rs569969398 Chr.14: 104937764-104937764 on Build GRCh38
rs781046331 Chr.14: 104938033-104938033 on Build GRCh38
rs866764204 Chr.14: 104937769-104937769 on Build GRCh38
rs1010850118 Chr.14: 104937971-104937971 on Build GRCh38
rs372176047 Chr.14: 104938053-104938053 on Build GRCh38
rs920221336 Chr.14: 104937828-104937828 on Build GRCh38
rs557930881 Chr.14: 104937948-104937948 on Build GRCh38
rs768229280 Chr.14: 104937984-104937984 on Build GRCh38

View Full Product Details