Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104992841-104993341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf79
Entrez Gene ID
122616
Gene Name
chromosome 14 open reading frame 79
Gene Aliases
-
UniGene
Hs.27183
Gene Chromosome Location
Chr.14: 104985801-105000310 on Build GRCh38
Transcripts
NM_174891.3 XM_005267317.3 XM_005267318.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5920660 c.700-5C>T p.? Chr.14: 104992943-104992943 on Build GRCh38
6614336 c.782G>A p.C261Y Chr.14: 104993030-104993030 on Build GRCh38
6355448 c.720C>T p.G240G Chr.14: 104992968-104992968 on Build GRCh38
3494255 c.763C>T p.L255F Chr.14: 104993011-104993011 on Build GRCh38
7283645 c.769G>A p.V257I Chr.14: 104993017-104993017 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953535826 Chr.14: 104993233-104993233 on Build GRCh38
rs755649629 Chr.14: 104992976-104992976 on Build GRCh38
rs541460585 Chr.14: 104993107-104993107 on Build GRCh38
rs966527807 Chr.14: 104993282-104993282 on Build GRCh38
rs763587137 Chr.14: 104993058-104993058 on Build GRCh38
rs926491746 Chr.14: 104993201-104993201 on Build GRCh38
rs553515417 Chr.14: 104992875-104992875 on Build GRCh38
rs376699203 Chr.14: 104993019-104993019 on Build GRCh38
rs966061786 Chr.14: 104992883-104992883 on Build GRCh38
rs116352190 Chr.14: 104993267-104993267 on Build GRCh38
rs895595016 Chr.14: 104993144-104993144 on Build GRCh38
rs772771782 Chr.14: 104993135-104993135 on Build GRCh38
rs779606303 Chr.14: 104992977-104992977 on Build GRCh38
rs775572576 Chr.14: 104993052-104993052 on Build GRCh38
rs142561418 Chr.14: 104993289-104993289 on Build GRCh38
rs752619296 Chr.14: 104993115-104993115 on Build GRCh38
rs760304582 Chr.14: 104993106-104993106 on Build GRCh38
rs367841290 Chr.14: 104993100-104993100 on Build GRCh38
rs900766039 Chr.14: 104993145-104993145 on Build GRCh38
rs775918317 Chr.14: 104992943-104992943 on Build GRCh38
rs372320531 Chr.14: 104993018-104993018 on Build GRCh38
rs890769859 Chr.14: 104993316-104993316 on Build GRCh38
rs757377749 Chr.14: 104993050-104993050 on Build GRCh38
rs373560879 Chr.14: 104992955-104992955 on Build GRCh38
rs746088734 Chr.14: 104993008-104993008 on Build GRCh38
rs763495910 Chr.14: 104992948-104992948 on Build GRCh38
rs926560477 Chr.14: 104993049-104993049 on Build GRCh38
rs748068963 Chr.14: 104993063-104993064 on Build GRCh38
rs754866898 Chr.14: 104992983-104992983 on Build GRCh38
rs955145889 Chr.14: 104992930-104992930 on Build GRCh38
rs774978210 Chr.14: 104993098-104993098 on Build GRCh38
rs1039214293 Chr.14: 104993272-104993272 on Build GRCh38
rs750106224 Chr.14: 104993101-104993101 on Build GRCh38
rs988123308 Chr.14: 104993291-104993291 on Build GRCh38
rs1042471516 Chr.14: 104993258-104993258 on Build GRCh38
rs747260131 Chr.14: 104992898-104992898 on Build GRCh38
rs201948594 Chr.14: 104993124-104993124 on Build GRCh38
rs775588758 Chr.14: 104993061-104993063 on Build GRCh38
rs374333558 Chr.14: 104992963-104992963 on Build GRCh38
rs1055536048 Chr.14: 104993118-104993119 on Build GRCh38
rs771089395 Chr.14: 104992914-104992914 on Build GRCh38
rs771821067 Chr.14: 104993000-104993000 on Build GRCh38
rs917911686 Chr.14: 104993247-104993247 on Build GRCh38
rs201325152 Chr.14: 104992920-104992920 on Build GRCh38
rs913107347 Chr.14: 104993013-104993013 on Build GRCh38
rs546318117 Chr.14: 104993210-104993210 on Build GRCh38
rs746360340 Chr.14: 104993131-104993131 on Build GRCh38
rs769238451 Chr.14: 104993088-104993088 on Build GRCh38
rs758196097 Chr.14: 104993116-104993116 on Build GRCh38
rs776887803 Chr.14: 104992918-104992918 on Build GRCh38
rs200250144 Chr.14: 104992972-104992972 on Build GRCh38
rs1029329888 Chr.14: 104993155-104993155 on Build GRCh38
rs181306438 Chr.14: 104992990-104992990 on Build GRCh38
rs900713701 Chr.14: 104993275-104993275 on Build GRCh38
rs988584155 Chr.14: 104992928-104992928 on Build GRCh38
rs532581009 Chr.14: 104993199-104993199 on Build GRCh38
rs925793595 Chr.14: 104993110-104993110 on Build GRCh38
rs539550549 Chr.14: 104992949-104992949 on Build GRCh38
rs767126176 Chr.14: 104992968-104992968 on Build GRCh38
rs781335167 Chr.14: 104993006-104993006 on Build GRCh38
rs2819467 Chr.14: 104993172-104993172 on Build GRCh38
rs751933118 Chr.14: 104992957-104992957 on Build GRCh38
rs982978541 Chr.14: 104992965-104992965 on Build GRCh38
rs973879900 Chr.14: 104993190-104993190 on Build GRCh38
rs959315458 Chr.14: 104993219-104993219 on Build GRCh38
rs555306106 Chr.14: 104993108-104993108 on Build GRCh38
rs945380401 Chr.14: 104993249-104993249 on Build GRCh38
rs756226905 Chr.14: 104992891-104992891 on Build GRCh38
rs747982650 Chr.14: 104992996-104992996 on Build GRCh38
rs992507374 Chr.14: 104993242-104993242 on Build GRCh38
rs955240566 Chr.14: 104993164-104993164 on Build GRCh38
rs575145828 Chr.14: 104993146-104993146 on Build GRCh38
rs747412181 Chr.14: 104993271-104993271 on Build GRCh38
rs1015311695 Chr.14: 104993178-104993178 on Build GRCh38
rs1052475348 Chr.14: 104993296-104993296 on Build GRCh38
rs531733422 Chr.14: 104993283-104993283 on Build GRCh38
rs148860636 Chr.14: 104993188-104993188 on Build GRCh38
rs762314997 Chr.14: 104993099-104993099 on Build GRCh38
rs766934394 Chr.14: 104993168-104993171 on Build GRCh38
rs576549151 Chr.14: 104992919-104992919 on Build GRCh38

View Full Product Details