Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105011772-105012281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA4
Entrez Gene ID
55038
Gene Name
cell division cycle associated 4
Gene Aliases
HEPP, SEI-3/HEPP
UniGene
Hs.34045
Gene Chromosome Location
Chr.14: 105009573-105021088 on Build GRCh38
Transcripts
NM_017955.3 NM_145701.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5832734 c.41_42insA p.E15fs*114 Chr.14: 105011888-105011889 on Build GRCh38
4432157 c.101G>A p.R34Q Chr.14: 105011829-105011829 on Build GRCh38
5847693 c.83C>T p.S28F Chr.14: 105011847-105011847 on Build GRCh38
4567229 c.60_61CC>AT p.(=) Chr.14: 105011869-105011870 on Build GRCh38
7174125 c.40G>C p.E14Q Chr.14: 105011890-105011890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202086356 Chr.14: 105011933-105011933 on Build GRCh38
rs373156203 Chr.14: 105011884-105011884 on Build GRCh38
rs986368642 Chr.14: 105012049-105012049 on Build GRCh38
rs369501220 Chr.14: 105011882-105011882 on Build GRCh38
rs144801862 Chr.14: 105012012-105012012 on Build GRCh38
rs898893206 Chr.14: 105012235-105012235 on Build GRCh38
rs772846501 Chr.14: 105011976-105011976 on Build GRCh38
rs772002709 Chr.14: 105011970-105011970 on Build GRCh38
rs142906536 Chr.14: 105012044-105012044 on Build GRCh38
rs765015889 Chr.14: 105012127-105012127 on Build GRCh38
rs571432004 Chr.14: 105012122-105012122 on Build GRCh38
rs45618437 Chr.14: 105011948-105011948 on Build GRCh38
rs188548704 Chr.14: 105011984-105011984 on Build GRCh38
rs989629329 Chr.14: 105012033-105012033 on Build GRCh38
rs200800804 Chr.14: 105011812-105011812 on Build GRCh38
rs752763454 Chr.14: 105011891-105011891 on Build GRCh38
rs374846280 Chr.14: 105011945-105011945 on Build GRCh38
rs533607836 Chr.14: 105011983-105011983 on Build GRCh38
rs755680204 Chr.14: 105011949-105011949 on Build GRCh38
rs1012139593 Chr.14: 105012004-105012004 on Build GRCh38
rs886259928 Chr.14: 105012200-105012201 on Build GRCh38
rs779154349 Chr.14: 105011898-105011898 on Build GRCh38
rs145318953 Chr.14: 105011829-105011829 on Build GRCh38
rs768754038 Chr.14: 105011863-105011863 on Build GRCh38
rs748796689 Chr.14: 105011958-105011958 on Build GRCh38
rs889472867 Chr.14: 105011939-105011939 on Build GRCh38
rs1041144274 Chr.14: 105012093-105012093 on Build GRCh38
rs759631223 Chr.14: 105011885-105011885 on Build GRCh38
rs1037516012 Chr.14: 105012221-105012221 on Build GRCh38
rs748028645 Chr.14: 105011969-105011969 on Build GRCh38
rs755553746 Chr.14: 105011949-105011949 on Build GRCh38
rs757587945 Chr.14: 105011881-105011881 on Build GRCh38
rs551736845 Chr.14: 105012064-105012064 on Build GRCh38
rs140201029 Chr.14: 105011794-105011794 on Build GRCh38
rs752057194 Chr.14: 105011919-105011919 on Build GRCh38
rs757625004 Chr.14: 105011920-105011920 on Build GRCh38
rs888294569 Chr.14: 105012172-105012172 on Build GRCh38
rs754095552 Chr.14: 105011810-105011810 on Build GRCh38
rs757606420 Chr.14: 105011832-105011832 on Build GRCh38
rs137892580 Chr.14: 105011852-105011852 on Build GRCh38
rs746563866 Chr.14: 105012007-105012007 on Build GRCh38
rs918309934 Chr.14: 105012107-105012107 on Build GRCh38
rs4073698 Chr.14: 105012247-105012247 on Build GRCh38
rs377230780 Chr.14: 105011979-105011979 on Build GRCh38
rs369116805 Chr.14: 105012025-105012025 on Build GRCh38
rs752289412 Chr.14: 105011879-105011880 on Build GRCh38
rs758793382 Chr.14: 105011823-105011823 on Build GRCh38
rs1028578568 Chr.14: 105012258-105012258 on Build GRCh38
rs143314664 Chr.14: 105011864-105011864 on Build GRCh38
rs989956346 Chr.14: 105012043-105012043 on Build GRCh38
rs563877475 Chr.14: 105012005-105012005 on Build GRCh38
rs567414075 Chr.14: 105012239-105012239 on Build GRCh38
rs982543488 Chr.14: 105012011-105012011 on Build GRCh38
rs111738285 Chr.14: 105012071-105012071 on Build GRCh38
rs564615325 Chr.14: 105011968-105011968 on Build GRCh38
rs749278109 Chr.14: 105011861-105011861 on Build GRCh38
rs566079310 Chr.14: 105012037-105012037 on Build GRCh38
rs550518896 Chr.14: 105012196-105012196 on Build GRCh38
rs772946197 Chr.14: 105012189-105012189 on Build GRCh38
rs747426142 Chr.14: 105011830-105011830 on Build GRCh38
rs927958166 Chr.14: 105012094-105012094 on Build GRCh38
rs745447408 Chr.14: 105011846-105011846 on Build GRCh38
rs879522567 Chr.14: 105012088-105012088 on Build GRCh38
rs768264159 Chr.14: 105011960-105011960 on Build GRCh38
rs778788310 Chr.14: 105011822-105011822 on Build GRCh38
rs912171922 Chr.14: 105012065-105012065 on Build GRCh38
rs1027242904 Chr.14: 105011972-105011972 on Build GRCh38
rs929719647 Chr.14: 105012113-105012113 on Build GRCh38
rs940559364 Chr.14: 105012201-105012201 on Build GRCh38
rs780889852 Chr.14: 105011845-105011845 on Build GRCh38
rs148556426 Chr.14: 105012026-105012026 on Build GRCh38
rs957039765 Chr.14: 105012023-105012023 on Build GRCh38
rs764644055 Chr.14: 105011901-105011901 on Build GRCh38
rs35339875 Chr.14: 105012111-105012112 on Build GRCh38
rs193135763 Chr.14: 105012195-105012195 on Build GRCh38
rs764715608 Chr.14: 105011976-105011977 on Build GRCh38
rs765392183 Chr.14: 105011890-105011890 on Build GRCh38
rs906497494 Chr.14: 105012240-105012240 on Build GRCh38
rs779725263 Chr.14: 105011953-105011953 on Build GRCh38

View Full Product Details