Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105474542-105475079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIP2
Entrez Gene ID
1397
Gene Name
cysteine rich protein 2
Gene Aliases
CRIP, CRP2, ESP1
UniGene
Hs.534309
Gene Chromosome Location
Chr.14: 105472938-105480170 on Build GRCh38
Transcripts
NM_001270837.1 NM_001270841.1 NM_001312.3 NR_073081.1 NR_073082.1 NR_073083.1

SNP Detail

SNP ID Chromosome Location
rs782616987 Chr.14: 105474929-105474929 on Build GRCh38
rs148013668 Chr.14: 105474836-105474836 on Build GRCh38
rs781960994 Chr.14: 105474846-105474846 on Build GRCh38
rs782631571 Chr.14: 105474828-105474828 on Build GRCh38
rs782536262 Chr.14: 105474934-105474934 on Build GRCh38
rs782768323 Chr.14: 105474881-105474881 on Build GRCh38
rs982883458 Chr.14: 105474954-105474954 on Build GRCh38
rs973988561 Chr.14: 105474985-105474985 on Build GRCh38
rs937838015 Chr.14: 105475018-105475018 on Build GRCh38
rs182146787 Chr.14: 105474859-105474859 on Build GRCh38
rs782443172 Chr.14: 105474904-105474904 on Build GRCh38
rs782042376 Chr.14: 105474852-105474852 on Build GRCh38
rs945384872 Chr.14: 105475054-105475054 on Build GRCh38
rs587652277 Chr.14: 105474997-105474997 on Build GRCh38
rs111378860 Chr.14: 105474803-105474803 on Build GRCh38
rs587720379 Chr.14: 105474895-105474895 on Build GRCh38
rs782588488 Chr.14: 105474831-105474831 on Build GRCh38
rs781959094 Chr.14: 105474855-105474855 on Build GRCh38
rs782634104 Chr.14: 105474935-105474935 on Build GRCh38
rs372425630 Chr.14: 105474890-105474890 on Build GRCh38
rs587699417 Chr.14: 105475014-105475014 on Build GRCh38
rs782209719 Chr.14: 105474933-105474933 on Build GRCh38
rs782696166 Chr.14: 105474803-105474804 on Build GRCh38
rs587769419 Chr.14: 105474591-105474591 on Build GRCh38
rs908446488 Chr.14: 105474956-105474956 on Build GRCh38
rs962658335 Chr.14: 105474971-105474971 on Build GRCh38
rs782541124 Chr.14: 105474830-105474830 on Build GRCh38
rs587653614 Chr.14: 105475040-105475040 on Build GRCh38
rs1042818718 Chr.14: 105475055-105475055 on Build GRCh38
rs782265452 Chr.14: 105474850-105474850 on Build GRCh38
rs587604519 Chr.14: 105475035-105475035 on Build GRCh38
rs782295478 Chr.14: 105474829-105474829 on Build GRCh38
rs781811749 Chr.14: 105474901-105474901 on Build GRCh38
rs782427002 Chr.14: 105474785-105474785 on Build GRCh38
rs782063025 Chr.14: 105474854-105474854 on Build GRCh38
rs587597401 Chr.14: 105474995-105474995 on Build GRCh38
rs782529998 Chr.14: 105474822-105474822 on Build GRCh38
rs781854536 Chr.14: 105474886-105474886 on Build GRCh38
rs141796112 Chr.14: 105474871-105474871 on Build GRCh38
rs782294069 Chr.14: 105474937-105474937 on Build GRCh38
rs781820656 Chr.14: 105474809-105474809 on Build GRCh38
rs782356184 Chr.14: 105474840-105474840 on Build GRCh38
rs113499763 Chr.14: 105474725-105474725 on Build GRCh38

View Full Product Details