Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105488318-105488808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIP1
Entrez Gene ID
1396
Gene Name
cysteine rich protein 1
Gene Aliases
CRHP, CRIP, CRP-1, CRP1
UniGene
Hs.70327
Gene Chromosome Location
Chr.14: 105486920-105488789 on Build GRCh38
Transcripts
NM_001311.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3765742 c.173C>T p.A58V Chr.14: 105488368-105488368 on Build GRCh38
6635502 c.194-11_194-9delCTT p.? Chr.14: 105488460-105488462 on Build GRCh38
3748577 c.147A>C p.K49N Chr.14: 105488342-105488342 on Build GRCh38
4049576 c.193G>T p.G65C Chr.14: 105488388-105488388 on Build GRCh38
4049575 c.182T>C p.F61S Chr.14: 105488377-105488377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55633823 Chr.14: 105488368-105488368 on Build GRCh38
rs781968541 Chr.14: 105488533-105488533 on Build GRCh38
rs932446913 Chr.14: 105488697-105488697 on Build GRCh38
rs782250550 Chr.14: 105488472-105488472 on Build GRCh38
rs782078881 Chr.14: 105488731-105488731 on Build GRCh38
rs781936877 Chr.14: 105488557-105488557 on Build GRCh38
rs935918728 Chr.14: 105488481-105488481 on Build GRCh38
rs781994054 Chr.14: 105488456-105488456 on Build GRCh38
rs59732071 Chr.14: 105488612-105488612 on Build GRCh38
rs781913799 Chr.14: 105488417-105488417 on Build GRCh38
rs587712607 Chr.14: 105488425-105488425 on Build GRCh38
rs200341901 Chr.14: 105488535-105488535 on Build GRCh38
rs940090262 Chr.14: 105488747-105488747 on Build GRCh38
rs782315278 Chr.14: 105488463-105488463 on Build GRCh38
rs782056548 Chr.14: 105488412-105488412 on Build GRCh38
rs587713950 Chr.14: 105488341-105488341 on Build GRCh38
rs587741402 Chr.14: 105488578-105488578 on Build GRCh38
rs373010106 Chr.14: 105488410-105488410 on Build GRCh38
rs587662378 Chr.14: 105488637-105488637 on Build GRCh38
rs587624602 Chr.14: 105488403-105488404 on Build GRCh38
rs781873741 Chr.14: 105488424-105488424 on Build GRCh38
rs782378236 Chr.14: 105488647-105488647 on Build GRCh38
rs782587529 Chr.14: 105488444-105488444 on Build GRCh38
rs551006867 Chr.14: 105488484-105488484 on Build GRCh38
rs587606939 Chr.14: 105488431-105488431 on Build GRCh38
rs782558569 Chr.14: 105488518-105488518 on Build GRCh38
rs782180723 Chr.14: 105488392-105488392 on Build GRCh38
rs1012675858 Chr.14: 105488616-105488616 on Build GRCh38
rs200883038 Chr.14: 105488359-105488359 on Build GRCh38
rs202000136 Chr.14: 105488460-105488460 on Build GRCh38
rs781944229 Chr.14: 105488469-105488469 on Build GRCh38
rs782615467 Chr.14: 105488479-105488479 on Build GRCh38
rs587611651 Chr.14: 105488600-105488600 on Build GRCh38
rs782336997 Chr.14: 105488413-105488413 on Build GRCh38
rs782604672 Chr.14: 105488396-105488396 on Build GRCh38
rs782472779 Chr.14: 105488500-105488500 on Build GRCh38
rs781962611 Chr.14: 105488668-105488668 on Build GRCh38
rs912680585 Chr.14: 105488735-105488735 on Build GRCh38
rs587679022 Chr.14: 105488566-105488566 on Build GRCh38
rs782802692 Chr.14: 105488416-105488416 on Build GRCh38
rs782021511 Chr.14: 105488541-105488541 on Build GRCh38
rs1128308 Chr.14: 105488345-105488345 on Build GRCh38
rs782222991 Chr.14: 105488454-105488454 on Build GRCh38
rs782082513 Chr.14: 105488558-105488558 on Build GRCh38
rs921103947 Chr.14: 105488681-105488681 on Build GRCh38
rs201961496 Chr.14: 105488490-105488490 on Build GRCh38
rs587677025 Chr.14: 105488784-105488784 on Build GRCh38
rs55633823 Chr.14: 105488368-105488368 on Build GRCh38
rs782022706 Chr.14: 105488680-105488680 on Build GRCh38
rs144550612 Chr.14: 105488485-105488485 on Build GRCh38
rs113160808 Chr.14: 105488519-105488519 on Build GRCh38
rs1037013212 Chr.14: 105488756-105488756 on Build GRCh38
rs782205997 Chr.14: 105488528-105488528 on Build GRCh38
rs782110616 Chr.14: 105488415-105488415 on Build GRCh38
rs782132838 Chr.14: 105488625-105488625 on Build GRCh38
rs782390379 Chr.14: 105488536-105488536 on Build GRCh38
rs782695231 Chr.14: 105488544-105488544 on Build GRCh38
rs782082040 Chr.14: 105488470-105488470 on Build GRCh38
rs782172947 Chr.14: 105488436-105488436 on Build GRCh38
rs200056933 Chr.14: 105488439-105488439 on Build GRCh38
rs200533449 Chr.14: 105488491-105488491 on Build GRCh38
rs924280000 Chr.14: 105488661-105488661 on Build GRCh38
rs762972577 Chr.14: 105488786-105488786 on Build GRCh38
rs138781112 Chr.14: 105488478-105488478 on Build GRCh38
rs782014612 Chr.14: 105488351-105488351 on Build GRCh38
rs7145303 Chr.14: 105488402-105488402 on Build GRCh38
rs142384687 Chr.14: 105488366-105488366 on Build GRCh38
rs782351069 Chr.14: 105488529-105488529 on Build GRCh38
rs587761042 Chr.14: 105488619-105488619 on Build GRCh38
rs781845027 Chr.14: 105488561-105488561 on Build GRCh38
rs587772737 Chr.14: 105488726-105488726 on Build GRCh38
rs782573055 Chr.14: 105488522-105488522 on Build GRCh38
rs781825871 Chr.14: 105488419-105488419 on Build GRCh38
rs781813302 Chr.14: 105488498-105488498 on Build GRCh38
rs141808882 Chr.14: 105488480-105488480 on Build GRCh38
rs782055319 Chr.14: 105488418-105488418 on Build GRCh38
rs200421273 Chr.14: 105488543-105488543 on Build GRCh38
rs587721357 Chr.14: 105488674-105488674 on Build GRCh38
rs781882431 Chr.14: 105488380-105488380 on Build GRCh38
rs782482745 Chr.14: 105488420-105488420 on Build GRCh38
rs977111713 Chr.14: 105488631-105488631 on Build GRCh38
rs782738199 Chr.14: 105488560-105488560 on Build GRCh38
rs782633985 Chr.14: 105488422-105488422 on Build GRCh38
rs782637357 Chr.14: 105488510-105488510 on Build GRCh38
rs782694651 Chr.14: 105488389-105488389 on Build GRCh38
rs782252951 Chr.14: 105488515-105488515 on Build GRCh38
rs782467361 Chr.14: 105488394-105488394 on Build GRCh38
rs949662997 Chr.14: 105488453-105488453 on Build GRCh38
rs372511951 Chr.14: 105488339-105488339 on Build GRCh38
rs971130874 Chr.14: 105488620-105488620 on Build GRCh38
rs782535705 Chr.14: 105488383-105488383 on Build GRCh38
rs995750701 Chr.14: 105488579-105488579 on Build GRCh38
rs782709731 Chr.14: 105488761-105488761 on Build GRCh38
rs782368147 Chr.14: 105488455-105488455 on Build GRCh38
rs782412472 Chr.14: 105488408-105488408 on Build GRCh38
rs782307574 Chr.14: 105488574-105488574 on Build GRCh38

View Full Product Details