Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 26544564-26545063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB3
Entrez Gene ID
2562
Gene Name
gamma-aminobutyric acid type A receptor beta3 subunit
Gene Aliases
ECA5
UniGene
Hs.302352
Gene Chromosome Location
Chr.15: 26543546-26773788 on Build GRCh38
Transcripts
NM_000814.5 NM_001191320.1 NM_001191321.2 NM_001278631.1 NM_021912.4 XM_011521428.2

SNP Detail

SNP ID Chromosome Location
rs947167053 Chr.15: 26544905-26544905 on Build GRCh38
rs552882518 Chr.15: 26544859-26544859 on Build GRCh38
rs140835040 Chr.15: 26544762-26544762 on Build GRCh38
rs184100810 Chr.15: 26544975-26544975 on Build GRCh38
rs547830778 Chr.15: 26544759-26544759 on Build GRCh38
rs551991431 Chr.15: 26544738-26544738 on Build GRCh38
rs186781745 Chr.15: 26544799-26544799 on Build GRCh38
rs1022660735 Chr.15: 26544619-26544620 on Build GRCh38
rs867693875 Chr.15: 26544824-26544824 on Build GRCh38
rs970198380 Chr.15: 26544641-26544641 on Build GRCh38
rs998603411 Chr.15: 26544948-26544948 on Build GRCh38
rs148266551 Chr.15: 26544935-26544935 on Build GRCh38
rs577681212 Chr.15: 26544949-26544949 on Build GRCh38
rs534322248 Chr.15: 26544624-26544624 on Build GRCh38
rs933179800 Chr.15: 26544604-26544604 on Build GRCh38
rs993680990 Chr.15: 26544884-26544884 on Build GRCh38
rs961416765 Chr.15: 26544845-26544845 on Build GRCh38
rs1035959197 Chr.15: 26544752-26544752 on Build GRCh38
rs988586671 Chr.15: 26545000-26545000 on Build GRCh38
rs28537095 Chr.15: 26544767-26544767 on Build GRCh38
rs116454913 Chr.15: 26544936-26544936 on Build GRCh38
rs888950903 Chr.15: 26544659-26544659 on Build GRCh38
rs1011367694 Chr.15: 26544723-26544723 on Build GRCh38
rs571410924 Chr.15: 26545028-26545031 on Build GRCh38
rs191581252 Chr.15: 26544904-26544904 on Build GRCh38
rs910590518 Chr.15: 26544790-26544790 on Build GRCh38
rs956036509 Chr.15: 26544999-26544999 on Build GRCh38
rs146998015 Chr.15: 26544898-26544900 on Build GRCh38
rs528165999 Chr.15: 26544758-26544758 on Build GRCh38
rs998903593 Chr.15: 26544749-26544749 on Build GRCh38
rs12917525 Chr.15: 26544966-26544966 on Build GRCh38
rs896050333 Chr.15: 26544916-26544916 on Build GRCh38
rs925774309 Chr.15: 26544733-26544733 on Build GRCh38
rs536961974 Chr.15: 26544796-26544796 on Build GRCh38
rs922469543 Chr.15: 26545016-26545016 on Build GRCh38
rs2912584 Chr.15: 26544742-26544742 on Build GRCh38
rs774120870 Chr.15: 26544629-26544629 on Build GRCh38
rs141378716 Chr.15: 26544993-26544993 on Build GRCh38
rs766170500 Chr.15: 26544974-26544974 on Build GRCh38
rs570554541 Chr.15: 26544810-26544810 on Build GRCh38
rs56398838 Chr.15: 26544597-26544597 on Build GRCh38
rs533542628 Chr.15: 26544734-26544734 on Build GRCh38
rs923155283 Chr.15: 26544589-26544589 on Build GRCh38
rs553695518 Chr.15: 26544772-26544773 on Build GRCh38
rs539578205 Chr.15: 26544849-26544849 on Build GRCh38
rs969788985 Chr.15: 26544953-26544954 on Build GRCh38
rs1013154787 Chr.15: 26545007-26545007 on Build GRCh38
rs1023139532 Chr.15: 26545019-26545019 on Build GRCh38
rs748007939 Chr.15: 26544717-26544717 on Build GRCh38
rs933274305 Chr.15: 26544860-26544860 on Build GRCh38
rs1050328010 Chr.15: 26544871-26544871 on Build GRCh38
rs760668429 Chr.15: 26544838-26544838 on Build GRCh38

View Full Product Details