Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 26937101-26937599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA5
Entrez Gene ID
2558
Gene Name
gamma-aminobutyric acid type A receptor alpha5 subunit
Gene Aliases
-
UniGene
Hs.24969
Gene Chromosome Location
Chr.15: 26866719-26949210 on Build GRCh38
Transcripts
NM_000810.3 NM_001165037.1 XM_005268258.2 XM_006720459.2 XM_017022055.1 XM_017022056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3956490 c.681G>C p.L227L Chr.15: 26937285-26937285 on Build GRCh38
5453140 c.606T>C p.V202V Chr.15: 26937210-26937210 on Build GRCh38
3956489 c.681G>A p.L227L Chr.15: 26937285-26937285 on Build GRCh38
6015666 c.615G>A p.W205* Chr.15: 26937219-26937219 on Build GRCh38
6293278 c.620A>G p.N207S Chr.15: 26937224-26937224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761530483 Chr.15: 26937373-26937373 on Build GRCh38
rs769772156 Chr.15: 26937302-26937302 on Build GRCh38
rs529004661 Chr.15: 26937361-26937361 on Build GRCh38
rs776868902 Chr.15: 26937290-26937290 on Build GRCh38
rs748424282 Chr.15: 26937142-26937142 on Build GRCh38
rs773598738 Chr.15: 26937320-26937321 on Build GRCh38
rs780724444 Chr.15: 26937576-26937576 on Build GRCh38
rs946542791 Chr.15: 26937488-26937488 on Build GRCh38
rs754056464 Chr.15: 26937448-26937448 on Build GRCh38
rs574250487 Chr.15: 26937155-26937155 on Build GRCh38
rs748560384 Chr.15: 26937232-26937232 on Build GRCh38
rs776753706 Chr.15: 26937188-26937188 on Build GRCh38
rs1029341769 Chr.15: 26937328-26937328 on Build GRCh38
rs111726286 Chr.15: 26937263-26937263 on Build GRCh38
rs1014795336 Chr.15: 26937575-26937575 on Build GRCh38
rs369047132 Chr.15: 26937257-26937257 on Build GRCh38
rs149156018 Chr.15: 26937189-26937189 on Build GRCh38
rs939263817 Chr.15: 26937535-26937535 on Build GRCh38
rs771305498 Chr.15: 26937353-26937353 on Build GRCh38
rs143301914 Chr.15: 26937344-26937344 on Build GRCh38
rs755540999 Chr.15: 26937226-26937226 on Build GRCh38
rs935109173 Chr.15: 26937502-26937503 on Build GRCh38
rs781495330 Chr.15: 26937354-26937354 on Build GRCh38
rs758841630 Chr.15: 26937346-26937346 on Build GRCh38
rs916827947 Chr.15: 26937395-26937395 on Build GRCh38
rs745970247 Chr.15: 26937357-26937357 on Build GRCh38
rs948798362 Chr.15: 26937567-26937567 on Build GRCh38
rs985136956 Chr.15: 26937432-26937432 on Build GRCh38
rs751197696 Chr.15: 26937213-26937213 on Build GRCh38
rs915042396 Chr.15: 26937240-26937240 on Build GRCh38
rs565133391 Chr.15: 26937343-26937343 on Build GRCh38
rs377729786 Chr.15: 26937336-26937336 on Build GRCh38
rs763792178 Chr.15: 26937323-26937323 on Build GRCh38
rs181785772 Chr.15: 26937207-26937207 on Build GRCh38
rs200310381 Chr.15: 26937225-26937225 on Build GRCh38
rs771187193 Chr.15: 26937259-26937259 on Build GRCh38
rs767116452 Chr.15: 26937338-26937338 on Build GRCh38
rs1057191257 Chr.15: 26937543-26937543 on Build GRCh38
rs769748036 Chr.15: 26937358-26937358 on Build GRCh38
rs374261310 Chr.15: 26937345-26937345 on Build GRCh38
rs375240262 Chr.15: 26937208-26937208 on Build GRCh38
rs374318313 Chr.15: 26937297-26937297 on Build GRCh38
rs773252977 Chr.15: 26937150-26937150 on Build GRCh38
rs754287701 Chr.15: 26937274-26937274 on Build GRCh38
rs978317662 Chr.15: 26937504-26937504 on Build GRCh38
rs961910127 Chr.15: 26937372-26937372 on Build GRCh38
rs772314090 Chr.15: 26937248-26937248 on Build GRCh38
rs757922277 Chr.15: 26937279-26937279 on Build GRCh38
rs189562874 Chr.15: 26937360-26937360 on Build GRCh38
rs769292515 Chr.15: 26937319-26937320 on Build GRCh38
rs372141813 Chr.15: 26937135-26937135 on Build GRCh38
rs972730565 Chr.15: 26937163-26937163 on Build GRCh38
rs946628458 Chr.15: 26937246-26937246 on Build GRCh38
rs61999613 Chr.15: 26937509-26937509 on Build GRCh38
rs760898058 Chr.15: 26937156-26937156 on Build GRCh38
rs1042285046 Chr.15: 26937255-26937255 on Build GRCh38
rs901753362 Chr.15: 26937288-26937288 on Build GRCh38
rs778210544 Chr.15: 26937252-26937252 on Build GRCh38
rs140682 Chr.15: 26937210-26937210 on Build GRCh38
rs775231235 Chr.15: 26937315-26937315 on Build GRCh38
rs749942488 Chr.15: 26937339-26937339 on Build GRCh38
rs569072408 Chr.15: 26937423-26937423 on Build GRCh38
rs778263902 Chr.15: 26937348-26937348 on Build GRCh38
rs751231443 Chr.15: 26937438-26937438 on Build GRCh38
rs551680323 Chr.15: 26937531-26937531 on Build GRCh38
rs772345251 Chr.15: 26937149-26937149 on Build GRCh38
rs201843662 Chr.15: 26937364-26937364 on Build GRCh38
rs368209283 Chr.15: 26937199-26937199 on Build GRCh38
rs1018598218 Chr.15: 26937287-26937287 on Build GRCh38
rs779223735 Chr.15: 26937228-26937228 on Build GRCh38
rs762861847 Chr.15: 26937317-26937317 on Build GRCh38
rs371001870 Chr.15: 26937351-26937351 on Build GRCh38
rs895787497 Chr.15: 26937544-26937544 on Build GRCh38
rs922263611 Chr.15: 26937374-26937374 on Build GRCh38
rs186262515 Chr.15: 26937214-26937214 on Build GRCh38
rs766567820 Chr.15: 26937178-26937178 on Build GRCh38

View Full Product Details