Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 26939847-26940345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA5
Entrez Gene ID
2558
Gene Name
gamma-aminobutyric acid type A receptor alpha5 subunit
Gene Aliases
-
UniGene
Hs.24969
Gene Chromosome Location
Chr.15: 26866719-26949210 on Build GRCh38
Transcripts
NM_000810.3 NM_001165037.1 XM_005268258.2 XM_006720459.2 XM_017022055.1 XM_017022056.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6251658 c.843G>T p.L281L Chr.15: 26940043-26940043 on Build GRCh38
700319 c.873T>G p.V291V Chr.15: 26940073-26940073 on Build GRCh38
4633961 c.839G>A p.W280* Chr.15: 26940039-26940039 on Build GRCh38
6219196 c.792C>G p.T264T Chr.15: 26939992-26939992 on Build GRCh38
5462432 c.725-8A>G p.? Chr.15: 26939917-26939917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559028221 Chr.15: 26940300-26940300 on Build GRCh38
rs745607505 Chr.15: 26939981-26939981 on Build GRCh38
rs1033956987 Chr.15: 26940160-26940160 on Build GRCh38
rs572228912 Chr.15: 26939974-26939974 on Build GRCh38
rs867999464 Chr.15: 26940011-26940011 on Build GRCh38
rs892593050 Chr.15: 26940116-26940116 on Build GRCh38
rs765407468 Chr.15: 26939920-26939920 on Build GRCh38
rs781181456 Chr.15: 26940164-26940164 on Build GRCh38
rs1025155116 Chr.15: 26940136-26940136 on Build GRCh38
rs777014928 Chr.15: 26940093-26940093 on Build GRCh38
rs547854807 Chr.15: 26939890-26939890 on Build GRCh38
rs373466562 Chr.15: 26940091-26940091 on Build GRCh38
rs756159250 Chr.15: 26940109-26940115 on Build GRCh38
rs778729060 Chr.15: 26939934-26939934 on Build GRCh38
rs917493258 Chr.15: 26940165-26940165 on Build GRCh38
rs761012597 Chr.15: 26939894-26939894 on Build GRCh38
rs144516237 Chr.15: 26940285-26940285 on Build GRCh38
rs375293570 Chr.15: 26939921-26939921 on Build GRCh38
rs757039936 Chr.15: 26939959-26939959 on Build GRCh38
rs772470476 Chr.15: 26939889-26939889 on Build GRCh38
rs775862695 Chr.15: 26940304-26940304 on Build GRCh38
rs369568018 Chr.15: 26939926-26939926 on Build GRCh38
rs773633746 Chr.15: 26940087-26940087 on Build GRCh38
rs771296427 Chr.15: 26940092-26940092 on Build GRCh38
rs747303013 Chr.15: 26940148-26940148 on Build GRCh38
rs753789146 Chr.15: 26939878-26939878 on Build GRCh38
rs796997732 Chr.15: 26940252-26940252 on Build GRCh38
rs561136227 Chr.15: 26940104-26940104 on Build GRCh38
rs1030009252 Chr.15: 26940067-26940067 on Build GRCh38
rs368673573 Chr.15: 26940013-26940013 on Build GRCh38
rs767843158 Chr.15: 26939896-26939897 on Build GRCh38
rs773756749 Chr.15: 26939891-26939891 on Build GRCh38
rs778129106 Chr.15: 26940135-26940135 on Build GRCh38
rs528634721 Chr.15: 26940301-26940301 on Build GRCh38
rs955202805 Chr.15: 26940082-26940082 on Build GRCh38
rs751506953 Chr.15: 26939943-26939943 on Build GRCh38
rs775161893 Chr.15: 26939881-26939881 on Build GRCh38
rs368433344 Chr.15: 26939904-26939904 on Build GRCh38
rs943303821 Chr.15: 26940005-26940005 on Build GRCh38
rs28551016 Chr.15: 26940159-26940159 on Build GRCh38
rs963949932 Chr.15: 26940167-26940167 on Build GRCh38
rs1000902749 Chr.15: 26940098-26940098 on Build GRCh38
rs779796035 Chr.15: 26940025-26940025 on Build GRCh38
rs754894074 Chr.15: 26939888-26939888 on Build GRCh38
rs765454531 Chr.15: 26940123-26940123 on Build GRCh38
rs1010086417 Chr.15: 26940058-26940058 on Build GRCh38
rs758442427 Chr.15: 26939935-26939935 on Build GRCh38
rs752878081 Chr.15: 26939910-26939910 on Build GRCh38
rs563294388 Chr.15: 26940191-26940191 on Build GRCh38
rs752440270 Chr.15: 26939936-26939936 on Build GRCh38
rs371995516 Chr.15: 26939910-26939910 on Build GRCh38
rs768209346 Chr.15: 26940055-26940055 on Build GRCh38
rs74006534 Chr.15: 26940144-26940144 on Build GRCh38
rs1025899601 Chr.15: 26940270-26940270 on Build GRCh38
rs748866335 Chr.15: 26940029-26940029 on Build GRCh38
rs41309256 Chr.15: 26940210-26940210 on Build GRCh38
rs548390755 Chr.15: 26940318-26940318 on Build GRCh38
rs376568358 Chr.15: 26939901-26939901 on Build GRCh38
rs770239309 Chr.15: 26940250-26940250 on Build GRCh38

View Full Product Details