Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30382936-30383450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRFAM7A
Entrez Gene ID
89832
Gene Name
CHRNA7 (exons 5-10) and FAM7A (exons A-E) fusion
Gene Aliases
CHRNA7, CHRNA7-DR1, D-10
UniGene
Hs.510853
Gene Chromosome Location
Chr.15: 30357770-30393849 on Build GRCh38
Transcripts
NM_139320.1 NM_148911.1 XM_005254750.2 XM_011522153.1 XM_011522155.2

SNP Detail

SNP ID Chromosome Location
rs4043148 Chr.15: 30383390-30383391 on Build GRCh38
rs767087680 Chr.15: 30383339-30383339 on Build GRCh38
rs917181553 Chr.15: 30383041-30383041 on Build GRCh38
rs755906401 Chr.15: 30383281-30383281 on Build GRCh38
rs773771160 Chr.15: 30383401-30383401 on Build GRCh38
rs373716749 Chr.15: 30383091-30383091 on Build GRCh38
rs112266675 Chr.15: 30383309-30383309 on Build GRCh38
rs765983923 Chr.15: 30383389-30383389 on Build GRCh38
rs371249914 Chr.15: 30383390-30383390 on Build GRCh38
rs770139221 Chr.15: 30383391-30383391 on Build GRCh38
rs755367799 Chr.15: 30383382-30383382 on Build GRCh38
rs780430375 Chr.15: 30383297-30383297 on Build GRCh38
rs958847140 Chr.15: 30383024-30383024 on Build GRCh38
rs3100759 Chr.15: 30383399-30383399 on Build GRCh38
rs377464096 Chr.15: 30383268-30383268 on Build GRCh38
rs757959379 Chr.15: 30383306-30383306 on Build GRCh38
rs758493209 Chr.15: 30383296-30383296 on Build GRCh38
rs201569187 Chr.15: 30383378-30383378 on Build GRCh38
rs748647270 Chr.15: 30383329-30383329 on Build GRCh38
rs747376372 Chr.15: 30383299-30383299 on Build GRCh38
rs778818814 Chr.15: 30383289-30383289 on Build GRCh38
rs570439947 Chr.15: 30383362-30383362 on Build GRCh38
rs745984236 Chr.15: 30383290-30383290 on Build GRCh38
rs773682600 Chr.15: 30383337-30383337 on Build GRCh38
rs753698111 Chr.15: 30383363-30383363 on Build GRCh38
rs747286636 Chr.15: 30383379-30383379 on Build GRCh38
rs781651933 Chr.15: 30383383-30383383 on Build GRCh38
rs775188957 Chr.15: 30383342-30383342 on Build GRCh38
rs750404721 Chr.15: 30383373-30383373 on Build GRCh38
rs745363590 Chr.15: 30383403-30383403 on Build GRCh38
rs796714144 Chr.15: 30382960-30382960 on Build GRCh38
rs290158 Chr.15: 30383046-30383046 on Build GRCh38
rs757094906 Chr.15: 30383371-30383371 on Build GRCh38
rs532949924 Chr.15: 30383372-30383372 on Build GRCh38
rs200486343 Chr.15: 30383349-30383349 on Build GRCh38
rs967174248 Chr.15: 30383065-30383065 on Build GRCh38
rs202209324 Chr.15: 30383302-30383302 on Build GRCh38
rs2564621 Chr.15: 30383350-30383350 on Build GRCh38
rs992739125 Chr.15: 30383037-30383038 on Build GRCh38
rs200181885 Chr.15: 30383403-30383404 on Build GRCh38
rs754469476 Chr.15: 30383391-30383392 on Build GRCh38
rs753829424 Chr.15: 30383283-30383283 on Build GRCh38
rs750142634 Chr.15: 30383278-30383281 on Build GRCh38
rs777259274 Chr.15: 30383314-30383314 on Build GRCh38
rs751103264 Chr.15: 30383391-30383391 on Build GRCh38
rs781771698 Chr.15: 30383404-30383404 on Build GRCh38
rs757337854 Chr.15: 30383286-30383286 on Build GRCh38
rs758573687 Chr.15: 30383377-30383377 on Build GRCh38
rs770360671 Chr.15: 30383331-30383331 on Build GRCh38

View Full Product Details