Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32615933-32616428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP11A
Entrez Gene ID
9824
Gene Name
Rho GTPase activating protein 11A
Gene Aliases
GAP (1-12)
UniGene
Hs.591130
Gene Chromosome Location
Chr.15: 32615144-32639949 on Build GRCh38
Transcripts
NM_001286479.1 NM_001286480.1 NM_014783.4 NM_199357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5848993 c.129+1G>A p.? Chr.15: 32616341-32616341 on Build GRCh38
126556 c.6G>A p.W2* Chr.15: 32616217-32616217 on Build GRCh38
5687470 c.33G>T p.L11L Chr.15: 32616244-32616244 on Build GRCh38
4578162 c.62T>C p.I21T Chr.15: 32616273-32616273 on Build GRCh38
4662710 c.80G>A p.R27H Chr.15: 32616291-32616291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767019297 Chr.15: 32616383-32616383 on Build GRCh38
rs986475136 Chr.15: 32616200-32616200 on Build GRCh38
rs1054603135 Chr.15: 32615997-32615997 on Build GRCh38
rs771026086 Chr.15: 32616189-32616189 on Build GRCh38
rs4353456 Chr.15: 32616405-32616405 on Build GRCh38
rs925506846 Chr.15: 32616176-32616176 on Build GRCh38
rs775625870 Chr.15: 32616336-32616336 on Build GRCh38
rs928939621 Chr.15: 32616279-32616279 on Build GRCh38
rs149871501 Chr.15: 32616267-32616267 on Build GRCh38
rs759280343 Chr.15: 32616353-32616353 on Build GRCh38
rs532723102 Chr.15: 32616334-32616334 on Build GRCh38
rs778973451 Chr.15: 32616291-32616291 on Build GRCh38
rs759772503 Chr.15: 32616197-32616197 on Build GRCh38
rs4470116 Chr.15: 32616234-32616234 on Build GRCh38
rs368253364 Chr.15: 32616387-32616387 on Build GRCh38
rs774361730 Chr.15: 32616191-32616191 on Build GRCh38
rs1034179880 Chr.15: 32616093-32616093 on Build GRCh38
rs200942090 Chr.15: 32616231-32616231 on Build GRCh38
rs749189484 Chr.15: 32616188-32616188 on Build GRCh38
rs777617949 Chr.15: 32616181-32616181 on Build GRCh38
rs936829747 Chr.15: 32616189-32616190 on Build GRCh38
rs575507338 Chr.15: 32616077-32616077 on Build GRCh38
rs751046100 Chr.15: 32616236-32616236 on Build GRCh38
rs368624593 Chr.15: 32616173-32616173 on Build GRCh38
rs749242605 Chr.15: 32616275-32616275 on Build GRCh38
rs962828796 Chr.15: 32616073-32616073 on Build GRCh38
rs4255759 Chr.15: 32616402-32616402 on Build GRCh38
rs754505443 Chr.15: 32616237-32616237 on Build GRCh38
rs562999699 Chr.15: 32616365-32616365 on Build GRCh38
rs772122169 Chr.15: 32616218-32616218 on Build GRCh38
rs1045264376 Chr.15: 32616400-32616400 on Build GRCh38
rs760922820 Chr.15: 32616357-32616357 on Build GRCh38
rs777500985 Chr.15: 32616273-32616273 on Build GRCh38
rs1050766670 Chr.15: 32616251-32616251 on Build GRCh38
rs865874118 Chr.15: 32616092-32616092 on Build GRCh38
rs762370333 Chr.15: 32616233-32616233 on Build GRCh38
rs968344650 Chr.15: 32616230-32616230 on Build GRCh38
rs374567702 Chr.15: 32616351-32616351 on Build GRCh38
rs571090423 Chr.15: 32615959-32615959 on Build GRCh38
rs769615783 Chr.15: 32616174-32616174 on Build GRCh38
rs924317020 Chr.15: 32616235-32616235 on Build GRCh38
rs752371262 Chr.15: 32616259-32616259 on Build GRCh38
rs917890052 Chr.15: 32616121-32616121 on Build GRCh38
rs773841329 Chr.15: 32616374-32616374 on Build GRCh38
rs1015440508 Chr.15: 32616064-32616064 on Build GRCh38
rs887757626 Chr.15: 32616089-32616089 on Build GRCh38
rs768933545 Chr.15: 32616358-32616358 on Build GRCh38
rs1045026861 Chr.15: 32616035-32616035 on Build GRCh38
rs959427155 Chr.15: 32616098-32616098 on Build GRCh38
rs770793060 Chr.15: 32616289-32616289 on Build GRCh38
rs139475473 Chr.15: 32616250-32616250 on Build GRCh38
rs372331657 Chr.15: 32616210-32616210 on Build GRCh38
rs761006120 Chr.15: 32616223-32616223 on Build GRCh38
rs556943450 Chr.15: 32616010-32616010 on Build GRCh38
rs746017840 Chr.15: 32616318-32616318 on Build GRCh38
rs953306301 Chr.15: 32616186-32616186 on Build GRCh38
rs1029613656 Chr.15: 32616018-32616018 on Build GRCh38
rs145284623 Chr.15: 32615963-32615963 on Build GRCh38
rs1004141007 Chr.15: 32616290-32616290 on Build GRCh38
rs1004109913 Chr.15: 32616056-32616056 on Build GRCh38
rs759015570 Chr.15: 32616380-32616380 on Build GRCh38
rs777087649 Chr.15: 32616359-32616359 on Build GRCh38
rs1057074951 Chr.15: 32615961-32615961 on Build GRCh38
rs186897838 Chr.15: 32616100-32616100 on Build GRCh38
rs899535993 Chr.15: 32616000-32616000 on Build GRCh38
rs774133034 Chr.15: 32616352-32616352 on Build GRCh38
rs765911064 Chr.15: 32616367-32616367 on Build GRCh38
rs193237097 Chr.15: 32616159-32616159 on Build GRCh38
rs764499874 Chr.15: 32616226-32616226 on Build GRCh38
rs781044661 Chr.15: 32616171-32616171 on Build GRCh38
rs565405976 Chr.15: 32616272-32616272 on Build GRCh38
rs974218960 Chr.15: 32616075-32616075 on Build GRCh38

View Full Product Details