Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32766151-32766653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMN1
Entrez Gene ID
342184
Gene Name
formin 1
Gene Aliases
FMN, LD
UniGene
Hs.657649
Gene Chromosome Location
Chr.15: 32765544-33194765 on Build GRCh38
Transcripts
NM_001103184.3 NM_001277313.1 XM_011521504.2 XM_011521506.2 XM_011521509.2 XM_011521511.2 XM_017022130.1 XM_017022132.1 XM_017022133.1 XM_017022134.1 XM_017022135.1

SNP Detail

SNP ID Chromosome Location
rs998070288 Chr.15: 32766259-32766259 on Build GRCh38
rs969583587 Chr.15: 32766320-32766320 on Build GRCh38
rs921829041 Chr.15: 32766393-32766393 on Build GRCh38
rs573208604 Chr.15: 32766598-32766598 on Build GRCh38
rs1042549703 Chr.15: 32766444-32766444 on Build GRCh38
rs944634195 Chr.15: 32766371-32766371 on Build GRCh38
rs1054043528 Chr.15: 32766416-32766416 on Build GRCh38
rs928131157 Chr.15: 32766323-32766323 on Build GRCh38
rs112587170 Chr.15: 32766219-32766219 on Build GRCh38
rs1004106233 Chr.15: 32766465-32766465 on Build GRCh38
rs1017014852 Chr.15: 32766546-32766546 on Build GRCh38
rs181442732 Chr.15: 32766339-32766339 on Build GRCh38
rs960921282 Chr.15: 32766340-32766340 on Build GRCh38
rs557462742 Chr.15: 32766452-32766452 on Build GRCh38
rs202080717 Chr.15: 32766541-32766541 on Build GRCh38
rs534977052 Chr.15: 32766277-32766277 on Build GRCh38
rs1033958743 Chr.15: 32766480-32766480 on Build GRCh38
rs56756022 Chr.15: 32766533-32766533 on Build GRCh38
rs557364664 Chr.15: 32766211-32766211 on Build GRCh38
rs1258723 Chr.15: 32766387-32766387 on Build GRCh38
rs370395969 Chr.15: 32766533-32766534 on Build GRCh38
rs540587472 Chr.15: 32766621-32766621 on Build GRCh38
rs1006092953 Chr.15: 32766433-32766442 on Build GRCh38
rs891553300 Chr.15: 32766421-32766421 on Build GRCh38
rs564471650 Chr.15: 32766468-32766468 on Build GRCh38
rs749742598 Chr.15: 32766530-32766540 on Build GRCh38
rs914183279 Chr.15: 32766390-32766390 on Build GRCh38
rs911704693 Chr.15: 32766324-32766324 on Build GRCh38
rs994872565 Chr.15: 32766572-32766572 on Build GRCh38
rs935721537 Chr.15: 32766403-32766403 on Build GRCh38
rs12148620 Chr.15: 32766322-32766322 on Build GRCh38
rs12148200 Chr.15: 32766234-32766234 on Build GRCh38
rs575932139 Chr.15: 32766221-32766221 on Build GRCh38
rs1005216450 Chr.15: 32766545-32766545 on Build GRCh38
rs950745484 Chr.15: 32766305-32766305 on Build GRCh38
rs895383660 Chr.15: 32766524-32766524 on Build GRCh38
rs973382129 Chr.15: 32766236-32766236 on Build GRCh38
rs781082348 Chr.15: 32766293-32766293 on Build GRCh38
rs554794991 Chr.15: 32766569-32766569 on Build GRCh38
rs16958518 Chr.15: 32766190-32766190 on Build GRCh38
rs796286728 Chr.15: 32766286-32766286 on Build GRCh38
rs1025239843 Chr.15: 32766281-32766281 on Build GRCh38
rs200723918 Chr.15: 32766534-32766534 on Build GRCh38
rs769264402 Chr.15: 32766349-32766349 on Build GRCh38
rs575739518 Chr.15: 32766518-32766518 on Build GRCh38
rs796258894 Chr.15: 32766540-32766540 on Build GRCh38
rs758430358 Chr.15: 32766185-32766185 on Build GRCh38
rs983779727 Chr.15: 32766310-32766310 on Build GRCh38
rs55859863 Chr.15: 32766531-32766531 on Build GRCh38
rs945759329 Chr.15: 32766423-32766423 on Build GRCh38
rs1009722635 Chr.15: 32766178-32766178 on Build GRCh38
rs201266335 Chr.15: 32766540-32766540 on Build GRCh38
rs946761055 Chr.15: 32766187-32766187 on Build GRCh38
rs745677212 Chr.15: 32766530-32766531 on Build GRCh38
rs969132333 Chr.15: 32766569-32766573 on Build GRCh38
rs1034109485 Chr.15: 32766225-32766225 on Build GRCh38
rs950797831 Chr.15: 32766623-32766623 on Build GRCh38

View Full Product Details