Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32768048-32768590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMN1
Entrez Gene ID
342184
Gene Name
formin 1
Gene Aliases
FMN, LD
UniGene
Hs.657649
Gene Chromosome Location
Chr.15: 32765544-33194765 on Build GRCh38
Transcripts
NM_001103184.3 NM_001277313.1 XM_011521504.2 XM_011521506.2 XM_011521509.2 XM_011521511.2 XM_017022130.1 XM_017022132.1 XM_017022133.1 XM_017022134.1 XM_017022135.1

SNP Detail

SNP ID Chromosome Location
rs763101317 Chr.15: 32768507-32768507 on Build GRCh38
rs1014589318 Chr.15: 32768462-32768462 on Build GRCh38
rs948571711 Chr.15: 32768440-32768440 on Build GRCh38
rs962143151 Chr.15: 32768483-32768484 on Build GRCh38
rs28616589 Chr.15: 32768381-32768381 on Build GRCh38
rs1021868003 Chr.15: 32768366-32768366 on Build GRCh38
rs753456063 Chr.15: 32768202-32768202 on Build GRCh38
rs140390606 Chr.15: 32768390-32768390 on Build GRCh38
rs908066665 Chr.15: 32768081-32768081 on Build GRCh38
rs747657281 Chr.15: 32768306-32768306 on Build GRCh38
rs576989632 Chr.15: 32768523-32768523 on Build GRCh38
rs987568580 Chr.15: 32768073-32768073 on Build GRCh38
rs983945909 Chr.15: 32768453-32768453 on Build GRCh38
rs550670130 Chr.15: 32768214-32768214 on Build GRCh38
rs183580825 Chr.15: 32768405-32768405 on Build GRCh38
rs1008527064 Chr.15: 32768460-32768460 on Build GRCh38
rs544476706 Chr.15: 32768529-32768529 on Build GRCh38
rs1030928888 Chr.15: 32768319-32768319 on Build GRCh38
rs779860334 Chr.15: 32768145-32768145 on Build GRCh38
rs11856949 Chr.15: 32768320-32768320 on Build GRCh38
rs550184413 Chr.15: 32768190-32768192 on Build GRCh38
rs374955284 Chr.15: 32768352-32768352 on Build GRCh38
rs1036665323 Chr.15: 32768286-32768286 on Build GRCh38
rs562876556 Chr.15: 32768247-32768247 on Build GRCh38
rs1049536137 Chr.15: 32768552-32768552 on Build GRCh38
rs1003913523 Chr.15: 32768285-32768285 on Build GRCh38
rs958967371 Chr.15: 32768513-32768513 on Build GRCh38
rs552755387 Chr.15: 32768355-32768355 on Build GRCh38
rs540108247 Chr.15: 32768293-32768293 on Build GRCh38
rs562678818 Chr.15: 32768532-32768532 on Build GRCh38
rs977385409 Chr.15: 32768103-32768103 on Build GRCh38
rs902329484 Chr.15: 32768268-32768268 on Build GRCh38
rs114753203 Chr.15: 32768505-32768505 on Build GRCh38
rs1055774428 Chr.15: 32768370-32768370 on Build GRCh38
rs1024790679 Chr.15: 32768264-32768264 on Build GRCh38
rs940860188 Chr.15: 32768327-32768327 on Build GRCh38
rs933633676 Chr.15: 32768533-32768533 on Build GRCh38
rs186917684 Chr.15: 32768494-32768494 on Build GRCh38
rs190392513 Chr.15: 32768264-32768264 on Build GRCh38
rs28574716 Chr.15: 32768387-32768387 on Build GRCh38
rs974398595 Chr.15: 32768290-32768290 on Build GRCh38
rs529992035 Chr.15: 32768561-32768561 on Build GRCh38
rs879130620 Chr.15: 32768118-32768118 on Build GRCh38
rs1037942920 Chr.15: 32768157-32768157 on Build GRCh38
rs1028887801 Chr.15: 32768436-32768436 on Build GRCh38
rs895869311 Chr.15: 32768237-32768237 on Build GRCh38
rs1048363150 Chr.15: 32768274-32768274 on Build GRCh38
rs938742221 Chr.15: 32768191-32768191 on Build GRCh38
rs568482092 Chr.15: 32768393-32768396 on Build GRCh38
rs920816459 Chr.15: 32768354-32768354 on Build GRCh38
rs548275454 Chr.15: 32768289-32768289 on Build GRCh38
rs775044110 Chr.15: 32768494-32768498 on Build GRCh38
rs866607984 Chr.15: 32768401-32768401 on Build GRCh38
rs1014360674 Chr.15: 32768257-32768257 on Build GRCh38
rs915719934 Chr.15: 32768239-32768239 on Build GRCh38
rs1020561 Chr.15: 32768400-32768400 on Build GRCh38
rs76542275 Chr.15: 32768377-32768377 on Build GRCh38
rs62002631 Chr.15: 32768379-32768379 on Build GRCh38
rs2840190 Chr.15: 32768206-32768206 on Build GRCh38
rs939879805 Chr.15: 32768506-32768506 on Build GRCh38
rs866022414 Chr.15: 32768467-32768467 on Build GRCh38
rs564990906 Chr.15: 32768102-32768102 on Build GRCh38
rs572982243 Chr.15: 32768456-32768456 on Build GRCh38
rs888360978 Chr.15: 32768280-32768280 on Build GRCh38

View Full Product Details