Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34140285-34140805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNBL1
Entrez Gene ID
79768
Gene Name
katanin regulatory subunit B1 like 1
Gene Aliases
C15orf29
UniGene
Hs.633566
Gene Chromosome Location
Chr.15: 34140674-34210146 on Build GRCh38
Transcripts
NM_024713.2 XM_017022571.1 XM_017022572.1 XM_017022573.1 XM_017022574.1 XM_017022575.1

SNP Detail

SNP ID Chromosome Location
rs1038507786 Chr.15: 34140563-34140563 on Build GRCh38
rs899662261 Chr.15: 34140370-34140370 on Build GRCh38
rs769957427 Chr.15: 34140594-34140594 on Build GRCh38
rs915606502 Chr.15: 34140427-34140427 on Build GRCh38
rs563146399 Chr.15: 34140648-34140648 on Build GRCh38
rs141432347 Chr.15: 34140370-34140371 on Build GRCh38
rs548562441 Chr.15: 34140778-34140778 on Build GRCh38
rs752640512 Chr.15: 34140415-34140415 on Build GRCh38
rs917441437 Chr.15: 34140526-34140526 on Build GRCh38
rs899940515 Chr.15: 34140661-34140661 on Build GRCh38
rs758321493 Chr.15: 34140484-34140484 on Build GRCh38
rs73376158 Chr.15: 34140653-34140653 on Build GRCh38
rs886831963 Chr.15: 34140493-34140493 on Build GRCh38
rs867781144 Chr.15: 34140406-34140406 on Build GRCh38
rs1004147675 Chr.15: 34140428-34140428 on Build GRCh38
rs992089932 Chr.15: 34140486-34140486 on Build GRCh38
rs577349092 Chr.15: 34140456-34140456 on Build GRCh38
rs749464511 Chr.15: 34140725-34140725 on Build GRCh38
rs573953929 Chr.15: 34140421-34140421 on Build GRCh38
rs936023232 Chr.15: 34140610-34140610 on Build GRCh38
rs997115059 Chr.15: 34140404-34140404 on Build GRCh38
rs950317711 Chr.15: 34140554-34140554 on Build GRCh38
rs977675704 Chr.15: 34140565-34140565 on Build GRCh38
rs879075573 Chr.15: 34140394-34140394 on Build GRCh38
rs796248780 Chr.15: 34140368-34140369 on Build GRCh38
rs780430623 Chr.15: 34140722-34140722 on Build GRCh38
rs745971754 Chr.15: 34140557-34140557 on Build GRCh38
rs1028568459 Chr.15: 34140658-34140658 on Build GRCh38
rs781181752 Chr.15: 34140530-34140530 on Build GRCh38
rs544834239 Chr.15: 34140544-34140544 on Build GRCh38
rs1045907769 Chr.15: 34140483-34140483 on Build GRCh38
rs924565388 Chr.15: 34140590-34140590 on Build GRCh38
rs543103904 Chr.15: 34140738-34140738 on Build GRCh38
rs1021472847 Chr.15: 34140729-34140729 on Build GRCh38
rs925881254 Chr.15: 34140324-34140324 on Build GRCh38
rs891146758 Chr.15: 34140673-34140673 on Build GRCh38
rs949729092 Chr.15: 34140457-34140457 on Build GRCh38
rs1033422916 Chr.15: 34140476-34140476 on Build GRCh38
rs565017804 Chr.15: 34140423-34140423 on Build GRCh38

View Full Product Details