Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34141077-34141589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNBL1
Entrez Gene ID
79768
Gene Name
katanin regulatory subunit B1 like 1
Gene Aliases
C15orf29
UniGene
Hs.633566
Gene Chromosome Location
Chr.15: 34140674-34210146 on Build GRCh38
Transcripts
NM_024713.2 XM_017022571.1 XM_017022572.1 XM_017022573.1 XM_017022574.1 XM_017022575.1

SNP Detail

SNP ID Chromosome Location
rs150902114 Chr.15: 34141410-34141410 on Build GRCh38
rs768747135 Chr.15: 34141399-34141399 on Build GRCh38
rs546196032 Chr.15: 34141559-34141559 on Build GRCh38
rs55792000 Chr.15: 34141113-34141113 on Build GRCh38
rs773669330 Chr.15: 34141340-34141340 on Build GRCh38
rs17817770 Chr.15: 34141146-34141146 on Build GRCh38
rs1012705983 Chr.15: 34141538-34141538 on Build GRCh38
rs1051192151 Chr.15: 34141107-34141107 on Build GRCh38
rs577034281 Chr.15: 34141193-34141193 on Build GRCh38
rs762267584 Chr.15: 34141252-34141252 on Build GRCh38
rs572726574 Chr.15: 34141118-34141118 on Build GRCh38
rs987282301 Chr.15: 34141489-34141489 on Build GRCh38
rs563083001 Chr.15: 34141375-34141375 on Build GRCh38
rs527671238 Chr.15: 34141496-34141496 on Build GRCh38
rs1043025868 Chr.15: 34141380-34141380 on Build GRCh38
rs139382345 Chr.15: 34141419-34141419 on Build GRCh38
rs957582636 Chr.15: 34141397-34141397 on Build GRCh38
rs544768500 Chr.15: 34141326-34141326 on Build GRCh38
rs1013710817 Chr.15: 34141319-34141319 on Build GRCh38
rs772655531 Chr.15: 34141260-34141260 on Build GRCh38
rs895200481 Chr.15: 34141499-34141499 on Build GRCh38
rs1024762788 Chr.15: 34141334-34141335 on Build GRCh38
rs560869363 Chr.15: 34141445-34141445 on Build GRCh38
rs981760106 Chr.15: 34141176-34141176 on Build GRCh38
rs999050506 Chr.15: 34141357-34141357 on Build GRCh38
rs761760593 Chr.15: 34141217-34141217 on Build GRCh38
rs894817860 Chr.15: 34141284-34141284 on Build GRCh38
rs944342796 Chr.15: 34141346-34141346 on Build GRCh38
rs181832917 Chr.15: 34141184-34141184 on Build GRCh38
rs773844333 Chr.15: 34141201-34141202 on Build GRCh38
rs961319690 Chr.15: 34141201-34141201 on Build GRCh38
rs973844543 Chr.15: 34141245-34141245 on Build GRCh38
rs1049890526 Chr.15: 34141269-34141269 on Build GRCh38
rs554891952 Chr.15: 34141114-34141114 on Build GRCh38
rs1031863750 Chr.15: 34141367-34141367 on Build GRCh38

View Full Product Details