Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34355397-34355890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUTM1
Entrez Gene ID
256646
Gene Name
NUT midline carcinoma family member 1
Gene Aliases
C15orf55, FAM22H, NUT
UniGene
Hs.525769
Gene Chromosome Location
Chr.15: 34343315-34357737 on Build GRCh38
Transcripts
NM_001284292.1 NM_001284293.1 NM_175741.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
349024 c.1423G>A p.E475K Chr.15: 34355515-34355515 on Build GRCh38
5673112 c.1699C>T p.P567S Chr.15: 34355791-34355791 on Build GRCh38
1707979 c.1504G>A p.E502K Chr.15: 34355596-34355596 on Build GRCh38
1730726 c.1772A>G p.N591S Chr.15: 34355864-34355864 on Build GRCh38
6030687 c.1594C>T p.R532W Chr.15: 34355686-34355686 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763263640 Chr.15: 34355738-34355738 on Build GRCh38
rs765336791 Chr.15: 34355733-34355733 on Build GRCh38
rs755304945 Chr.15: 34355748-34355748 on Build GRCh38
rs745591416 Chr.15: 34355809-34355809 on Build GRCh38
rs747772079 Chr.15: 34355468-34355471 on Build GRCh38
rs201182294 Chr.15: 34355649-34355649 on Build GRCh38
rs773608332 Chr.15: 34355734-34355734 on Build GRCh38
rs144692942 Chr.15: 34355668-34355668 on Build GRCh38
rs771898661 Chr.15: 34355699-34355699 on Build GRCh38
rs759359911 Chr.15: 34355627-34355627 on Build GRCh38
rs752592889 Chr.15: 34355563-34355563 on Build GRCh38
rs771808676 Chr.15: 34355630-34355630 on Build GRCh38
rs61737339 Chr.15: 34355502-34355502 on Build GRCh38
rs780970802 Chr.15: 34355462-34355465 on Build GRCh38
rs769831562 Chr.15: 34355827-34355827 on Build GRCh38
rs35304501 Chr.15: 34355746-34355747 on Build GRCh38
rs548347433 Chr.15: 34355524-34355524 on Build GRCh38
rs746440939 Chr.15: 34355477-34355477 on Build GRCh38
rs768479594 Chr.15: 34355716-34355716 on Build GRCh38
rs747690434 Chr.15: 34355501-34355501 on Build GRCh38
rs944181631 Chr.15: 34355499-34355499 on Build GRCh38
rs888255982 Chr.15: 34355685-34355685 on Build GRCh38
rs775263884 Chr.15: 34355704-34355704 on Build GRCh38
rs757693918 Chr.15: 34355469-34355469 on Build GRCh38
rs761819633 Chr.15: 34355728-34355728 on Build GRCh38
rs769469309 Chr.15: 34355616-34355616 on Build GRCh38
rs772716400 Chr.15: 34355483-34355483 on Build GRCh38
rs372633609 Chr.15: 34355825-34355825 on Build GRCh38
rs369277860 Chr.15: 34355820-34355820 on Build GRCh38
rs145288462 Chr.15: 34355853-34355853 on Build GRCh38
rs763068881 Chr.15: 34355852-34355852 on Build GRCh38
rs968830702 Chr.15: 34355856-34355856 on Build GRCh38
rs759847785 Chr.15: 34355859-34355859 on Build GRCh38
rs751986279 Chr.15: 34355743-34355743 on Build GRCh38
rs113797264 Chr.15: 34355692-34355692 on Build GRCh38
rs758056107 Chr.15: 34355813-34355813 on Build GRCh38
rs764289245 Chr.15: 34355644-34355644 on Build GRCh38
rs916671060 Chr.15: 34355750-34355750 on Build GRCh38
rs895138270 Chr.15: 34355545-34355545 on Build GRCh38
rs571494372 Chr.15: 34355819-34355819 on Build GRCh38
rs187724721 Chr.15: 34355581-34355581 on Build GRCh38
rs560114865 Chr.15: 34355525-34355525 on Build GRCh38
rs988931364 Chr.15: 34355605-34355605 on Build GRCh38
rs549820703 Chr.15: 34355650-34355650 on Build GRCh38
rs776588383 Chr.15: 34355721-34355721 on Build GRCh38
rs779611382 Chr.15: 34355816-34355816 on Build GRCh38
rs778366625 Chr.15: 34355806-34355806 on Build GRCh38
rs774677346 Chr.15: 34355855-34355855 on Build GRCh38
rs529647220 Chr.15: 34355507-34355507 on Build GRCh38
rs773412361 Chr.15: 34355848-34355848 on Build GRCh38
rs150270874 Chr.15: 34355557-34355557 on Build GRCh38
rs34830783 Chr.15: 34355810-34355811 on Build GRCh38
rs78773193 Chr.15: 34355771-34355771 on Build GRCh38
rs775459257 Chr.15: 34355638-34355638 on Build GRCh38
rs1003207471 Chr.15: 34355798-34355798 on Build GRCh38
rs777144164 Chr.15: 34355617-34355617 on Build GRCh38
rs985716018 Chr.15: 34355430-34355430 on Build GRCh38
rs373788763 Chr.15: 34355572-34355572 on Build GRCh38
rs764062518 Chr.15: 34355576-34355576 on Build GRCh38
rs879231996 Chr.15: 34355648-34355648 on Build GRCh38
rs766908337 Chr.15: 34355739-34355739 on Build GRCh38
rs748753641 Chr.15: 34355688-34355688 on Build GRCh38
rs774003817 Chr.15: 34355537-34355537 on Build GRCh38
rs778006027 Chr.15: 34355452-34355452 on Build GRCh38
rs770204074 Chr.15: 34355473-34355473 on Build GRCh38
rs746745624 Chr.15: 34355715-34355715 on Build GRCh38
rs747495135 Chr.15: 34355612-34355612 on Build GRCh38
rs750088521 Chr.15: 34355604-34355604 on Build GRCh38
rs111885943 Chr.15: 34355618-34355618 on Build GRCh38
rs938575247 Chr.15: 34355817-34355817 on Build GRCh38
rs769443711 Chr.15: 34355675-34355709 on Build GRCh38
rs78073896 Chr.15: 34355611-34355611 on Build GRCh38
rs760769993 Chr.15: 34355571-34355571 on Build GRCh38
rs1026935508 Chr.15: 34355456-34355456 on Build GRCh38
rs74010082 Chr.15: 34355455-34355455 on Build GRCh38
rs1019668788 Chr.15: 34355437-34355437 on Build GRCh38
rs375124795 Chr.15: 34355474-34355474 on Build GRCh38
rs989495162 Chr.15: 34355727-34355727 on Build GRCh38
rs760578224 Chr.15: 34355640-34355640 on Build GRCh38
rs756769925 Chr.15: 34355795-34355795 on Build GRCh38
rs750622387 Chr.15: 34355652-34355652 on Build GRCh38
rs767942265 Chr.15: 34355759-34355759 on Build GRCh38
rs952592093 Chr.15: 34355496-34355496 on Build GRCh38
rs759270034 Chr.15: 34355556-34355556 on Build GRCh38
rs770505100 Chr.15: 34355622-34355622 on Build GRCh38
rs745501360 Chr.15: 34355698-34355698 on Build GRCh38
rs778560900 Chr.15: 34355695-34355695 on Build GRCh38
rs750818723 Chr.15: 34355601-34355601 on Build GRCh38
rs772965325 Chr.15: 34355512-34355512 on Build GRCh38
rs755426877 Chr.15: 34355686-34355686 on Build GRCh38
rs781682846 Chr.15: 34355687-34355687 on Build GRCh38
rs200891265 Chr.15: 34355500-34355500 on Build GRCh38
rs1046839152 Chr.15: 34355777-34355777 on Build GRCh38
rs757404618 Chr.15: 34355583-34355583 on Build GRCh38
rs747104257 Chr.15: 34355786-34355786 on Build GRCh38
rs779135171 Chr.15: 34355591-34355591 on Build GRCh38
rs114366417 Chr.15: 34355674-34355674 on Build GRCh38
rs370333183 Chr.15: 34355538-34355538 on Build GRCh38
rs756752810 Chr.15: 34355689-34355689 on Build GRCh38

View Full Product Details