Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34791024-34791522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTC1
Entrez Gene ID
70
Gene Name
actin, alpha, cardiac muscle 1
Gene Aliases
ACTC, ASD5, CMD1R, CMH11, LVNC4
UniGene
Hs.118127
Gene Chromosome Location
Chr.15: 34788096-34795726 on Build GRCh38
Transcripts
NM_005159.4 NR_120329.1

Gene Symbol
LOC101928174
Entrez Gene ID
101928174
Gene Name
uncharacterized LOC101928174
Gene Aliases
-
UniGene
Hs.680670
Gene Chromosome Location
Chr.15: 34755084-34812923 on Build GRCh38
Transcripts
NM_005159.4 NR_120329.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2268293 c.818C>T p.S273F Chr.15: 34791286-34791286 on Build GRCh38
6592871 c.879G>A p.K293K Chr.15: 34791225-34791225 on Build GRCh38
5944097 c.925_935del11 p.P309fs*1 Chr.15: 34791169-34791179 on Build GRCh38
5007317 c.941G>A p.R314H Chr.15: 34791163-34791163 on Build GRCh38
6592874 c.809-1G>A p.? Chr.15: 34791296-34791296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992571253 Chr.15: 34791344-34791348 on Build GRCh38
rs138347995 Chr.15: 34791201-34791201 on Build GRCh38
rs886038880 Chr.15: 34791134-34791134 on Build GRCh38
rs886074601 Chr.15: 34791454-34791454 on Build GRCh38
rs750091318 Chr.15: 34791322-34791323 on Build GRCh38
rs121912673 Chr.15: 34791163-34791163 on Build GRCh38
rs777680760 Chr.15: 34791104-34791104 on Build GRCh38
rs730880403 Chr.15: 34791152-34791152 on Build GRCh38
rs775774706 Chr.15: 34791340-34791354 on Build GRCh38
rs201456215 Chr.15: 34791308-34791323 on Build GRCh38
rs113062201 Chr.15: 34791336-34791353 on Build GRCh38
rs992270722 Chr.15: 34791458-34791458 on Build GRCh38
rs780884645 Chr.15: 34791092-34791092 on Build GRCh38
rs727504379 Chr.15: 34791238-34791238 on Build GRCh38
rs764887759 Chr.15: 34791310-34791325 on Build GRCh38
rs397517074 Chr.15: 34791186-34791186 on Build GRCh38
rs1049511214 Chr.15: 34791189-34791189 on Build GRCh38
rs766013863 Chr.15: 34791246-34791246 on Build GRCh38
rs981470702 Chr.15: 34791399-34791399 on Build GRCh38
rs149246511 Chr.15: 34791308-34791313 on Build GRCh38
rs140570640 Chr.15: 34791308-34791325 on Build GRCh38
rs144481225 Chr.15: 34791349-34791350 on Build GRCh38
rs757926447 Chr.15: 34791331-34791331 on Build GRCh38
rs759156523 Chr.15: 34791259-34791259 on Build GRCh38
rs778914066 Chr.15: 34791162-34791162 on Build GRCh38
rs998009318 Chr.15: 34791352-34791352 on Build GRCh38
rs751044644 Chr.15: 34791257-34791257 on Build GRCh38
rs765934670 Chr.15: 34791330-34791331 on Build GRCh38
rs886051089 Chr.15: 34791126-34791126 on Build GRCh38
rs764640888 Chr.15: 34791073-34791073 on Build GRCh38
rs756060664 Chr.15: 34791099-34791099 on Build GRCh38
rs368010998 Chr.15: 34791261-34791261 on Build GRCh38
rs866701109 Chr.15: 34791169-34791169 on Build GRCh38
rs543030688 Chr.15: 34791164-34791164 on Build GRCh38
rs561081869 Chr.15: 34791174-34791174 on Build GRCh38
rs754163265 Chr.15: 34791354-34791354 on Build GRCh38
rs532941484 Chr.15: 34791461-34791461 on Build GRCh38
rs987792186 Chr.15: 34791058-34791058 on Build GRCh38
rs545762703 Chr.15: 34791054-34791054 on Build GRCh38
rs1057518530 Chr.15: 34791196-34791196 on Build GRCh38
rs112469845 Chr.15: 34791204-34791432 on Build GRCh38
rs764090723 Chr.15: 34791307-34791307 on Build GRCh38
rs200369344 Chr.15: 34791078-34791078 on Build GRCh38
rs531880202 Chr.15: 34791496-34791496 on Build GRCh38
rs939520463 Chr.15: 34791129-34791129 on Build GRCh38
rs367842936 Chr.15: 34791335-34791336 on Build GRCh38
rs2307493 Chr.15: 34791177-34791177 on Build GRCh38
rs771011464 Chr.15: 34791137-34791137 on Build GRCh38
rs397517073 Chr.15: 34791254-34791254 on Build GRCh38
rs374685988 Chr.15: 34791337-34791338 on Build GRCh38
rs863225303 Chr.15: 34791210-34791210 on Build GRCh38
rs774238821 Chr.15: 34791306-34791307 on Build GRCh38
rs917890683 Chr.15: 34791352-34791353 on Build GRCh38
rs121912675 Chr.15: 34791215-34791215 on Build GRCh38
rs730880410 Chr.15: 34791118-34791118 on Build GRCh38
rs115415793 Chr.15: 34791362-34791362 on Build GRCh38
rs750991515 Chr.15: 34791333-34791351 on Build GRCh38
rs113565005 Chr.15: 34791297-34791297 on Build GRCh38
rs968331443 Chr.15: 34791391-34791391 on Build GRCh38
rs2307494 Chr.15: 34791050-34791050 on Build GRCh38
rs762550009 Chr.15: 34791081-34791081 on Build GRCh38
rs752499457 Chr.15: 34791304-34791304 on Build GRCh38
rs756470577 Chr.15: 34791348-34791348 on Build GRCh38
rs956424463 Chr.15: 34791372-34791372 on Build GRCh38
rs59431308 Chr.15: 34791353-34791354 on Build GRCh38
rs760450469 Chr.15: 34791342-34791343 on Build GRCh38
rs1033929114 Chr.15: 34791439-34791439 on Build GRCh38
rs867493541 Chr.15: 34791214-34791214 on Build GRCh38
rs1038835043 Chr.15: 34791406-34791406 on Build GRCh38
rs67669808 Chr.15: 34791338-34791353 on Build GRCh38
rs368741225 Chr.15: 34791086-34791086 on Build GRCh38
rs730880404 Chr.15: 34791136-34791136 on Build GRCh38
rs762354030 Chr.15: 34791231-34791231 on Build GRCh38
rs1022349632 Chr.15: 34791380-34791380 on Build GRCh38
rs781651067 Chr.15: 34791345-34791347 on Build GRCh38
rs1009253273 Chr.15: 34791374-34791374 on Build GRCh38
rs112623657 Chr.15: 34791113-34791113 on Build GRCh38
rs755521922 Chr.15: 34791343-34791343 on Build GRCh38
rs538052265 Chr.15: 34791283-34791283 on Build GRCh38
rs767415522 Chr.15: 34791340-34791341 on Build GRCh38
rs767357797 Chr.15: 34791307-34791308 on Build GRCh38
rs749154513 Chr.15: 34791108-34791108 on Build GRCh38
rs368549973 Chr.15: 34791387-34791387 on Build GRCh38
rs796944904 Chr.15: 34791348-34791349 on Build GRCh38
rs869025354 Chr.15: 34791250-34791250 on Build GRCh38
rs878999773 Chr.15: 34791308-34791309 on Build GRCh38
rs752512209 Chr.15: 34791098-34791098 on Build GRCh38
rs587780846 Chr.15: 34791307-34791307 on Build GRCh38
rs767180775 Chr.15: 34791303-34791303 on Build GRCh38
rs900297467 Chr.15: 34791420-34791420 on Build GRCh38
rs748575010 Chr.15: 34791346-34791348 on Build GRCh38
rs867699976 Chr.15: 34791224-34791224 on Build GRCh38
rs730880401 Chr.15: 34791292-34791292 on Build GRCh38
rs572654192 Chr.15: 34791308-34791311 on Build GRCh38
rs565588533 Chr.15: 34791448-34791448 on Build GRCh38
rs113513162 Chr.15: 34791274-34791274 on Build GRCh38
rs899704796 Chr.15: 34791349-34791349 on Build GRCh38
rs771953658 Chr.15: 34791387-34791387 on Build GRCh38
rs770947748 Chr.15: 34791338-34791352 on Build GRCh38
rs730880402 Chr.15: 34791218-34791218 on Build GRCh38
rs771899463 Chr.15: 34791338-34791355 on Build GRCh38
rs777285554 Chr.15: 34791204-34791204 on Build GRCh38
rs886051090 Chr.15: 34791308-34791319 on Build GRCh38
rs765049172 Chr.15: 34791309-34791311 on Build GRCh38
rs749257180 Chr.15: 34791336-34791352 on Build GRCh38
rs371877455 Chr.15: 34791088-34791088 on Build GRCh38
rs531195437 Chr.15: 34791085-34791085 on Build GRCh38

View Full Product Details