Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34873736-34874242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQR
Entrez Gene ID
9716
Gene Name
aquarius intron-binding spliceosomal factor
Gene Aliases
IBP160, fSAP164
UniGene
Hs.510958
Gene Chromosome Location
Chr.15: 34856351-34969794 on Build GRCh38
Transcripts
NM_014691.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1197100 c.3434A>G p.N1145S Chr.15: 34873991-34873991 on Build GRCh38
459052 c.3556C>T p.Q1186* Chr.15: 34873869-34873869 on Build GRCh38
4915861 c.3579T>C p.P1193P Chr.15: 34873846-34873846 on Build GRCh38
4974036 c.3583C>T p.P1195S Chr.15: 34873842-34873842 on Build GRCh38
4969999 c.3572C>T p.S1191F Chr.15: 34873853-34873853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772773793 Chr.15: 34873957-34873957 on Build GRCh38
rs552562479 Chr.15: 34874069-34874069 on Build GRCh38
rs567234646 Chr.15: 34874041-34874041 on Build GRCh38
rs747425434 Chr.15: 34873975-34873975 on Build GRCh38
rs981102436 Chr.15: 34874168-34874168 on Build GRCh38
rs913560557 Chr.15: 34874089-34874089 on Build GRCh38
rs777911104 Chr.15: 34874113-34874114 on Build GRCh38
rs770356308 Chr.15: 34874016-34874016 on Build GRCh38
rs776357687 Chr.15: 34873808-34873808 on Build GRCh38
rs961588946 Chr.15: 34874206-34874206 on Build GRCh38
rs769456293 Chr.15: 34873976-34873976 on Build GRCh38
rs201488950 Chr.15: 34873859-34873859 on Build GRCh38
rs988235021 Chr.15: 34874087-34874087 on Build GRCh38
rs777899236 Chr.15: 34873787-34873787 on Build GRCh38
rs577199973 Chr.15: 34874078-34874078 on Build GRCh38
rs1007024789 Chr.15: 34873768-34873768 on Build GRCh38
rs35971071 Chr.15: 34873793-34873794 on Build GRCh38
rs368882712 Chr.15: 34873846-34873846 on Build GRCh38
rs891151533 Chr.15: 34873953-34873953 on Build GRCh38
rs536852805 Chr.15: 34873911-34873911 on Build GRCh38
rs377705723 Chr.15: 34873956-34873956 on Build GRCh38
rs559279839 Chr.15: 34874023-34874023 on Build GRCh38
rs376967251 Chr.15: 34873825-34873825 on Build GRCh38
rs371314055 Chr.15: 34874010-34874010 on Build GRCh38
rs530024579 Chr.15: 34874139-34874139 on Build GRCh38
rs534562036 Chr.15: 34874065-34874065 on Build GRCh38
rs772705373 Chr.15: 34873805-34873805 on Build GRCh38
rs141632217 Chr.15: 34873764-34873764 on Build GRCh38
rs926170605 Chr.15: 34874146-34874146 on Build GRCh38
rs751470443 Chr.15: 34873850-34873850 on Build GRCh38
rs1040306323 Chr.15: 34873989-34873989 on Build GRCh38
rs201986214 Chr.15: 34873806-34873806 on Build GRCh38
rs200560139 Chr.15: 34873972-34873972 on Build GRCh38
rs971119169 Chr.15: 34874116-34874116 on Build GRCh38
rs892893241 Chr.15: 34874066-34874066 on Build GRCh38
rs569861650 Chr.15: 34873894-34873894 on Build GRCh38
rs777900641 Chr.15: 34873903-34873903 on Build GRCh38
rs942860572 Chr.15: 34874194-34874194 on Build GRCh38
rs1015858035 Chr.15: 34874199-34874199 on Build GRCh38
rs1012689397 Chr.15: 34874067-34874067 on Build GRCh38
rs1043332519 Chr.15: 34874129-34874130 on Build GRCh38
rs749379877 Chr.15: 34873792-34873792 on Build GRCh38
rs908788878 Chr.15: 34873815-34873815 on Build GRCh38
rs772770845 Chr.15: 34873993-34873993 on Build GRCh38
rs984045743 Chr.15: 34873866-34873866 on Build GRCh38
rs373250842 Chr.15: 34873914-34873914 on Build GRCh38
rs779418157 Chr.15: 34873921-34873921 on Build GRCh38
rs746529344 Chr.15: 34873897-34873897 on Build GRCh38
rs762410262 Chr.15: 34873832-34873832 on Build GRCh38
rs761552330 Chr.15: 34873816-34873816 on Build GRCh38
rs974254310 Chr.15: 34874208-34874208 on Build GRCh38
rs548864767 Chr.15: 34873926-34873926 on Build GRCh38
rs769389023 Chr.15: 34873821-34873821 on Build GRCh38
rs753917760 Chr.15: 34873905-34873905 on Build GRCh38
rs550781537 Chr.15: 34873887-34873887 on Build GRCh38

View Full Product Details