Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 36657713-36658227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf41
Entrez Gene ID
84529
Gene Name
chromosome 15 open reading frame 41
Gene Aliases
HH114
UniGene
Hs.48348
Gene Chromosome Location
Chr.15: 36579603-36810260 on Build GRCh38
Transcripts
NM_001130010.2 NM_001290232.1 NM_001290233.1 NM_001321756.1 NM_001321757.1 NM_001321758.1 NM_001321759.1 NM_001321760.1 NM_001321761.1 NM_032499.5 XM_011522111.1 XM_017022676.1 XM_017022677.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
960953 c.284C>T p.A95V Chr.15: 36657843-36657843 on Build GRCh38
5971371 c.290C>T p.S97L Chr.15: 36657849-36657849 on Build GRCh38
960952 c.274-1G>A p.? Chr.15: 36657832-36657832 on Build GRCh38
960954 c.301C>T p.R101W Chr.15: 36657860-36657860 on Build GRCh38
4645193 c.286C>T p.P96S Chr.15: 36657845-36657845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868279090 Chr.15: 36657944-36657944 on Build GRCh38
rs1054686869 Chr.15: 36657934-36657934 on Build GRCh38
rs879296321 Chr.15: 36657976-36657976 on Build GRCh38
rs980364023 Chr.15: 36658047-36658047 on Build GRCh38
rs79877354 Chr.15: 36658082-36658082 on Build GRCh38
rs552740790 Chr.15: 36658006-36658006 on Build GRCh38
rs370077292 Chr.15: 36658101-36658101 on Build GRCh38
rs747142710 Chr.15: 36657928-36657928 on Build GRCh38
rs76993439 Chr.15: 36657783-36657783 on Build GRCh38
rs372337437 Chr.15: 36657844-36657844 on Build GRCh38
rs548735216 Chr.15: 36657789-36657789 on Build GRCh38
rs770982288 Chr.15: 36657829-36657829 on Build GRCh38
rs145749465 Chr.15: 36657780-36657780 on Build GRCh38
rs1002587553 Chr.15: 36658174-36658174 on Build GRCh38
rs768850711 Chr.15: 36657942-36657942 on Build GRCh38
rs755539718 Chr.15: 36657800-36657800 on Build GRCh38
rs367989849 Chr.15: 36657859-36657859 on Build GRCh38
rs13329086 Chr.15: 36658160-36658160 on Build GRCh38
rs777429485 Chr.15: 36657806-36657806 on Build GRCh38
rs756863819 Chr.15: 36657884-36657884 on Build GRCh38
rs753397786 Chr.15: 36657883-36657883 on Build GRCh38
rs6495863 Chr.15: 36657751-36657751 on Build GRCh38
rs1020002844 Chr.15: 36658156-36658156 on Build GRCh38
rs756408340 Chr.15: 36657917-36657917 on Build GRCh38
rs190555032 Chr.15: 36658123-36658123 on Build GRCh38
rs765704243 Chr.15: 36657861-36657861 on Build GRCh38
rs201022722 Chr.15: 36657857-36657857 on Build GRCh38
rs758085699 Chr.15: 36657905-36657905 on Build GRCh38
rs755848656 Chr.15: 36657871-36657871 on Build GRCh38
rs749183771 Chr.15: 36657816-36657816 on Build GRCh38
rs958019408 Chr.15: 36657927-36657927 on Build GRCh38
rs527937388 Chr.15: 36657987-36657987 on Build GRCh38
rs373459296 Chr.15: 36658142-36658142 on Build GRCh38
rs745616406 Chr.15: 36657836-36657836 on Build GRCh38
rs1048221132 Chr.15: 36657835-36657835 on Build GRCh38
rs1030778452 Chr.15: 36657890-36657890 on Build GRCh38
rs148963775 Chr.15: 36658143-36658143 on Build GRCh38
rs766798444 Chr.15: 36657866-36657866 on Build GRCh38
rs752227872 Chr.15: 36657868-36657868 on Build GRCh38
rs562926148 Chr.15: 36657766-36657766 on Build GRCh38
rs760095205 Chr.15: 36657826-36657826 on Build GRCh38
rs745835771 Chr.15: 36657892-36657892 on Build GRCh38
rs969047534 Chr.15: 36658019-36658019 on Build GRCh38
rs773255408 Chr.15: 36657771-36657771 on Build GRCh38
rs587777101 Chr.15: 36657840-36657840 on Build GRCh38
rs763900403 Chr.15: 36657878-36657878 on Build GRCh38
rs919854988 Chr.15: 36657767-36657767 on Build GRCh38
rs556551397 Chr.15: 36658117-36658117 on Build GRCh38
rs776904462 Chr.15: 36657953-36657953 on Build GRCh38
rs775521817 Chr.15: 36657843-36657843 on Build GRCh38
rs777848881 Chr.15: 36657936-36657937 on Build GRCh38
rs767847503 Chr.15: 36658085-36658085 on Build GRCh38
rs931389662 Chr.15: 36658135-36658135 on Build GRCh38
rs867928419 Chr.15: 36657788-36657788 on Build GRCh38
rs776243356 Chr.15: 36658133-36658133 on Build GRCh38
rs770467860 Chr.15: 36657831-36657831 on Build GRCh38
rs553489250 Chr.15: 36658175-36658175 on Build GRCh38
rs779902933 Chr.15: 36657921-36657921 on Build GRCh38
rs201835784 Chr.15: 36657860-36657860 on Build GRCh38
rs566962772 Chr.15: 36657790-36657790 on Build GRCh38

View Full Product Details