Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38474043-38474562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98B
Entrez Gene ID
283742
Gene Name
family with sequence similarity 98 member B
Gene Aliases
-
UniGene
Hs.625289
Gene Chromosome Location
Chr.15: 38454127-38487710 on Build GRCh38
Transcripts
NM_173611.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1301057 c.710G>C p.G237A Chr.15: 38474279-38474279 on Build GRCh38
7299230 c.613-10A>G p.? Chr.15: 38474172-38474172 on Build GRCh38
1301058 c.710G>C p.G237A Chr.15: 38474279-38474279 on Build GRCh38
960989 c.620T>C p.L207P Chr.15: 38474189-38474189 on Build GRCh38
4441356 c.683G>A p.R228Q Chr.15: 38474252-38474252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051769641 Chr.15: 38474459-38474459 on Build GRCh38
rs539133168 Chr.15: 38474527-38474527 on Build GRCh38
rs780343072 Chr.15: 38474325-38474325 on Build GRCh38
rs747782955 Chr.15: 38474329-38474329 on Build GRCh38
rs907836354 Chr.15: 38474084-38474084 on Build GRCh38
rs986108023 Chr.15: 38474317-38474317 on Build GRCh38
rs62002940 Chr.15: 38474377-38474377 on Build GRCh38
rs753895894 Chr.15: 38474288-38474288 on Build GRCh38
rs181737697 Chr.15: 38474347-38474347 on Build GRCh38
rs1005864251 Chr.15: 38474503-38474503 on Build GRCh38
rs567645575 Chr.15: 38474318-38474318 on Build GRCh38
rs752884539 Chr.15: 38474434-38474434 on Build GRCh38
rs564151026 Chr.15: 38474085-38474085 on Build GRCh38
rs779209212 Chr.15: 38474159-38474159 on Build GRCh38
rs748825537 Chr.15: 38474334-38474334 on Build GRCh38
rs1057488319 Chr.15: 38474383-38474383 on Build GRCh38
rs961834619 Chr.15: 38474090-38474090 on Build GRCh38
rs759611968 Chr.15: 38474349-38474349 on Build GRCh38
rs746414936 Chr.15: 38474315-38474315 on Build GRCh38
rs767458011 Chr.15: 38474257-38474257 on Build GRCh38
rs535881213 Chr.15: 38474231-38474231 on Build GRCh38
rs772476138 Chr.15: 38474168-38474168 on Build GRCh38
rs772735059 Chr.15: 38474333-38474333 on Build GRCh38
rs961947042 Chr.15: 38474526-38474526 on Build GRCh38
rs760304570 Chr.15: 38474135-38474135 on Build GRCh38
rs776316579 Chr.15: 38474172-38474172 on Build GRCh38
rs887440709 Chr.15: 38474445-38474445 on Build GRCh38
rs1041976077 Chr.15: 38474332-38474332 on Build GRCh38
rs984386956 Chr.15: 38474356-38474356 on Build GRCh38
rs34337256 Chr.15: 38474433-38474434 on Build GRCh38
rs200539145 Chr.15: 38474263-38474263 on Build GRCh38
rs770612384 Chr.15: 38474343-38474343 on Build GRCh38
rs141118770 Chr.15: 38474408-38474410 on Build GRCh38
rs146385062 Chr.15: 38474218-38474218 on Build GRCh38
rs191315583 Chr.15: 38474226-38474226 on Build GRCh38
rs933332127 Chr.15: 38474441-38474441 on Build GRCh38
rs947410446 Chr.15: 38474324-38474324 on Build GRCh38
rs779372591 Chr.15: 38474312-38474312 on Build GRCh38
rs759380769 Chr.15: 38474251-38474251 on Build GRCh38
rs974769304 Chr.15: 38474395-38474395 on Build GRCh38
rs764175172 Chr.15: 38474281-38474281 on Build GRCh38
rs913341785 Chr.15: 38474467-38474467 on Build GRCh38
rs757712904 Chr.15: 38474300-38474300 on Build GRCh38
rs747825087 Chr.15: 38474185-38474185 on Build GRCh38
rs755691104 Chr.15: 38474307-38474307 on Build GRCh38
rs139746650 Chr.15: 38474234-38474234 on Build GRCh38
rs757774111 Chr.15: 38474158-38474158 on Build GRCh38
rs752958089 Chr.15: 38474265-38474265 on Build GRCh38
rs201530859 Chr.15: 38474269-38474269 on Build GRCh38
rs921898496 Chr.15: 38474411-38474411 on Build GRCh38
rs891931016 Chr.15: 38474134-38474134 on Build GRCh38
rs549262978 Chr.15: 38474266-38474266 on Build GRCh38
rs756349331 Chr.15: 38474270-38474270 on Build GRCh38
rs904800831 Chr.15: 38474280-38474280 on Build GRCh38
rs752323122 Chr.15: 38474148-38474154 on Build GRCh38
rs749789126 Chr.15: 38474157-38474157 on Build GRCh38
rs923349554 Chr.15: 38474149-38474149 on Build GRCh38
rs746253357 Chr.15: 38474164-38474164 on Build GRCh38
rs376806825 Chr.15: 38474215-38474215 on Build GRCh38
rs753326585 Chr.15: 38474392-38474392 on Build GRCh38
rs771885419 Chr.15: 38474350-38474350 on Build GRCh38
rs763555178 Chr.15: 38474409-38474409 on Build GRCh38
rs578000598 Chr.15: 38474481-38474481 on Build GRCh38
rs75101159 Chr.15: 38474079-38474080 on Build GRCh38
rs1001199981 Chr.15: 38474306-38474306 on Build GRCh38
rs1013977547 Chr.15: 38474342-38474342 on Build GRCh38
rs769335722 Chr.15: 38474214-38474214 on Build GRCh38
rs1014778624 Chr.15: 38474520-38474520 on Build GRCh38
rs770161410 Chr.15: 38474233-38474233 on Build GRCh38
rs959695627 Chr.15: 38474321-38474321 on Build GRCh38
rs140718010 Chr.15: 38474076-38474077 on Build GRCh38
rs777779570 Chr.15: 38474132-38474132 on Build GRCh38
rs896198049 Chr.15: 38474391-38474391 on Build GRCh38

View Full Product Details