Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38481181-38481723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98B
Entrez Gene ID
283742
Gene Name
family with sequence similarity 98 member B
Gene Aliases
-
UniGene
Hs.625289
Gene Chromosome Location
Chr.15: 38454127-38487710 on Build GRCh38
Transcripts
NM_173611.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3420259 c.795G>A p.T265T Chr.15: 38481357-38481357 on Build GRCh38
5519297 c.879C>T p.T293T Chr.15: 38481441-38481441 on Build GRCh38
135658 c.977C>T p.S326F Chr.15: 38481539-38481539 on Build GRCh38
2268677 c.868C>T p.R290W Chr.15: 38481430-38481430 on Build GRCh38
3420260 c.795G>A p.T265T Chr.15: 38481357-38481357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775161500 Chr.15: 38481241-38481241 on Build GRCh38
rs756841894 Chr.15: 38481213-38481213 on Build GRCh38
rs750510739 Chr.15: 38481415-38481415 on Build GRCh38
rs200367299 Chr.15: 38481571-38481571 on Build GRCh38
rs752154534 Chr.15: 38481431-38481431 on Build GRCh38
rs756929106 Chr.15: 38481581-38481581 on Build GRCh38
rs762915332 Chr.15: 38481558-38481558 on Build GRCh38
rs112596936 Chr.15: 38481442-38481442 on Build GRCh38
rs768324462 Chr.15: 38481513-38481513 on Build GRCh38
rs1014117588 Chr.15: 38481321-38481321 on Build GRCh38
rs774585657 Chr.15: 38481355-38481355 on Build GRCh38
rs987693057 Chr.15: 38481542-38481542 on Build GRCh38
rs139435266 Chr.15: 38481392-38481392 on Build GRCh38
rs557395856 Chr.15: 38481465-38481465 on Build GRCh38
rs755739118 Chr.15: 38481335-38481335 on Build GRCh38
rs185746275 Chr.15: 38481583-38481583 on Build GRCh38
rs778623343 Chr.15: 38481360-38481360 on Build GRCh38
rs762082697 Chr.15: 38481411-38481411 on Build GRCh38
rs766516491 Chr.15: 38481567-38481567 on Build GRCh38
rs779378459 Chr.15: 38481608-38481608 on Build GRCh38
rs766052033 Chr.15: 38481638-38481638 on Build GRCh38
rs763413844 Chr.15: 38481418-38481418 on Build GRCh38
rs746866742 Chr.15: 38481584-38481584 on Build GRCh38
rs146604215 Chr.15: 38481479-38481479 on Build GRCh38
rs960280944 Chr.15: 38481684-38481684 on Build GRCh38
rs542841170 Chr.15: 38481260-38481260 on Build GRCh38
rs760884102 Chr.15: 38481256-38481256 on Build GRCh38
rs539006816 Chr.15: 38481440-38481440 on Build GRCh38
rs763485245 Chr.15: 38481370-38481370 on Build GRCh38
rs1014500196 Chr.15: 38481650-38481650 on Build GRCh38
rs745740259 Chr.15: 38481383-38481383 on Build GRCh38
rs980965716 Chr.15: 38481444-38481444 on Build GRCh38
rs760145073 Chr.15: 38481573-38481573 on Build GRCh38
rs193044999 Chr.15: 38481402-38481402 on Build GRCh38
rs765245920 Chr.15: 38481414-38481414 on Build GRCh38
rs773038747 Chr.15: 38481562-38481562 on Build GRCh38
rs941717054 Chr.15: 38481467-38481467 on Build GRCh38
rs1046854941 Chr.15: 38481683-38481683 on Build GRCh38
rs1001254343 Chr.15: 38481222-38481222 on Build GRCh38
rs148441485 Chr.15: 38481530-38481530 on Build GRCh38
rs767603274 Chr.15: 38481551-38481551 on Build GRCh38
rs766813354 Chr.15: 38481423-38481423 on Build GRCh38
rs757667017 Chr.15: 38481585-38481586 on Build GRCh38
rs753368177 Chr.15: 38481687-38481687 on Build GRCh38
rs751904798 Chr.15: 38481329-38481329 on Build GRCh38
rs753114488 Chr.15: 38481580-38481580 on Build GRCh38
rs375025627 Chr.15: 38481582-38481582 on Build GRCh38
rs773231400 Chr.15: 38481546-38481546 on Build GRCh38
rs760446687 Chr.15: 38481590-38481590 on Build GRCh38
rs547607256 Chr.15: 38481577-38481577 on Build GRCh38
rs373769983 Chr.15: 38481283-38481283 on Build GRCh38
rs780403041 Chr.15: 38481317-38481317 on Build GRCh38
rs1016184412 Chr.15: 38481396-38481396 on Build GRCh38
rs780928732 Chr.15: 38481587-38481587 on Build GRCh38
rs1031858437 Chr.15: 38481641-38481641 on Build GRCh38
rs771662477 Chr.15: 38481472-38481472 on Build GRCh38
rs895601078 Chr.15: 38481617-38481617 on Build GRCh38
rs748305534 Chr.15: 38481533-38481533 on Build GRCh38
rs199750152 Chr.15: 38481293-38481293 on Build GRCh38
rs775630121 Chr.15: 38481277-38481277 on Build GRCh38
rs370314405 Chr.15: 38481635-38481635 on Build GRCh38
rs761851653 Chr.15: 38481270-38481270 on Build GRCh38
rs756982209 Chr.15: 38481446-38481446 on Build GRCh38
rs1032366853 Chr.15: 38481364-38481364 on Build GRCh38
rs376754001 Chr.15: 38481336-38481336 on Build GRCh38
rs200628550 Chr.15: 38481478-38481478 on Build GRCh38
rs770038154 Chr.15: 38481539-38481539 on Build GRCh38
rs758922386 Chr.15: 38481313-38481313 on Build GRCh38
rs374539540 Chr.15: 38481577-38481577 on Build GRCh38
rs771907405 Chr.15: 38481385-38481385 on Build GRCh38
rs373873719 Chr.15: 38481429-38481429 on Build GRCh38
rs768788629 Chr.15: 38481332-38481332 on Build GRCh38
rs754951308 Chr.15: 38481586-38481586 on Build GRCh38
rs374143929 Chr.15: 38481603-38481603 on Build GRCh38
rs375455088 Chr.15: 38481466-38481466 on Build GRCh38
rs777361855 Chr.15: 38481503-38481506 on Build GRCh38
rs775306766 Chr.15: 38481386-38481386 on Build GRCh38
rs150056145 Chr.15: 38481578-38481578 on Build GRCh38
rs947816822 Chr.15: 38481437-38481437 on Build GRCh38
rs776426294 Chr.15: 38481522-38481522 on Build GRCh38
rs917629930 Chr.15: 38481430-38481430 on Build GRCh38
rs761793572 Chr.15: 38481357-38481357 on Build GRCh38
rs747963954 Chr.15: 38481589-38481589 on Build GRCh38
rs377355881 Chr.15: 38481441-38481441 on Build GRCh38
rs1007938460 Chr.15: 38481285-38481285 on Build GRCh38

View Full Product Details