Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 39600776-39601292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSIP1
Entrez Gene ID
161835
Gene Name
fibrous sheath interacting protein 1
Gene Aliases
HSD10
UniGene
Hs.129598
Gene Chromosome Location
Chr.15: 39598643-39782838 on Build GRCh38
Transcripts
NM_001324338.1 NM_152597.4 XM_011521305.2 XM_011521306.1 XM_011521311.2 XM_017021972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4054252 c.1712C>G p.T571S Chr.15: 39600914-39600914 on Build GRCh38
961050 c.1710G>T p.E570D Chr.15: 39600916-39600916 on Build GRCh38
961049 c.1738G>T p.E580* Chr.15: 39600888-39600888 on Build GRCh38
3420263 c.1710G>A p.E570E Chr.15: 39600916-39600916 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1050323226 Chr.15: 39601181-39601181 on Build GRCh38
rs760293477 Chr.15: 39600919-39600919 on Build GRCh38
rs771671728 Chr.15: 39600904-39600904 on Build GRCh38
rs149871498 Chr.15: 39601104-39601107 on Build GRCh38
rs888107178 Chr.15: 39601149-39601149 on Build GRCh38
rs189770705 Chr.15: 39601116-39601116 on Build GRCh38
rs576991013 Chr.15: 39600818-39600818 on Build GRCh38
rs533234934 Chr.15: 39601118-39601118 on Build GRCh38
rs765073210 Chr.15: 39600852-39600852 on Build GRCh38
rs748676748 Chr.15: 39600884-39600884 on Build GRCh38
rs770272309 Chr.15: 39600889-39600889 on Build GRCh38
rs954113112 Chr.15: 39601031-39601031 on Build GRCh38
rs539679056 Chr.15: 39600828-39600828 on Build GRCh38
rs778967027 Chr.15: 39601170-39601170 on Build GRCh38
rs1028207751 Chr.15: 39601027-39601027 on Build GRCh38
rs907529956 Chr.15: 39601103-39601103 on Build GRCh38
rs1037284878 Chr.15: 39601117-39601117 on Build GRCh38
rs372181128 Chr.15: 39600974-39600974 on Build GRCh38
rs889978847 Chr.15: 39601183-39601183 on Build GRCh38
rs1036745927 Chr.15: 39601175-39601175 on Build GRCh38
rs1032705616 Chr.15: 39601038-39601038 on Build GRCh38
rs200066501 Chr.15: 39600890-39600890 on Build GRCh38
rs755059526 Chr.15: 39600881-39600881 on Build GRCh38
rs377133313 Chr.15: 39600882-39600882 on Build GRCh38
rs780448719 Chr.15: 39600872-39600872 on Build GRCh38
rs536785985 Chr.15: 39600897-39600897 on Build GRCh38
rs541711332 Chr.15: 39600995-39600995 on Build GRCh38
rs747049316 Chr.15: 39600877-39600877 on Build GRCh38
rs549986661 Chr.15: 39601248-39601248 on Build GRCh38
rs991019264 Chr.15: 39601064-39601064 on Build GRCh38
rs562121630 Chr.15: 39601068-39601068 on Build GRCh38
rs73391256 Chr.15: 39600845-39600845 on Build GRCh38
rs150431848 Chr.15: 39601094-39601094 on Build GRCh38
rs749673377 Chr.15: 39600901-39600901 on Build GRCh38
rs758632843 Chr.15: 39600860-39600860 on Build GRCh38
rs761739768 Chr.15: 39600955-39600955 on Build GRCh38
rs776067567 Chr.15: 39600947-39600947 on Build GRCh38
rs774929666 Chr.15: 39600839-39600839 on Build GRCh38
rs981843790 Chr.15: 39601095-39601095 on Build GRCh38
rs769985462 Chr.15: 39600888-39600888 on Build GRCh38
rs957892209 Chr.15: 39601055-39601055 on Build GRCh38
rs572386634 Chr.15: 39601075-39601075 on Build GRCh38
rs763641732 Chr.15: 39600928-39600928 on Build GRCh38
rs574248587 Chr.15: 39601230-39601230 on Build GRCh38
rs894725389 Chr.15: 39600846-39600846 on Build GRCh38
rs555948124 Chr.15: 39601129-39601129 on Build GRCh38
rs900931298 Chr.15: 39601262-39601262 on Build GRCh38
rs757195285 Chr.15: 39600848-39600848 on Build GRCh38
rs775091136 Chr.15: 39600908-39600908 on Build GRCh38
rs749084263 Chr.15: 39600969-39600969 on Build GRCh38
rs760261829 Chr.15: 39600842-39600842 on Build GRCh38
rs750244327 Chr.15: 39600854-39600854 on Build GRCh38
rs999428090 Chr.15: 39601026-39601026 on Build GRCh38
rs965207345 Chr.15: 39600971-39600971 on Build GRCh38
rs1003923080 Chr.15: 39601159-39601159 on Build GRCh38
rs938815669 Chr.15: 39601109-39601109 on Build GRCh38
rs371045004 Chr.15: 39600933-39600933 on Build GRCh38
rs376302906 Chr.15: 39600958-39600958 on Build GRCh38

View Full Product Details