Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 39739586-39740096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSIP1
Entrez Gene ID
161835
Gene Name
fibrous sheath interacting protein 1
Gene Aliases
HSD10
UniGene
Hs.129598
Gene Chromosome Location
Chr.15: 39598643-39782838 on Build GRCh38
Transcripts
NM_001324338.1 NM_152597.4 XM_011521305.2 XM_011521306.1 XM_011521307.2 XM_011521309.2 XM_011521311.2 XM_017021972.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5679493 c.656-1G>T p.? Chr.15: 39739790-39739790 on Build GRCh38
5608916 c.770G>A p.R257K Chr.15: 39739675-39739675 on Build GRCh38
5499557 c.705G>T p.K235N Chr.15: 39739740-39739740 on Build GRCh38
4930087 c.767A>G p.K256R Chr.15: 39739678-39739678 on Build GRCh38
108401 c.701G>A p.G234E Chr.15: 39739744-39739744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144656037 Chr.15: 39739752-39739752 on Build GRCh38
rs773834629 Chr.15: 39739683-39739683 on Build GRCh38
rs917086680 Chr.15: 39739933-39739933 on Build GRCh38
rs895595705 Chr.15: 39739629-39739629 on Build GRCh38
rs1039488316 Chr.15: 39739763-39739763 on Build GRCh38
rs886822032 Chr.15: 39740027-39740027 on Build GRCh38
rs761611252 Chr.15: 39739824-39739825 on Build GRCh38
rs781023155 Chr.15: 39739624-39739624 on Build GRCh38
rs377061239 Chr.15: 39739634-39739634 on Build GRCh38
rs753519612 Chr.15: 39740036-39740036 on Build GRCh38
rs371457746 Chr.15: 39739728-39739728 on Build GRCh38
rs112100398 Chr.15: 39739639-39739639 on Build GRCh38
rs771837678 Chr.15: 39739796-39739796 on Build GRCh38
rs778518233 Chr.15: 39739656-39739656 on Build GRCh38
rs573263701 Chr.15: 39739881-39739881 on Build GRCh38
rs553469228 Chr.15: 39739714-39739714 on Build GRCh38
rs1016472435 Chr.15: 39739717-39739717 on Build GRCh38
rs573375897 Chr.15: 39739716-39739716 on Build GRCh38
rs761984862 Chr.15: 39739839-39739839 on Build GRCh38
rs540567248 Chr.15: 39740020-39740020 on Build GRCh38
rs890757217 Chr.15: 39739729-39739729 on Build GRCh38
rs761727154 Chr.15: 39740052-39740052 on Build GRCh38
rs575932235 Chr.15: 39739801-39739801 on Build GRCh38
rs763380904 Chr.15: 39739719-39739719 on Build GRCh38
rs745702889 Chr.15: 39739661-39739661 on Build GRCh38
rs925893355 Chr.15: 39739891-39739891 on Build GRCh38
rs201576082 Chr.15: 39739692-39739692 on Build GRCh38
rs182112366 Chr.15: 39739915-39739915 on Build GRCh38
rs937226167 Chr.15: 39739893-39739893 on Build GRCh38
rs956636182 Chr.15: 39739860-39739860 on Build GRCh38
rs757099167 Chr.15: 39739773-39739773 on Build GRCh38
rs747845768 Chr.15: 39739633-39739633 on Build GRCh38
rs536498351 Chr.15: 39739791-39739791 on Build GRCh38
rs143015687 Chr.15: 39739859-39739859 on Build GRCh38
rs949886483 Chr.15: 39739940-39739940 on Build GRCh38
rs148586423 Chr.15: 39739769-39739769 on Build GRCh38
rs762169076 Chr.15: 39739711-39739711 on Build GRCh38
rs1005668745 Chr.15: 39740028-39740028 on Build GRCh38
rs188638136 Chr.15: 39739816-39739816 on Build GRCh38
rs377291889 Chr.15: 39739775-39739775 on Build GRCh38
rs186651659 Chr.15: 39740024-39740024 on Build GRCh38
rs770910359 Chr.15: 39739644-39739644 on Build GRCh38
rs769805169 Chr.15: 39739841-39739841 on Build GRCh38
rs930215920 Chr.15: 39740068-39740068 on Build GRCh38
rs775663967 Chr.15: 39739670-39739670 on Build GRCh38
rs371733198 Chr.15: 39739777-39739777 on Build GRCh38
rs760944022 Chr.15: 39739687-39739687 on Build GRCh38
rs145528419 Chr.15: 39739744-39739744 on Build GRCh38
rs540996260 Chr.15: 39739812-39739812 on Build GRCh38
rs771892500 Chr.15: 39739663-39739663 on Build GRCh38
rs749180635 Chr.15: 39739641-39739641 on Build GRCh38
rs779889814 Chr.15: 39739798-39739798 on Build GRCh38
rs763814154 Chr.15: 39740025-39740025 on Build GRCh38
rs776776764 Chr.15: 39739701-39739701 on Build GRCh38
rs367899903 Chr.15: 39740017-39740017 on Build GRCh38
rs1046786857 Chr.15: 39739662-39739662 on Build GRCh38
rs143016905 Chr.15: 39739743-39739743 on Build GRCh38
rs775161841 Chr.15: 39739848-39739848 on Build GRCh38
rs768715909 Chr.15: 39739689-39739689 on Build GRCh38

View Full Product Details