Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40169927-40170429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1B
Entrez Gene ID
701
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase B
Gene Aliases
BUB1beta, BUBR1, Bub1A, MAD3L, MVA1, SSK1, hBUBR1
UniGene
Hs.513645
Gene Chromosome Location
Chr.15: 40161009-40221136 on Build GRCh38
Transcripts
NM_001211.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1730528 c.190T>A p.Y64N Chr.15: 40170072-40170072 on Build GRCh38
6077243 c.238A>T p.R80W Chr.15: 40170120-40170120 on Build GRCh38
4666426 c.186T>C p.F62F Chr.15: 40170068-40170068 on Build GRCh38
7140749 c.200G>T p.R67L Chr.15: 40170082-40170082 on Build GRCh38
6077242 c.238A>T p.R80W Chr.15: 40170120-40170120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989150125 Chr.15: 40169959-40169959 on Build GRCh38
rs372984239 Chr.15: 40170200-40170200 on Build GRCh38
rs770065443 Chr.15: 40170018-40170018 on Build GRCh38
rs869301849 Chr.15: 40170183-40170183 on Build GRCh38
rs184525311 Chr.15: 40170388-40170388 on Build GRCh38
rs989899832 Chr.15: 40169992-40169992 on Build GRCh38
rs1015878697 Chr.15: 40170247-40170247 on Build GRCh38
rs193241496 Chr.15: 40170306-40170306 on Build GRCh38
rs748354192 Chr.15: 40170016-40170016 on Build GRCh38
rs945912773 Chr.15: 40170024-40170024 on Build GRCh38
rs370352952 Chr.15: 40170148-40170148 on Build GRCh38
rs945047943 Chr.15: 40170328-40170328 on Build GRCh38
rs1012913493 Chr.15: 40170155-40170155 on Build GRCh38
rs781034323 Chr.15: 40170082-40170082 on Build GRCh38
rs547531900 Chr.15: 40170287-40170287 on Build GRCh38
rs767911076 Chr.15: 40170034-40170034 on Build GRCh38
rs967119927 Chr.15: 40170254-40170254 on Build GRCh38
rs910447433 Chr.15: 40170349-40170349 on Build GRCh38
rs986088552 Chr.15: 40170317-40170317 on Build GRCh38
rs1037722616 Chr.15: 40170358-40170358 on Build GRCh38
rs1054967966 Chr.15: 40170345-40170345 on Build GRCh38
rs747976070 Chr.15: 40170089-40170089 on Build GRCh38
rs750033644 Chr.15: 40170164-40170164 on Build GRCh38
rs775977964 Chr.15: 40170142-40170142 on Build GRCh38
rs200483866 Chr.15: 40170059-40170059 on Build GRCh38
rs377039578 Chr.15: 40170167-40170167 on Build GRCh38
rs16970425 Chr.15: 40170027-40170027 on Build GRCh38
rs989762306 Chr.15: 40170240-40170240 on Build GRCh38
rs1037816374 Chr.15: 40170131-40170131 on Build GRCh38
rs200884355 Chr.15: 40170081-40170081 on Build GRCh38
rs958609879 Chr.15: 40169966-40169966 on Build GRCh38
rs769980774 Chr.15: 40170091-40170091 on Build GRCh38
rs941895890 Chr.15: 40170357-40170357 on Build GRCh38
rs759974563 Chr.15: 40170029-40170029 on Build GRCh38
rs751094435 Chr.15: 40170179-40170179 on Build GRCh38
rs897922423 Chr.15: 40170144-40170144 on Build GRCh38
rs1034028990 Chr.15: 40169951-40169951 on Build GRCh38
rs760085736 Chr.15: 40170116-40170116 on Build GRCh38
rs746902649 Chr.15: 40170069-40170069 on Build GRCh38
rs760199559 Chr.15: 40170363-40170363 on Build GRCh38
rs138390503 Chr.15: 40170239-40170239 on Build GRCh38
rs906662010 Chr.15: 40170196-40170196 on Build GRCh38
rs143888330 Chr.15: 40170280-40170280 on Build GRCh38
rs779439053 Chr.15: 40170066-40170066 on Build GRCh38
rs953784144 Chr.15: 40170256-40170256 on Build GRCh38
rs535738783 Chr.15: 40170383-40170383 on Build GRCh38
rs754911446 Chr.15: 40170180-40170180 on Build GRCh38
rs753071863 Chr.15: 40170039-40170039 on Build GRCh38
rs770986034 Chr.15: 40170107-40170107 on Build GRCh38
rs1048487615 Chr.15: 40170384-40170384 on Build GRCh38
rs945289006 Chr.15: 40170030-40170030 on Build GRCh38
rs548540749 Chr.15: 40170101-40170101 on Build GRCh38
rs750125592 Chr.15: 40170055-40170055 on Build GRCh38
rs539421592 Chr.15: 40170332-40170332 on Build GRCh38
rs977102345 Chr.15: 40170265-40170265 on Build GRCh38
rs765895703 Chr.15: 40170173-40170173 on Build GRCh38
rs372718695 Chr.15: 40170051-40170051 on Build GRCh38
rs772497259 Chr.15: 40170120-40170120 on Build GRCh38
rs906743871 Chr.15: 40170405-40170405 on Build GRCh38
rs545833887 Chr.15: 40170021-40170021 on Build GRCh38
rs756941170 Chr.15: 40170042-40170042 on Build GRCh38
rs556117748 Chr.15: 40170342-40170342 on Build GRCh38
rs978147562 Chr.15: 40170330-40170330 on Build GRCh38
rs1021217396 Chr.15: 40170250-40170250 on Build GRCh38
rs569750437 Chr.15: 40170366-40170366 on Build GRCh38
rs966008137 Chr.15: 40170249-40170249 on Build GRCh38
rs777922307 Chr.15: 40170094-40170094 on Build GRCh38
rs869100891 Chr.15: 40170084-40170085 on Build GRCh38
rs773847890 Chr.15: 40170203-40170203 on Build GRCh38
rs113437650 Chr.15: 40170125-40170125 on Build GRCh38
rs774353130 Chr.15: 40170111-40170111 on Build GRCh38
rs773243532 Chr.15: 40170019-40170019 on Build GRCh38
rs781038212 Chr.15: 40170197-40170197 on Build GRCh38

View Full Product Details