Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40331975-40332484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf52
Entrez Gene ID
388115
Gene Name
chromosome 15 open reading frame 52
Gene Aliases
-
UniGene
Hs.32433
Gene Chromosome Location
Chr.15: 40331452-40340967 on Build GRCh38
Transcripts
NM_207380.2

SNP Detail

SNP ID Chromosome Location
rs1044130323 Chr.15: 40332181-40332181 on Build GRCh38
rs905337062 Chr.15: 40332190-40332190 on Build GRCh38
rs925860338 Chr.15: 40332082-40332082 on Build GRCh38
rs1026293142 Chr.15: 40332251-40332251 on Build GRCh38
rs768338719 Chr.15: 40332011-40332011 on Build GRCh38
rs77194071 Chr.15: 40332293-40332293 on Build GRCh38
rs767735586 Chr.15: 40332255-40332255 on Build GRCh38
rs895499516 Chr.15: 40332158-40332158 on Build GRCh38
rs892819810 Chr.15: 40332226-40332226 on Build GRCh38
rs766283724 Chr.15: 40332073-40332073 on Build GRCh38
rs116336794 Chr.15: 40332014-40332014 on Build GRCh38
rs978651376 Chr.15: 40332045-40332046 on Build GRCh38
rs953612385 Chr.15: 40332319-40332319 on Build GRCh38
rs145324448 Chr.15: 40332235-40332235 on Build GRCh38
rs778236501 Chr.15: 40332322-40332322 on Build GRCh38
rs972273140 Chr.15: 40332392-40332392 on Build GRCh38
rs1017693014 Chr.15: 40332302-40332302 on Build GRCh38
rs530036698 Chr.15: 40332423-40332423 on Build GRCh38
rs1016244492 Chr.15: 40332331-40332331 on Build GRCh38
rs561075992 Chr.15: 40332317-40332317 on Build GRCh38
rs909446679 Chr.15: 40332027-40332027 on Build GRCh38
rs987571553 Chr.15: 40332025-40332025 on Build GRCh38
rs1010079823 Chr.15: 40332236-40332236 on Build GRCh38
rs925838881 Chr.15: 40332339-40332339 on Build GRCh38
rs544365843 Chr.15: 40332315-40332315 on Build GRCh38
rs940899359 Chr.15: 40332035-40332035 on Build GRCh38
rs1045484983 Chr.15: 40332144-40332144 on Build GRCh38
rs144179803 Chr.15: 40332311-40332312 on Build GRCh38
rs997797633 Chr.15: 40332312-40332312 on Build GRCh38
rs970913462 Chr.15: 40332461-40332461 on Build GRCh38
rs751698994 Chr.15: 40332157-40332157 on Build GRCh38
rs962049708 Chr.15: 40332375-40332375 on Build GRCh38
rs755048236 Chr.15: 40332230-40332230 on Build GRCh38
rs773455201 Chr.15: 40332096-40332097 on Build GRCh38
rs964828571 Chr.15: 40332304-40332305 on Build GRCh38
rs1016453985 Chr.15: 40332209-40332209 on Build GRCh38
rs7165582 Chr.15: 40332040-40332040 on Build GRCh38
rs573134542 Chr.15: 40332106-40332106 on Build GRCh38
rs1053186301 Chr.15: 40332150-40332150 on Build GRCh38
rs1054070883 Chr.15: 40332207-40332207 on Build GRCh38
rs993775105 Chr.15: 40332234-40332234 on Build GRCh38
rs139987593 Chr.15: 40332173-40332173 on Build GRCh38
rs774156261 Chr.15: 40332036-40332036 on Build GRCh38
rs756462143 Chr.15: 40332304-40332304 on Build GRCh38
rs150050325 Chr.15: 40332095-40332096 on Build GRCh38
rs905641879 Chr.15: 40332168-40332168 on Build GRCh38
rs534214550 Chr.15: 40332041-40332041 on Build GRCh38

View Full Product Details